Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37733338-37733868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
UniGene
Hs.163990
Gene Chromosome Location
Chr.9: 37602979-37746904 on Build GRCh38
Transcripts
NM_014907.2 XM_011517803.2 XM_011517804.2 XM_011517805.2 XM_011517806.2 XM_017014480.1 XM_017014481.1 XM_017014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4374353 c.613G>A p.G205R Chr.9: 37733483-37733483 on Build GRCh38
6115904 c.1021A>G p.I341V Chr.9: 37733498-37733498 on Build GRCh38
4374355 c.672G>T p.K224N Chr.9: 37733542-37733542 on Build GRCh38
6667048 c.1083C>T p.H361H Chr.9: 37733560-37733560 on Build GRCh38
487463 c.1008A>G p.G336G Chr.9: 37733485-37733485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138526022 Chr.9: 37733778-37733778 on Build GRCh38
rs775846532 Chr.9: 37733426-37733426 on Build GRCh38
rs971677151 Chr.9: 37733666-37733666 on Build GRCh38
rs772024773 Chr.9: 37733812-37733812 on Build GRCh38
rs768873921 Chr.9: 37733536-37733536 on Build GRCh38
rs780352243 Chr.9: 37733787-37733787 on Build GRCh38
rs369514409 Chr.9: 37733768-37733768 on Build GRCh38
rs979040535 Chr.9: 37733781-37733781 on Build GRCh38
rs372911565 Chr.9: 37733619-37733619 on Build GRCh38
rs762549033 Chr.9: 37733611-37733611 on Build GRCh38
rs769128597 Chr.9: 37733790-37733790 on Build GRCh38
rs914540861 Chr.9: 37733732-37733732 on Build GRCh38
rs761504225 Chr.9: 37733448-37733448 on Build GRCh38
rs753949850 Chr.9: 37733772-37733772 on Build GRCh38
rs764862793 Chr.9: 37733454-37733454 on Build GRCh38
rs148144624 Chr.9: 37733517-37733517 on Build GRCh38
rs199872242 Chr.9: 37733794-37733794 on Build GRCh38
rs111749017 Chr.9: 37733830-37733831 on Build GRCh38
rs777075450 Chr.9: 37733480-37733481 on Build GRCh38
rs963157872 Chr.9: 37733672-37733672 on Build GRCh38
rs774068296 Chr.9: 37733795-37733795 on Build GRCh38
rs770585299 Chr.9: 37733724-37733724 on Build GRCh38
rs775964178 Chr.9: 37733711-37733711 on Build GRCh38
rs756473213 Chr.9: 37733516-37733516 on Build GRCh38
rs776047208 Chr.9: 37733591-37733591 on Build GRCh38
rs747532862 Chr.9: 37733788-37733788 on Build GRCh38
rs751482715 Chr.9: 37733478-37733478 on Build GRCh38
rs748206120 Chr.9: 37733500-37733500 on Build GRCh38
rs776161036 Chr.9: 37733446-37733446 on Build GRCh38
rs766108689 Chr.9: 37733615-37733615 on Build GRCh38
rs766146826 Chr.9: 37733708-37733708 on Build GRCh38
rs1043110713 Chr.9: 37733368-37733368 on Build GRCh38
rs760022479 Chr.9: 37733433-37733433 on Build GRCh38
rs566568909 Chr.9: 37733360-37733360 on Build GRCh38
rs200774800 Chr.9: 37733626-37733626 on Build GRCh38
rs141088968 Chr.9: 37733596-37733596 on Build GRCh38
rs772852140 Chr.9: 37733599-37733599 on Build GRCh38
rs1016533181 Chr.9: 37733574-37733574 on Build GRCh38
rs1034446265 Chr.9: 37733688-37733688 on Build GRCh38
rs769322565 Chr.9: 37733710-37733710 on Build GRCh38
rs933498908 Chr.9: 37733468-37733468 on Build GRCh38
rs764295228 Chr.9: 37733771-37733771 on Build GRCh38
rs372000128 Chr.9: 37733707-37733707 on Build GRCh38
rs749464327 Chr.9: 37733523-37733523 on Build GRCh38
rs532373146 Chr.9: 37733465-37733465 on Build GRCh38
rs368540362 Chr.9: 37733549-37733549 on Build GRCh38
rs901419685 Chr.9: 37733609-37733609 on Build GRCh38
rs185914709 Chr.9: 37733635-37733635 on Build GRCh38
rs933516297 Chr.9: 37733829-37733829 on Build GRCh38
rs140800271 Chr.9: 37733755-37733755 on Build GRCh38
rs781177368 Chr.9: 37733491-37733491 on Build GRCh38
rs1041895719 Chr.9: 37733382-37733382 on Build GRCh38
rs199563469 Chr.9: 37733793-37733793 on Build GRCh38
rs377074505 Chr.9: 37733636-37733636 on Build GRCh38
rs369673683 Chr.9: 37733705-37733705 on Build GRCh38
rs759854327 Chr.9: 37733633-37733635 on Build GRCh38
rs967196038 Chr.9: 37733775-37733775 on Build GRCh38
rs759046042 Chr.9: 37733804-37733804 on Build GRCh38
rs1006136871 Chr.9: 37733660-37733660 on Build GRCh38
rs777085801 Chr.9: 37733620-37733620 on Build GRCh38
rs78896005 Chr.9: 37733839-37733839 on Build GRCh38
rs766127373 Chr.9: 37733474-37733474 on Build GRCh38
rs771204432 Chr.9: 37733545-37733545 on Build GRCh38
rs202015898 Chr.9: 37733710-37733711 on Build GRCh38
rs763547855 Chr.9: 37733444-37733444 on Build GRCh38
rs772612722 Chr.9: 37733579-37733579 on Build GRCh38
rs374356516 Chr.9: 37733505-37733505 on Build GRCh38
rs772660042 Chr.9: 37733712-37733712 on Build GRCh38
rs756277147 Chr.9: 37733622-37733622 on Build GRCh38
rs753143578 Chr.9: 37733709-37733709 on Build GRCh38
rs767160958 Chr.9: 37733754-37733754 on Build GRCh38
rs545280550 Chr.9: 37733592-37733592 on Build GRCh38
rs762645776 Chr.9: 37733683-37733683 on Build GRCh38
rs750274043 Chr.9: 37733457-37733457 on Build GRCh38
rs774079326 Chr.9: 37733727-37733727 on Build GRCh38
rs967395709 Chr.9: 37733737-37733737 on Build GRCh38
rs779197357 Chr.9: 37733640-37733640 on Build GRCh38
rs902041082 Chr.9: 37733422-37733422 on Build GRCh38
rs759363152 Chr.9: 37733745-37733745 on Build GRCh38
rs538015666 Chr.9: 37733647-37733647 on Build GRCh38
rs887253855 Chr.9: 37733657-37733657 on Build GRCh38
rs746388440 Chr.9: 37733569-37733569 on Build GRCh38
rs767115433 Chr.9: 37733777-37733777 on Build GRCh38
rs754895066 Chr.9: 37733487-37733487 on Build GRCh38
rs376747897 Chr.9: 37733720-37733720 on Build GRCh38

View Full Product Details