Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 134789057-134789433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM122B
Entrez Gene ID
159090
Gene Name
family with sequence similarity 122B
Gene Aliases
SPACIA2
UniGene
Hs.404706
Gene Chromosome Location
Chr.X: 134769566-134797232 on Build GRCh38
Transcripts
NM_001166599.2 NM_001166600.2 NM_001170756.1 NM_001170757.1 NM_145284.5 XM_006724730.2 XM_006724732.2 XM_006724733.3 XM_011531282.1 XM_011531283.1 XM_011531284.1 XM_011531285.1 XM_011531286.1 XM_011531287.1 XM_011531288.1 XM_011531289.1 XM_011531290.1 XM_011531291.1 XM_011531292.1 XM_011531293.1 XM_011531294.1 XM_017029306.1 XM_017029307.1

SNP Detail

SNP ID Chromosome Location
rs764300213 Chr.X: 134789146-134789146 on Build GRCh38
rs1011991708 Chr.X: 134789320-134789320 on Build GRCh38
rs767776388 Chr.X: 134789394-134789394 on Build GRCh38
rs770751499 Chr.X: 134789209-134789209 on Build GRCh38
rs557056064 Chr.X: 134789308-134789308 on Build GRCh38
rs1021679024 Chr.X: 134789334-134789334 on Build GRCh38
rs763443114 Chr.X: 134789135-134789135 on Build GRCh38
rs377296666 Chr.X: 134789328-134789328 on Build GRCh38
rs778964213 Chr.X: 134789187-134789187 on Build GRCh38
rs745441825 Chr.X: 134789230-134789230 on Build GRCh38
rs200988823 Chr.X: 134789190-134789190 on Build GRCh38
rs375316140 Chr.X: 134789284-134789284 on Build GRCh38
rs760667614 Chr.X: 134789329-134789329 on Build GRCh38
rs757420254 Chr.X: 134789160-134789160 on Build GRCh38
rs759089577 Chr.X: 134789126-134789126 on Build GRCh38
rs757069116 Chr.X: 134789195-134789196 on Build GRCh38
rs774657485 Chr.X: 134789312-134789312 on Build GRCh38
rs150146844 Chr.X: 134789358-134789358 on Build GRCh38
rs891839913 Chr.X: 134789290-134789290 on Build GRCh38
rs1054267388 Chr.X: 134789332-134789332 on Build GRCh38
rs773487698 Chr.X: 134789316-134789316 on Build GRCh38
rs769184633 Chr.X: 134789269-134789269 on Build GRCh38
rs138947407 Chr.X: 134789106-134789106 on Build GRCh38
rs914494087 Chr.X: 134789367-134789367 on Build GRCh38
rs756120865 Chr.X: 134789169-134789169 on Build GRCh38
rs777665722 Chr.X: 134789204-134789204 on Build GRCh38
rs776071037 Chr.X: 134789282-134789282 on Build GRCh38
rs1002818581 Chr.X: 134789184-134789184 on Build GRCh38
rs746591286 Chr.X: 134789095-134789095 on Build GRCh38
rs751795110 Chr.X: 134789150-134789150 on Build GRCh38
rs965169742 Chr.X: 134789335-134789335 on Build GRCh38
rs547436971 Chr.X: 134789390-134789390 on Build GRCh38
rs372860931 Chr.X: 134789205-134789205 on Build GRCh38
rs17317849 Chr.X: 134789109-134789109 on Build GRCh38
rs776587668 Chr.X: 134789373-134789374 on Build GRCh38
rs758215206 Chr.X: 134789203-134789203 on Build GRCh38
rs767614958 Chr.X: 134789164-134789164 on Build GRCh38
rs775848587 Chr.X: 134789128-134789128 on Build GRCh38
rs750541645 Chr.X: 134789167-134789167 on Build GRCh38
rs765826136 Chr.X: 134789127-134789127 on Build GRCh38
rs776404895 Chr.X: 134789212-134789212 on Build GRCh38

View Full Product Details