Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 78596968-78597554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA3
Entrez Gene ID
306
Gene Name
annexin A3
Gene Aliases
ANX3
UniGene
Hs.480042
Gene Chromosome Location
Chr.4: 78551588-78610451 on Build GRCh38
Transcripts
NM_005139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1431246 c.703C>T p.H235Y Chr.4: 78597387-78597387 on Build GRCh38
393684 c.635-1G>T p.? Chr.4: 78597318-78597318 on Build GRCh38
481560 c.718C>A p.L240M Chr.4: 78597402-78597402 on Build GRCh38
6261240 c.724G>T p.A242S Chr.4: 78597408-78597408 on Build GRCh38
3606296 c.657C>T p.I219I Chr.4: 78597341-78597341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201370863 Chr.4: 78597500-78597500 on Build GRCh38
rs992020777 Chr.4: 78597347-78597347 on Build GRCh38
rs544019292 Chr.4: 78597472-78597472 on Build GRCh38
rs947937988 Chr.4: 78597008-78597008 on Build GRCh38
rs995697655 Chr.4: 78597102-78597102 on Build GRCh38
rs6811432 Chr.4: 78597140-78597140 on Build GRCh38
rs5948 Chr.4: 78597340-78597340 on Build GRCh38
rs950463745 Chr.4: 78597218-78597218 on Build GRCh38
rs181593447 Chr.4: 78597321-78597321 on Build GRCh38
rs948163950 Chr.4: 78597471-78597471 on Build GRCh38
rs773594353 Chr.4: 78597401-78597401 on Build GRCh38
rs544680965 Chr.4: 78597030-78597030 on Build GRCh38
rs200781276 Chr.4: 78597276-78597276 on Build GRCh38
rs796739014 Chr.4: 78597495-78597495 on Build GRCh38
rs752297719 Chr.4: 78597308-78597308 on Build GRCh38
rs780472674 Chr.4: 78597342-78597342 on Build GRCh38
rs1013136303 Chr.4: 78597031-78597031 on Build GRCh38
rs1056975332 Chr.4: 78597019-78597019 on Build GRCh38
rs930959479 Chr.4: 78597475-78597475 on Build GRCh38
rs746181482 Chr.4: 78597329-78597329 on Build GRCh38
rs569205950 Chr.4: 78597242-78597242 on Build GRCh38
rs994743944 Chr.4: 78597511-78597511 on Build GRCh38
rs748302781 Chr.4: 78597388-78597388 on Build GRCh38
rs73828043 Chr.4: 78597284-78597284 on Build GRCh38
rs772148534 Chr.4: 78597397-78597397 on Build GRCh38
rs1019083860 Chr.4: 78597264-78597264 on Build GRCh38
rs1026257042 Chr.4: 78597524-78597524 on Build GRCh38
rs749307848 Chr.4: 78597273-78597273 on Build GRCh38
rs561037114 Chr.4: 78597512-78597512 on Build GRCh38
rs760493179 Chr.4: 78597403-78597403 on Build GRCh38
rs547366308 Chr.4: 78597050-78597050 on Build GRCh38
rs1019169610 Chr.4: 78597188-78597188 on Build GRCh38
rs764635603 Chr.4: 78597305-78597305 on Build GRCh38
rs560067429 Chr.4: 78597125-78597125 on Build GRCh38
rs759521730 Chr.4: 78597453-78597453 on Build GRCh38
rs935173037 Chr.4: 78597344-78597344 on Build GRCh38
rs1056482210 Chr.4: 78597413-78597413 on Build GRCh38
rs753844970 Chr.4: 78597297-78597297 on Build GRCh38
rs370924287 Chr.4: 78597277-78597277 on Build GRCh38
rs886836947 Chr.4: 78597526-78597526 on Build GRCh38
rs766373060 Chr.4: 78597295-78597295 on Build GRCh38
rs532484518 Chr.4: 78597246-78597246 on Build GRCh38
rs886610093 Chr.4: 78597156-78597156 on Build GRCh38
rs1047264240 Chr.4: 78597487-78597487 on Build GRCh38
rs76109076 Chr.4: 78597283-78597283 on Build GRCh38
rs112391761 Chr.4: 78597350-78597350 on Build GRCh38
rs538516109 Chr.4: 78597331-78597331 on Build GRCh38
rs537983688 Chr.4: 78597275-78597275 on Build GRCh38
rs375891610 Chr.4: 78597456-78597456 on Build GRCh38
rs117293676 Chr.4: 78597425-78597425 on Build GRCh38
rs5947 Chr.4: 78597320-78597320 on Build GRCh38
rs758063772 Chr.4: 78597345-78597345 on Build GRCh38
rs372028565 Chr.4: 78597208-78597208 on Build GRCh38
rs749791814 Chr.4: 78597368-78597368 on Build GRCh38
rs6854623 Chr.4: 78597058-78597058 on Build GRCh38
rs776623242 Chr.4: 78597442-78597442 on Build GRCh38
rs769265929 Chr.4: 78597385-78597385 on Build GRCh38
rs373769010 Chr.4: 78597287-78597287 on Build GRCh38
rs371527894 Chr.4: 78597178-78597178 on Build GRCh38
rs778839003 Chr.4: 78597386-78597386 on Build GRCh38
rs552621277 Chr.4: 78597240-78597240 on Build GRCh38
rs755028590 Chr.4: 78597302-78597302 on Build GRCh38

View Full Product Details