Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74725738-74726315 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFM2
Entrez Gene ID
84340
Gene Name
G elongation factor mitochondrial 2
Gene Aliases
EF-G2mt, EFG2, MRRF2, MST027, MSTP027, RRF2, RRF2mt, hEFG2, mEF-G 2
UniGene
Hs.277154
Gene Chromosome Location
Chr.5: 74721204-74767371 on Build GRCh38
Transcripts
NM_001281302.1 NM_032380.4 NM_170691.2 NR_104006.1 XM_017009986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4885599 c.1915C>A p.P639T Chr.5: 74725753-74725753 on Build GRCh38
1070022 c.1756A>C p.R586R Chr.5: 74726097-74726097 on Build GRCh38
1070021 c.1766T>C p.V589A Chr.5: 74726087-74726087 on Build GRCh38
7448587 c.1762C>T p.L588F Chr.5: 74726091-74726091 on Build GRCh38
311433 c.1826T>A p.F609Y Chr.5: 74726027-74726027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115273629 Chr.5: 74725930-74725930 on Build GRCh38
rs914924380 Chr.5: 74726053-74726053 on Build GRCh38
rs778262362 Chr.5: 74726188-74726188 on Build GRCh38
rs1052166118 Chr.5: 74725813-74725813 on Build GRCh38
rs1027015939 Chr.5: 74725882-74725882 on Build GRCh38
rs758363436 Chr.5: 74726177-74726177 on Build GRCh38
rs775689852 Chr.5: 74725794-74725794 on Build GRCh38
rs766242615 Chr.5: 74725967-74725967 on Build GRCh38
rs759417609 Chr.5: 74726111-74726111 on Build GRCh38
rs116684547 Chr.5: 74725968-74725968 on Build GRCh38
rs1057291653 Chr.5: 74725842-74725842 on Build GRCh38
rs746698267 Chr.5: 74726018-74726018 on Build GRCh38
rs939309990 Chr.5: 74726250-74726250 on Build GRCh38
rs3058458 Chr.5: 74726281-74726284 on Build GRCh38
rs764298909 Chr.5: 74725768-74725768 on Build GRCh38
rs764105920 Chr.5: 74725953-74725953 on Build GRCh38
rs76339998 Chr.5: 74726027-74726027 on Build GRCh38
rs141759476 Chr.5: 74726028-74726028 on Build GRCh38
rs140077535 Chr.5: 74726125-74726125 on Build GRCh38
rs760789545 Chr.5: 74725805-74725808 on Build GRCh38
rs553982420 Chr.5: 74725861-74725861 on Build GRCh38
rs1028155364 Chr.5: 74725985-74725985 on Build GRCh38
rs753724363 Chr.5: 74726096-74726096 on Build GRCh38
rs771908502 Chr.5: 74725800-74725800 on Build GRCh38
rs964340945 Chr.5: 74726048-74726050 on Build GRCh38
rs1044888497 Chr.5: 74725852-74725852 on Build GRCh38
rs17856872 Chr.5: 74726072-74726072 on Build GRCh38
rs773835525 Chr.5: 74726201-74726201 on Build GRCh38
rs757369852 Chr.5: 74725957-74725957 on Build GRCh38
rs945022234 Chr.5: 74726060-74726060 on Build GRCh38
rs573186339 Chr.5: 74725851-74725851 on Build GRCh38
rs191767372 Chr.5: 74725836-74725836 on Build GRCh38
rs987924608 Chr.5: 74726201-74726203 on Build GRCh38
rs956571633 Chr.5: 74726159-74726159 on Build GRCh38
rs773011043 Chr.5: 74725801-74725801 on Build GRCh38
rs747656336 Chr.5: 74725795-74725795 on Build GRCh38
rs376216209 Chr.5: 74725925-74725925 on Build GRCh38
rs763012299 Chr.5: 74725951-74725951 on Build GRCh38
rs758660730 Chr.5: 74725765-74725765 on Build GRCh38
rs543243430 Chr.5: 74726243-74726243 on Build GRCh38
rs770347396 Chr.5: 74726051-74726051 on Build GRCh38
rs750247207 Chr.5: 74725773-74725773 on Build GRCh38
rs775618593 Chr.5: 74725944-74725944 on Build GRCh38
rs34845453 Chr.5: 74725973-74725973 on Build GRCh38
rs371379739 Chr.5: 74725989-74725989 on Build GRCh38
rs543482000 Chr.5: 74726006-74726006 on Build GRCh38
rs776814499 Chr.5: 74725917-74725917 on Build GRCh38
rs774271112 Chr.5: 74726088-74726088 on Build GRCh38
rs369168318 Chr.5: 74726047-74726047 on Build GRCh38
rs377216980 Chr.5: 74725775-74725775 on Build GRCh38
rs376500994 Chr.5: 74726000-74726000 on Build GRCh38
rs778603093 Chr.5: 74725787-74725787 on Build GRCh38
rs145997856 Chr.5: 74726030-74726030 on Build GRCh38
rs765702596 Chr.5: 74725932-74725932 on Build GRCh38
rs746228528 Chr.5: 74726049-74726049 on Build GRCh38
rs926419065 Chr.5: 74726231-74726231 on Build GRCh38
rs946412135 Chr.5: 74726215-74726215 on Build GRCh38
rs572302480 Chr.5: 74725883-74725883 on Build GRCh38
rs935125008 Chr.5: 74725807-74725807 on Build GRCh38
rs201580206 Chr.5: 74726038-74726038 on Build GRCh38
rs775994813 Chr.5: 74726067-74726067 on Build GRCh38
rs764931275 Chr.5: 74726113-74726116 on Build GRCh38
rs754940538 Chr.5: 74725783-74725783 on Build GRCh38
rs761686309 Chr.5: 74726055-74726055 on Build GRCh38
rs5868753 Chr.5: 74726022-74726027 on Build GRCh38
rs778979352 Chr.5: 74725981-74725981 on Build GRCh38
rs749268103 Chr.5: 74725780-74725780 on Build GRCh38
rs764127754 Chr.5: 74726019-74726020 on Build GRCh38
rs779087043 Chr.5: 74725864-74725864 on Build GRCh38
rs17852782 Chr.5: 74725990-74725990 on Build GRCh38
rs763385749 Chr.5: 74726076-74726076 on Build GRCh38
rs373327256 Chr.5: 74726132-74726132 on Build GRCh38
rs202067734 Chr.5: 74726108-74726108 on Build GRCh38
rs34172202 Chr.5: 74725998-74725998 on Build GRCh38
rs751813986 Chr.5: 74725954-74725954 on Build GRCh38
rs894628083 Chr.5: 74726225-74726225 on Build GRCh38
rs369990115 Chr.5: 74725779-74725779 on Build GRCh38
rs746574696 Chr.5: 74725903-74725903 on Build GRCh38
rs763287631 Chr.5: 74726218-74726218 on Build GRCh38
rs771360110 Chr.5: 74725916-74725916 on Build GRCh38
rs777105528 Chr.5: 74726013-74726013 on Build GRCh38
rs767624997 Chr.5: 74726095-74726095 on Build GRCh38
rs750112708 Chr.5: 74726176-74726177 on Build GRCh38
rs376628780 Chr.5: 74726149-74726149 on Build GRCh38
rs529214882 Chr.5: 74726093-74726093 on Build GRCh38
rs915117121 Chr.5: 74725826-74725826 on Build GRCh38
rs768742394 Chr.5: 74726080-74726080 on Build GRCh38
rs36108518 Chr.5: 74725966-74725968 on Build GRCh38
rs1056764798 Chr.5: 74726255-74726255 on Build GRCh38

View Full Product Details