Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 42565543-42566133 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHR
Entrez Gene ID
2690
Gene Name
growth hormone receptor
Gene Aliases
GHBP, GHIP
UniGene
Hs.125180
Gene Chromosome Location
Chr.5: 42423775-42721878 on Build GRCh38
Transcripts
NM_000163.4 NM_001242399.2 NM_001242400.2 NM_001242401.3 NM_001242402.2 NM_001242403.2 NM_001242404.2 NM_001242405.2 NM_001242406.2 NM_001242460.1 NM_001242462.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4999904 c.62G>A p.G21E Chr.5: 42565936-42565936 on Build GRCh38
5031458 c.40G>T p.G14* Chr.5: 42565914-42565914 on Build GRCh38
738740 c.61G>A p.G21R Chr.5: 42565935-42565935 on Build GRCh38
374501 c.52G>T p.A18S Chr.5: 42565926-42565926 on Build GRCh38
3381265 c.36G>T p.L12L Chr.5: 42565910-42565910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750933471 Chr.5: 42565890-42565890 on Build GRCh38
rs763288729 Chr.5: 42565870-42565870 on Build GRCh38
rs915173701 Chr.5: 42565714-42565714 on Build GRCh38
rs753400591 Chr.5: 42565818-42565818 on Build GRCh38
rs534206265 Chr.5: 42565568-42565568 on Build GRCh38
rs185241673 Chr.5: 42565868-42565868 on Build GRCh38
rs751331390 Chr.5: 42565705-42565705 on Build GRCh38
rs968247066 Chr.5: 42565619-42565619 on Build GRCh38
rs34838342 Chr.5: 42565884-42565884 on Build GRCh38
rs775348305 Chr.5: 42565646-42565646 on Build GRCh38
rs1009355322 Chr.5: 42565968-42565968 on Build GRCh38
rs768651333 Chr.5: 42565993-42565993 on Build GRCh38
rs754496739 Chr.5: 42565827-42565827 on Build GRCh38
rs111623983 Chr.5: 42566056-42566056 on Build GRCh38
rs369651256 Chr.5: 42565905-42565905 on Build GRCh38
rs529128423 Chr.5: 42566014-42566014 on Build GRCh38
rs139137947 Chr.5: 42565706-42565706 on Build GRCh38
rs547124461 Chr.5: 42565920-42565920 on Build GRCh38
rs771574196 Chr.5: 42565831-42565831 on Build GRCh38
rs1013547040 Chr.5: 42565574-42565574 on Build GRCh38
rs752025877 Chr.5: 42565875-42565875 on Build GRCh38
rs774446969 Chr.5: 42565982-42565982 on Build GRCh38
rs931201874 Chr.5: 42566090-42566090 on Build GRCh38
rs1008674325 Chr.5: 42565645-42565645 on Build GRCh38
rs372245866 Chr.5: 42565914-42565914 on Build GRCh38
rs80207133 Chr.5: 42565634-42565634 on Build GRCh38
rs992185320 Chr.5: 42565657-42565657 on Build GRCh38
rs542386921 Chr.5: 42565765-42565765 on Build GRCh38
rs12521020 Chr.5: 42565741-42565741 on Build GRCh38
rs898409340 Chr.5: 42565764-42565764 on Build GRCh38
rs745825846 Chr.5: 42565924-42565924 on Build GRCh38
rs966651629 Chr.5: 42566072-42566072 on Build GRCh38
rs561619571 Chr.5: 42566004-42566004 on Build GRCh38
rs964872770 Chr.5: 42566046-42566046 on Build GRCh38
rs993480083 Chr.5: 42565597-42565597 on Build GRCh38
rs183489953 Chr.5: 42565633-42565633 on Build GRCh38
rs878866020 Chr.5: 42565845-42565845 on Build GRCh38
rs749321748 Chr.5: 42565970-42565970 on Build GRCh38
rs768976063 Chr.5: 42565972-42565972 on Build GRCh38
rs899144611 Chr.5: 42565588-42565588 on Build GRCh38
rs372702393 Chr.5: 42565810-42565810 on Build GRCh38
rs767899669 Chr.5: 42565994-42565994 on Build GRCh38
rs762027736 Chr.5: 42565991-42565991 on Build GRCh38
rs775749224 Chr.5: 42565942-42565942 on Build GRCh38
rs939122321 Chr.5: 42565762-42565762 on Build GRCh38
rs10941579 Chr.5: 42566064-42566064 on Build GRCh38
rs969393107 Chr.5: 42566040-42566040 on Build GRCh38
rs779298515 Chr.5: 42566076-42566076 on Build GRCh38
rs769835036 Chr.5: 42565935-42565935 on Build GRCh38
rs371646052 Chr.5: 42565869-42565869 on Build GRCh38
rs1029168910 Chr.5: 42565610-42565610 on Build GRCh38
rs750507074 Chr.5: 42565861-42565861 on Build GRCh38
rs894724734 Chr.5: 42565571-42565571 on Build GRCh38
rs747188385 Chr.5: 42565867-42565868 on Build GRCh38
rs528933970 Chr.5: 42565899-42565899 on Build GRCh38
rs769555994 Chr.5: 42565586-42565586 on Build GRCh38
rs762698547 Chr.5: 42565658-42565658 on Build GRCh38
rs974957870 Chr.5: 42565592-42565592 on Build GRCh38
rs201804261 Chr.5: 42565865-42565865 on Build GRCh38
rs916032770 Chr.5: 42565804-42565804 on Build GRCh38
rs34089431 Chr.5: 42565725-42565725 on Build GRCh38
rs1047319513 Chr.5: 42565799-42565799 on Build GRCh38
rs756572945 Chr.5: 42565895-42565895 on Build GRCh38
rs992610313 Chr.5: 42565967-42565967 on Build GRCh38
rs977782044 Chr.5: 42565739-42565739 on Build GRCh38
rs577293595 Chr.5: 42566023-42566023 on Build GRCh38
rs543620503 Chr.5: 42565863-42565863 on Build GRCh38
rs374557559 Chr.5: 42565830-42565830 on Build GRCh38
rs538194703 Chr.5: 42566108-42566108 on Build GRCh38
rs764187966 Chr.5: 42565871-42565871 on Build GRCh38
rs564361091 Chr.5: 42565834-42565834 on Build GRCh38
rs908337629 Chr.5: 42566106-42566106 on Build GRCh38
rs571081984 Chr.5: 42566102-42566102 on Build GRCh38
rs763920964 Chr.5: 42565689-42565689 on Build GRCh38
rs34190598 Chr.5: 42565802-42565802 on Build GRCh38
rs746777165 Chr.5: 42565844-42565844 on Build GRCh38
rs552627632 Chr.5: 42565813-42565813 on Build GRCh38
rs376039279 Chr.5: 42565901-42565901 on Build GRCh38
rs964577920 Chr.5: 42565685-42565685 on Build GRCh38
rs931178397 Chr.5: 42565776-42565776 on Build GRCh38
rs539017521 Chr.5: 42566059-42566059 on Build GRCh38
rs756257972 Chr.5: 42565878-42565878 on Build GRCh38
rs866523407 Chr.5: 42565877-42565877 on Build GRCh38

View Full Product Details