Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 141655968-141656567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAP3
Entrez Gene ID
64411
Gene Name
ArfGAP with RhoGAP domain, ankyrin repeat and PH domain 3
Gene Aliases
CENTD3, DRAG1
UniGene
Hs.726187
Gene Chromosome Location
Chr.5: 141653401-141682233 on Build GRCh38
Transcripts
NM_022481.5 XM_005268497.1 XM_005268498.1 XM_005268499.1 XM_006714792.1 XM_011537677.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7000116 c.3806G>A p.R1269Q Chr.5: 141656260-141656260 on Build GRCh38
6529646 c.3882C>G p.F1294L Chr.5: 141656090-141656090 on Build GRCh38
3975294 c.3805C>T p.R1269W Chr.5: 141656261-141656261 on Build GRCh38
3697047 c.3847C>T p.R1283C Chr.5: 141656219-141656219 on Build GRCh38
3611768 c.3756T>C p.R1252R Chr.5: 141656537-141656537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746413805 Chr.5: 141656080-141656080 on Build GRCh38
rs750620055 Chr.5: 141656213-141656213 on Build GRCh38
rs766331562 Chr.5: 141656226-141656226 on Build GRCh38
rs779093646 Chr.5: 141656046-141656046 on Build GRCh38
rs199818610 Chr.5: 141656507-141656507 on Build GRCh38
rs904527499 Chr.5: 141655996-141655996 on Build GRCh38
rs766532762 Chr.5: 141656322-141656322 on Build GRCh38
rs75953884 Chr.5: 141656532-141656532 on Build GRCh38
rs867177043 Chr.5: 141656375-141656375 on Build GRCh38
rs959919471 Chr.5: 141656406-141656406 on Build GRCh38
rs768461512 Chr.5: 141656311-141656311 on Build GRCh38
rs773033723 Chr.5: 141656510-141656510 on Build GRCh38
rs747408158 Chr.5: 141656088-141656088 on Build GRCh38
rs1055378232 Chr.5: 141656305-141656305 on Build GRCh38
rs771642077 Chr.5: 141656279-141656279 on Build GRCh38
rs772681497 Chr.5: 141656083-141656083 on Build GRCh38
rs773830890 Chr.5: 141656118-141656118 on Build GRCh38
rs978869050 Chr.5: 141656452-141656452 on Build GRCh38
rs577518826 Chr.5: 141656341-141656341 on Build GRCh38
rs752150905 Chr.5: 141656228-141656228 on Build GRCh38
rs553896685 Chr.5: 141656387-141656387 on Build GRCh38
rs773604326 Chr.5: 141656317-141656317 on Build GRCh38
rs967651655 Chr.5: 141656448-141656448 on Build GRCh38
rs781188929 Chr.5: 141656072-141656072 on Build GRCh38
rs1036509824 Chr.5: 141656300-141656300 on Build GRCh38
rs931970956 Chr.5: 141656465-141656465 on Build GRCh38
rs368667708 Chr.5: 141656539-141656539 on Build GRCh38
rs770469996 Chr.5: 141656114-141656114 on Build GRCh38
rs897960556 Chr.5: 141656349-141656349 on Build GRCh38
rs762774245 Chr.5: 141656533-141656533 on Build GRCh38
rs547825734 Chr.5: 141656172-141656172 on Build GRCh38
rs374493401 Chr.5: 141656086-141656086 on Build GRCh38
rs750513785 Chr.5: 141656160-141656160 on Build GRCh38
rs890622957 Chr.5: 141656009-141656009 on Build GRCh38
rs757519531 Chr.5: 141656146-141656146 on Build GRCh38
rs200362515 Chr.5: 141656483-141656483 on Build GRCh38
rs928383893 Chr.5: 141656270-141656270 on Build GRCh38
rs2306339 Chr.5: 141656388-141656388 on Build GRCh38
rs778070017 Chr.5: 141656516-141656516 on Build GRCh38
rs776543575 Chr.5: 141656200-141656200 on Build GRCh38
rs867622204 Chr.5: 141656542-141656542 on Build GRCh38
rs775518944 Chr.5: 141656184-141656184 on Build GRCh38
rs551472008 Chr.5: 141656123-141656123 on Build GRCh38
rs932024934 Chr.5: 141656251-141656251 on Build GRCh38
rs375912874 Chr.5: 141656142-141656142 on Build GRCh38
rs963954078 Chr.5: 141656219-141656219 on Build GRCh38
rs996351579 Chr.5: 141656003-141656003 on Build GRCh38
rs768849519 Chr.5: 141656089-141656089 on Build GRCh38
rs993922766 Chr.5: 141656344-141656344 on Build GRCh38
rs150555451 Chr.5: 141656259-141656259 on Build GRCh38
rs866318488 Chr.5: 141656234-141656234 on Build GRCh38
rs747013493 Chr.5: 141656302-141656302 on Build GRCh38
rs772942683 Chr.5: 141656092-141656092 on Build GRCh38
rs751579549 Chr.5: 141656455-141656455 on Build GRCh38
rs755403355 Chr.5: 141656171-141656171 on Build GRCh38
rs368763410 Chr.5: 141656104-141656104 on Build GRCh38
rs768207195 Chr.5: 141656055-141656055 on Build GRCh38
rs770557886 Chr.5: 141656177-141656177 on Build GRCh38
rs201679785 Chr.5: 141656087-141656087 on Build GRCh38
rs1019309565 Chr.5: 141656090-141656090 on Build GRCh38
rs140445106 Chr.5: 141656198-141656198 on Build GRCh38
rs1001228317 Chr.5: 141656385-141656385 on Build GRCh38
rs1037055033 Chr.5: 141656522-141656522 on Build GRCh38
rs775100945 Chr.5: 141656056-141656057 on Build GRCh38
rs952393836 Chr.5: 141656383-141656383 on Build GRCh38
rs376948840 Chr.5: 141656242-141656242 on Build GRCh38
rs992745207 Chr.5: 141656441-141656441 on Build GRCh38
rs762180122 Chr.5: 141656209-141656209 on Build GRCh38
rs370511102 Chr.5: 141656456-141656456 on Build GRCh38
rs754142626 Chr.5: 141656145-141656145 on Build GRCh38
rs773918782 Chr.5: 141656180-141656180 on Build GRCh38
rs758890408 Chr.5: 141656161-141656161 on Build GRCh38
rs745462041 Chr.5: 141656267-141656267 on Build GRCh38
rs372058436 Chr.5: 141656169-141656169 on Build GRCh38
rs771425166 Chr.5: 141656181-141656181 on Build GRCh38
rs370152380 Chr.5: 141656005-141656005 on Build GRCh38
rs539283748 Chr.5: 141656261-141656261 on Build GRCh38
rs369156358 Chr.5: 141656143-141656143 on Build GRCh38
rs555466648 Chr.5: 141656397-141656397 on Build GRCh38
rs765548563 Chr.5: 141656211-141656211 on Build GRCh38
rs376087570 Chr.5: 141656490-141656490 on Build GRCh38
rs753144470 Chr.5: 141656492-141656492 on Build GRCh38
rs371302595 Chr.5: 141656124-141656124 on Build GRCh38
rs763056983 Chr.5: 141656318-141656318 on Build GRCh38
rs765453669 Chr.5: 141656159-141656159 on Build GRCh38
rs374506349 Chr.5: 141656538-141656538 on Build GRCh38
rs185928279 Chr.5: 141656173-141656173 on Build GRCh38
rs376162341 Chr.5: 141656530-141656530 on Build GRCh38
rs201242450 Chr.5: 141656194-141656194 on Build GRCh38
rs780627779 Chr.5: 141656167-141656167 on Build GRCh38
rs774830315 Chr.5: 141656298-141656298 on Build GRCh38
rs200516734 Chr.5: 141656212-141656212 on Build GRCh38
rs576422457 Chr.5: 141656484-141656484 on Build GRCh38
rs775222956 Chr.5: 141656125-141656125 on Build GRCh38
rs769666430 Chr.5: 141656316-141656316 on Build GRCh38
rs760487784 Chr.5: 141656197-141656197 on Build GRCh38
rs913033042 Chr.5: 141656445-141656445 on Build GRCh38
rs181324510 Chr.5: 141656170-141656170 on Build GRCh38
rs533661072 Chr.5: 141656384-141656384 on Build GRCh38
rs1043544302 Chr.5: 141656236-141656236 on Build GRCh38
rs762631922 Chr.5: 141656513-141656515 on Build GRCh38
rs757432961 Chr.5: 141656017-141656017 on Build GRCh38
rs13359221 Chr.5: 141656313-141656313 on Build GRCh38
rs779670102 Chr.5: 141656537-141656537 on Build GRCh38
rs755418658 Chr.5: 141656235-141656235 on Build GRCh38
rs758532450 Chr.5: 141656218-141656218 on Build GRCh38

View Full Product Details