Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31971167-31971736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DXO
Entrez Gene ID
1797
Gene Name
decapping exoribonuclease
Gene Aliases
DOM3L, DOM3Z, NG6, RAI1
UniGene
Hs.153299
Gene Chromosome Location
Chr.6: 31969811-31972290 on Build GRCh38
Transcripts
NM_005510.3 XM_006715005.3 XM_017010329.1 NR_026717.1

Gene Symbol
STK19
Entrez Gene ID
8859
Gene Name
serine/threonine kinase 19
Gene Aliases
D6S60, D6S60E, G11, HLA-RP1, RP1
UniGene
Hs.654371
Gene Chromosome Location
Chr.6: 31971175-31981446 on Build GRCh38
Transcripts
NM_005510.3 XM_006715005.3 XM_017010329.1 NR_026717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6429110 c.323G>A p.C108Y Chr.6: 31971353-31971353 on Build GRCh38
4778311 c.35A>C p.K12T Chr.6: 31971641-31971641 on Build GRCh38
7133796 c.289G>A p.E97K Chr.6: 31971387-31971387 on Build GRCh38
6456847 c.138T>G p.P46P Chr.6: 31971538-31971538 on Build GRCh38
483889 c.27A>G p.G9G Chr.6: 31971649-31971649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762497537 Chr.6: 31971190-31971190 on Build GRCh38
rs11558688 Chr.6: 31971621-31971621 on Build GRCh38
rs201613063 Chr.6: 31971338-31971338 on Build GRCh38
rs755964041 Chr.6: 31971457-31971457 on Build GRCh38
rs921004228 Chr.6: 31971600-31971600 on Build GRCh38
rs759610663 Chr.6: 31971420-31971420 on Build GRCh38
rs375449615 Chr.6: 31971454-31971454 on Build GRCh38
rs371984557 Chr.6: 31971411-31971411 on Build GRCh38
rs367678257 Chr.6: 31971710-31971710 on Build GRCh38
rs749356551 Chr.6: 31971452-31971452 on Build GRCh38
rs748979455 Chr.6: 31971459-31971459 on Build GRCh38
rs1048331374 Chr.6: 31971248-31971248 on Build GRCh38
rs775642249 Chr.6: 31971296-31971296 on Build GRCh38
rs775881905 Chr.6: 31971641-31971641 on Build GRCh38
rs768736059 Chr.6: 31971453-31971454 on Build GRCh38
rs754121281 Chr.6: 31971419-31971419 on Build GRCh38
rs778458569 Chr.6: 31971476-31971476 on Build GRCh38
rs766603641 Chr.6: 31971414-31971414 on Build GRCh38
rs369779413 Chr.6: 31971392-31971392 on Build GRCh38
rs764223713 Chr.6: 31971547-31971547 on Build GRCh38
rs140845331 Chr.6: 31971485-31971485 on Build GRCh38
rs773601388 Chr.6: 31971634-31971635 on Build GRCh38
rs763105492 Chr.6: 31971537-31971537 on Build GRCh38
rs767352315 Chr.6: 31971433-31971433 on Build GRCh38
rs755942402 Chr.6: 31971554-31971554 on Build GRCh38
rs747564963 Chr.6: 31971610-31971610 on Build GRCh38
rs761046928 Chr.6: 31971496-31971496 on Build GRCh38
rs762236357 Chr.6: 31971284-31971284 on Build GRCh38
rs767722945 Chr.6: 31971619-31971619 on Build GRCh38
rs779644491 Chr.6: 31971458-31971458 on Build GRCh38
rs753914062 Chr.6: 31971304-31971304 on Build GRCh38
rs150136570 Chr.6: 31971535-31971535 on Build GRCh38
rs752812055 Chr.6: 31971434-31971434 on Build GRCh38
rs769376829 Chr.6: 31971360-31971360 on Build GRCh38
rs2746395 Chr.6: 31971488-31971488 on Build GRCh38
rs2746396 Chr.6: 31971487-31971487 on Build GRCh38
rs199557774 Chr.6: 31971511-31971511 on Build GRCh38
rs147640853 Chr.6: 31971455-31971455 on Build GRCh38
rs753567260 Chr.6: 31971569-31971569 on Build GRCh38
rs930758003 Chr.6: 31971592-31971592 on Build GRCh38
rs912163807 Chr.6: 31971543-31971543 on Build GRCh38
rs888369714 Chr.6: 31971262-31971262 on Build GRCh38
rs774772156 Chr.6: 31971572-31971576 on Build GRCh38
rs767980181 Chr.6: 31971347-31971347 on Build GRCh38
rs932303712 Chr.6: 31971213-31971213 on Build GRCh38
rs868047896 Chr.6: 31971501-31971501 on Build GRCh38
rs41267118 Chr.6: 31971568-31971568 on Build GRCh38
rs1048222508 Chr.6: 31971607-31971607 on Build GRCh38
rs771331073 Chr.6: 31971620-31971620 on Build GRCh38
rs745656538 Chr.6: 31971350-31971350 on Build GRCh38
rs1036166351 Chr.6: 31971439-31971439 on Build GRCh38
rs200467482 Chr.6: 31971504-31971504 on Build GRCh38
rs750375402 Chr.6: 31971550-31971550 on Build GRCh38
rs958774028 Chr.6: 31971440-31971440 on Build GRCh38
rs774277486 Chr.6: 31971675-31971675 on Build GRCh38
rs746947329 Chr.6: 31971317-31971317 on Build GRCh38
rs759863016 Chr.6: 31971515-31971515 on Build GRCh38
rs754664557 Chr.6: 31971572-31971572 on Build GRCh38
rs201234020 Chr.6: 31971412-31971412 on Build GRCh38
rs771600493 Chr.6: 31971596-31971596 on Build GRCh38
rs369427683 Chr.6: 31971644-31971644 on Build GRCh38
rs755106840 Chr.6: 31971306-31971306 on Build GRCh38
rs149369670 Chr.6: 31971604-31971604 on Build GRCh38
rs574553922 Chr.6: 31971273-31971273 on Build GRCh38
rs897875733 Chr.6: 31971570-31971570 on Build GRCh38
rs1056694 Chr.6: 31971594-31971594 on Build GRCh38
rs756355852 Chr.6: 31971302-31971302 on Build GRCh38
rs767879196 Chr.6: 31971290-31971290 on Build GRCh38
rs1023243249 Chr.6: 31971390-31971390 on Build GRCh38
rs764278833 Chr.6: 31971656-31971656 on Build GRCh38
rs548330202 Chr.6: 31971708-31971708 on Build GRCh38
rs753714393 Chr.6: 31971712-31971712 on Build GRCh38
rs375528246 Chr.6: 31971363-31971363 on Build GRCh38
rs575269411 Chr.6: 31971370-31971370 on Build GRCh38
rs564844657 Chr.6: 31971427-31971427 on Build GRCh38
rs772615090 Chr.6: 31971493-31971493 on Build GRCh38
rs778650301 Chr.6: 31971696-31971696 on Build GRCh38
rs754787619 Chr.6: 31971714-31971714 on Build GRCh38
rs773730664 Chr.6: 31971495-31971495 on Build GRCh38
rs1008974407 Chr.6: 31971207-31971207 on Build GRCh38
rs541936724 Chr.6: 31971282-31971282 on Build GRCh38
rs768213256 Chr.6: 31971385-31971385 on Build GRCh38
rs756834755 Chr.6: 31971690-31971690 on Build GRCh38
rs746190343 Chr.6: 31971631-31971631 on Build GRCh38
rs779097572 Chr.6: 31971308-31971308 on Build GRCh38
rs1012046591 Chr.6: 31971667-31971667 on Build GRCh38
rs747749087 Chr.6: 31971588-31971588 on Build GRCh38
rs766590118 Chr.6: 31971303-31971303 on Build GRCh38
rs762336647 Chr.6: 31971612-31971615 on Build GRCh38
rs1012141352 Chr.6: 31971375-31971375 on Build GRCh38
rs372732739 Chr.6: 31971585-31971585 on Build GRCh38
rs768281949 Chr.6: 31971473-31971473 on Build GRCh38
rs202178198 Chr.6: 31971296-31971296 on Build GRCh38
rs371295094 Chr.6: 31971346-31971346 on Build GRCh38
rs750238921 Chr.6: 31971456-31971456 on Build GRCh38
rs763231141 Chr.6: 31971653-31971653 on Build GRCh38
rs1002990582 Chr.6: 31971355-31971355 on Build GRCh38
rs768006071 Chr.6: 31971405-31971405 on Build GRCh38
rs11558689 Chr.6: 31971639-31971639 on Build GRCh38
rs765552878 Chr.6: 31971517-31971517 on Build GRCh38
rs762362820 Chr.6: 31971393-31971393 on Build GRCh38
rs777122516 Chr.6: 31971628-31971628 on Build GRCh38

View Full Product Details