Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 111694404-111694989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FYN
Entrez Gene ID
2534
Gene Name
FYN proto-oncogene, Src family tyrosine kinase
Gene Aliases
SLK, SYN, p59-FYN
UniGene
Hs.390567
Gene Chromosome Location
Chr.6: 111660332-111873452 on Build GRCh38
Transcripts
NM_002037.5 NM_153047.3 NM_153048.3 XM_005266890.3 XM_005266892.3 XM_017010650.1 XM_017010651.1 XM_017010652.1 XM_017010653.1 XM_017010654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668085 c.1148G>T p.R383L Chr.6: 111694500-111694500 on Build GRCh38
1496221 c.1211C>T p.A404V Chr.6: 111694428-111694428 on Build GRCh38
7326602 c.1158T>A p.Y386* Chr.6: 111694490-111694490 on Build GRCh38
4685942 c.1207A>G p.I403V Chr.6: 111694432-111694432 on Build GRCh38
4685941 c.1216A>G p.I406V Chr.6: 111694432-111694432 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs869063474 Chr.6: 111694622-111694622 on Build GRCh38
rs1020256786 Chr.6: 111694911-111694911 on Build GRCh38
rs371036122 Chr.6: 111694654-111694654 on Build GRCh38
rs115315762 Chr.6: 111694888-111694888 on Build GRCh38
rs775593554 Chr.6: 111694554-111694554 on Build GRCh38
rs745335589 Chr.6: 111694631-111694631 on Build GRCh38
rs550786502 Chr.6: 111694585-111694585 on Build GRCh38
rs144806893 Chr.6: 111694564-111694564 on Build GRCh38
rs760744833 Chr.6: 111694556-111694556 on Build GRCh38
rs774455447 Chr.6: 111694756-111694756 on Build GRCh38
rs755454152 Chr.6: 111694504-111694504 on Build GRCh38
rs981679231 Chr.6: 111694958-111694958 on Build GRCh38
rs148135487 Chr.6: 111694949-111694949 on Build GRCh38
rs138034632 Chr.6: 111694500-111694500 on Build GRCh38
rs935701361 Chr.6: 111694561-111694561 on Build GRCh38
rs778156013 Chr.6: 111694511-111694511 on Build GRCh38
rs368283898 Chr.6: 111694754-111694754 on Build GRCh38
rs778100987 Chr.6: 111694612-111694612 on Build GRCh38
rs548386810 Chr.6: 111694921-111694921 on Build GRCh38
rs897080714 Chr.6: 111694746-111694746 on Build GRCh38
rs776429653 Chr.6: 111694708-111694708 on Build GRCh38
rs754772337 Chr.6: 111694562-111694562 on Build GRCh38
rs763124560 Chr.6: 111694451-111694451 on Build GRCh38
rs182477528 Chr.6: 111694804-111694804 on Build GRCh38
rs779481913 Chr.6: 111694761-111694761 on Build GRCh38
rs866410829 Chr.6: 111694537-111694537 on Build GRCh38
rs771687922 Chr.6: 111694640-111694640 on Build GRCh38
rs1040286611 Chr.6: 111694598-111694598 on Build GRCh38
rs35334544 Chr.6: 111694807-111694808 on Build GRCh38
rs1050977081 Chr.6: 111694819-111694819 on Build GRCh38
rs766475559 Chr.6: 111694472-111694472 on Build GRCh38
rs199592252 Chr.6: 111694536-111694536 on Build GRCh38
rs746940763 Chr.6: 111694658-111694658 on Build GRCh38
rs376029302 Chr.6: 111694876-111694876 on Build GRCh38
rs768772210 Chr.6: 111694687-111694687 on Build GRCh38
rs376486970 Chr.6: 111694522-111694522 on Build GRCh38
rs373532821 Chr.6: 111694505-111694505 on Build GRCh38
rs559426859 Chr.6: 111694576-111694576 on Build GRCh38
rs146616109 Chr.6: 111694449-111694449 on Build GRCh38
rs374665881 Chr.6: 111694610-111694610 on Build GRCh38
rs759563917 Chr.6: 111694584-111694584 on Build GRCh38
rs765507873 Chr.6: 111694444-111694444 on Build GRCh38
rs751647014 Chr.6: 111694474-111694474 on Build GRCh38
rs1036974418 Chr.6: 111694665-111694665 on Build GRCh38
rs756490869 Chr.6: 111694507-111694507 on Build GRCh38
rs376867074 Chr.6: 111694565-111694565 on Build GRCh38
rs201834690 Chr.6: 111694578-111694578 on Build GRCh38
rs748934675 Chr.6: 111694520-111694520 on Build GRCh38
rs772098337 Chr.6: 111694542-111694542 on Build GRCh38
rs58876844 Chr.6: 111694895-111694895 on Build GRCh38
rs753177376 Chr.6: 111694597-111694597 on Build GRCh38
rs804186 Chr.6: 111694620-111694620 on Build GRCh38
rs567684325 Chr.6: 111694875-111694875 on Build GRCh38
rs551266964 Chr.6: 111694860-111694860 on Build GRCh38
rs804187 Chr.6: 111694538-111694538 on Build GRCh38
rs1033083915 Chr.6: 111694884-111694884 on Build GRCh38
rs778465060 Chr.6: 111694622-111694622 on Build GRCh38
rs533306619 Chr.6: 111694733-111694733 on Build GRCh38
rs887864078 Chr.6: 111694825-111694825 on Build GRCh38

View Full Product Details