Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 143394951-143395369 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHA1
Entrez Gene ID
2041
Gene Name
EPH receptor A1
Gene Aliases
EPH, EPHT, EPHT1
UniGene
Hs.89839
Gene Chromosome Location
Chr.7: 143390813-143408892 on Build GRCh38
Transcripts
NM_005232.4 XM_006715880.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3431334 c.2073C>A p.V691V Chr.7: 143395329-143395329 on Build GRCh38
3635252 c.2085A>T p.R695R Chr.7: 143395181-143395181 on Build GRCh38
3878957 c.2044C>A p.H682N Chr.7: 143395358-143395358 on Build GRCh38
5019760 c.2090C>T p.P697L Chr.7: 143395176-143395176 on Build GRCh38
5783131 c.2097G>A p.M699I Chr.7: 143395169-143395169 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755691716 Chr.7: 143395193-143395193 on Build GRCh38
rs768978982 Chr.7: 143395350-143395350 on Build GRCh38
rs752053540 Chr.7: 143395170-143395170 on Build GRCh38
rs894354444 Chr.7: 143395069-143395069 on Build GRCh38
rs745739492 Chr.7: 143395296-143395296 on Build GRCh38
rs779807002 Chr.7: 143395027-143395027 on Build GRCh38
rs765350929 Chr.7: 143395113-143395113 on Build GRCh38
rs762865705 Chr.7: 143394970-143394970 on Build GRCh38
rs117640091 Chr.7: 143395199-143395199 on Build GRCh38
rs994750699 Chr.7: 143395306-143395306 on Build GRCh38
rs796717483 Chr.7: 143395270-143395271 on Build GRCh38
rs765930702 Chr.7: 143395314-143395314 on Build GRCh38
rs774851071 Chr.7: 143395272-143395272 on Build GRCh38
rs754713826 Chr.7: 143395203-143395203 on Build GRCh38
rs774312704 Chr.7: 143394980-143394980 on Build GRCh38
rs144535927 Chr.7: 143395010-143395010 on Build GRCh38
rs758879954 Chr.7: 143395006-143395006 on Build GRCh38
rs778674067 Chr.7: 143395021-143395021 on Build GRCh38
rs576989035 Chr.7: 143395301-143395301 on Build GRCh38
rs139005342 Chr.7: 143395252-143395252 on Build GRCh38
rs375566010 Chr.7: 143395148-143395148 on Build GRCh38
rs760710752 Chr.7: 143394985-143394985 on Build GRCh38
rs768471877 Chr.7: 143395066-143395066 on Build GRCh38
rs200415307 Chr.7: 143395313-143395313 on Build GRCh38
rs770598035 Chr.7: 143395270-143395271 on Build GRCh38
rs749879930 Chr.7: 143395191-143395191 on Build GRCh38
rs147817243 Chr.7: 143395014-143395014 on Build GRCh38
rs117651735 Chr.7: 143395091-143395091 on Build GRCh38
rs1026597748 Chr.7: 143395311-143395311 on Build GRCh38
rs767880997 Chr.7: 143395302-143395302 on Build GRCh38
rs775806265 Chr.7: 143395226-143395226 on Build GRCh38
rs547080889 Chr.7: 143395024-143395024 on Build GRCh38
rs34044440 Chr.7: 143395063-143395064 on Build GRCh38
rs759684433 Chr.7: 143394994-143394994 on Build GRCh38
rs768560146 Chr.7: 143394973-143394973 on Build GRCh38
rs544450689 Chr.7: 143395329-143395329 on Build GRCh38
rs750970922 Chr.7: 143395308-143395308 on Build GRCh38
rs184351147 Chr.7: 143395234-143395234 on Build GRCh38
rs780580035 Chr.7: 143395017-143395017 on Build GRCh38
rs754624005 Chr.7: 143395315-143395315 on Build GRCh38
rs769943843 Chr.7: 143395220-143395220 on Build GRCh38
rs748006734 Chr.7: 143395208-143395208 on Build GRCh38
rs747914291 Chr.7: 143395319-143395319 on Build GRCh38
rs145160235 Chr.7: 143395331-143395331 on Build GRCh38
rs34372369 Chr.7: 143395176-143395176 on Build GRCh38
rs777619076 Chr.7: 143395033-143395036 on Build GRCh38
rs763368841 Chr.7: 143395152-143395152 on Build GRCh38
rs200451105 Chr.7: 143395288-143395288 on Build GRCh38
rs776567061 Chr.7: 143395103-143395103 on Build GRCh38
rs954357442 Chr.7: 143395260-143395260 on Build GRCh38
rs778687443 Chr.7: 143395206-143395206 on Build GRCh38
rs150722351 Chr.7: 143395216-143395216 on Build GRCh38
rs764574963 Chr.7: 143395158-143395158 on Build GRCh38
rs371479433 Chr.7: 143395342-143395342 on Build GRCh38
rs746630136 Chr.7: 143395091-143395092 on Build GRCh38
rs370711332 Chr.7: 143395111-143395111 on Build GRCh38
rs149121680 Chr.7: 143394989-143394989 on Build GRCh38
rs548151465 Chr.7: 143395002-143395002 on Build GRCh38
rs757842125 Chr.7: 143395175-143395175 on Build GRCh38
rs371392412 Chr.7: 143395217-143395217 on Build GRCh38
rs745456870 Chr.7: 143395019-143395019 on Build GRCh38
rs766472808 Chr.7: 143394988-143394988 on Build GRCh38
rs373607991 Chr.7: 143395033-143395033 on Build GRCh38
rs758249861 Chr.7: 143395328-143395328 on Build GRCh38
rs748965673 Chr.7: 143395201-143395201 on Build GRCh38
rs1021907546 Chr.7: 143395332-143395332 on Build GRCh38
rs763282033 Chr.7: 143395229-143395229 on Build GRCh38
rs201356860 Chr.7: 143395073-143395073 on Build GRCh38
rs761755446 Chr.7: 143395075-143395075 on Build GRCh38
rs369122539 Chr.7: 143394999-143394999 on Build GRCh38

View Full Product Details