Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219968230-219968822 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPRS
Entrez Gene ID
2058
Gene Name
glutamyl-prolyl-tRNA synthetase
Gene Aliases
EARS, GLUPRORS, PARS, PIG32, QARS, QPRS
UniGene
Hs.497788
Gene Chromosome Location
Chr.1: 219968600-220046658 on Build GRCh38
Transcripts
NM_004446.2 XM_017000614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5673396 c.4531A>T p.S1511C Chr.1: 219968814-219968814 on Build GRCh38
5950189 c.4529G>A p.R1510H Chr.1: 219968816-219968816 on Build GRCh38
904068 c.4530C>T p.R1510R Chr.1: 219968815-219968815 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs958858191 Chr.1: 219968589-219968589 on Build GRCh38
rs991880813 Chr.1: 219968403-219968403 on Build GRCh38
rs200965352 Chr.1: 219968519-219968519 on Build GRCh38
rs1061242 Chr.1: 219968728-219968728 on Build GRCh38
rs780431884 Chr.1: 219968303-219968306 on Build GRCh38
rs951222029 Chr.1: 219968271-219968271 on Build GRCh38
rs141067518 Chr.1: 219968258-219968258 on Build GRCh38
rs947446457 Chr.1: 219968431-219968431 on Build GRCh38
rs537843175 Chr.1: 219968504-219968504 on Build GRCh38
rs922173187 Chr.1: 219968319-219968319 on Build GRCh38
rs533921328 Chr.1: 219968571-219968571 on Build GRCh38
rs111721811 Chr.1: 219968622-219968622 on Build GRCh38
rs1035552213 Chr.1: 219968581-219968581 on Build GRCh38
rs973550731 Chr.1: 219968757-219968757 on Build GRCh38
rs748368737 Chr.1: 219968769-219968769 on Build GRCh38
rs143197781 Chr.1: 219968260-219968260 on Build GRCh38
rs370451537 Chr.1: 219968772-219968772 on Build GRCh38
rs995179978 Chr.1: 219968510-219968510 on Build GRCh38
rs1022042129 Chr.1: 219968629-219968629 on Build GRCh38
rs1061248 Chr.1: 219968681-219968681 on Build GRCh38
rs538450423 Chr.1: 219968417-219968417 on Build GRCh38
rs933609242 Chr.1: 219968364-219968364 on Build GRCh38
rs3816994 Chr.1: 219968509-219968509 on Build GRCh38
rs560915014 Chr.1: 219968729-219968729 on Build GRCh38
rs954971458 Chr.1: 219968661-219968661 on Build GRCh38
rs546529268 Chr.1: 219968270-219968270 on Build GRCh38
rs889348025 Chr.1: 219968482-219968482 on Build GRCh38
rs770863251 Chr.1: 219968788-219968788 on Build GRCh38
rs56039838 Chr.1: 219968575-219968575 on Build GRCh38
rs910752633 Chr.1: 219968761-219968761 on Build GRCh38
rs776303537 Chr.1: 219968789-219968789 on Build GRCh38
rs569129494 Chr.1: 219968462-219968462 on Build GRCh38
rs920677012 Chr.1: 219968778-219968778 on Build GRCh38
rs777571239 Chr.1: 219968779-219968779 on Build GRCh38
rs958712432 Chr.1: 219968308-219968308 on Build GRCh38
rs1037726823 Chr.1: 219968450-219968450 on Build GRCh38
rs113536284 Chr.1: 219968481-219968481 on Build GRCh38
rs573432340 Chr.1: 219968637-219968637 on Build GRCh38
rs2030078 Chr.1: 219968291-219968291 on Build GRCh38
rs17006931 Chr.1: 219968436-219968436 on Build GRCh38
rs1045083762 Chr.1: 219968508-219968508 on Build GRCh38
rs991456935 Chr.1: 219968611-219968611 on Build GRCh38
rs943660889 Chr.1: 219968487-219968487 on Build GRCh38
rs779035465 Chr.1: 219968753-219968753 on Build GRCh38
rs2270713 Chr.1: 219968752-219968752 on Build GRCh38
rs746928268 Chr.1: 219968787-219968787 on Build GRCh38
rs182059848 Chr.1: 219968432-219968432 on Build GRCh38
rs575488479 Chr.1: 219968705-219968705 on Build GRCh38
rs566685948 Chr.1: 219968399-219968399 on Build GRCh38
rs1051993125 Chr.1: 219968474-219968474 on Build GRCh38
rs796650470 Chr.1: 219968374-219968375 on Build GRCh38
rs1039766488 Chr.1: 219968499-219968499 on Build GRCh38
rs759167401 Chr.1: 219968797-219968797 on Build GRCh38
rs561052571 Chr.1: 219968269-219968269 on Build GRCh38
rs899258891 Chr.1: 219968460-219968460 on Build GRCh38
rs148290712 Chr.1: 219968680-219968680 on Build GRCh38
rs546495595 Chr.1: 219968301-219968301 on Build GRCh38
rs574599956 Chr.1: 219968554-219968554 on Build GRCh38
rs60844853 Chr.1: 219968540-219968540 on Build GRCh38
rs924707521 Chr.1: 219968688-219968688 on Build GRCh38
rs1022017753 Chr.1: 219968580-219968583 on Build GRCh38
rs78654047 Chr.1: 219968580-219968580 on Build GRCh38
rs769398147 Chr.1: 219968799-219968799 on Build GRCh38
rs1030929310 Chr.1: 219968523-219968523 on Build GRCh38
rs1025356969 Chr.1: 219968558-219968558 on Build GRCh38

View Full Product Details