Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 44067543-44068001 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
UniGene
Hs.126706
Gene Chromosome Location
Chr.11: 44066118-44084237 on Build GRCh38
Transcripts
NM_001127219.1 NM_032592.3 XM_005253170.3 XM_005253172.2 XM_006718348.2 XM_011520406.2 XM_011520407.1 XM_011520410.2 XM_017018421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5988591 c.71T>A p.L24Q Chr.11: 44067698-44067698 on Build GRCh38
6352004 c.175G>A p.D59N Chr.11: 44067802-44067802 on Build GRCh38
6518874 c.271G>A p.E91K Chr.11: 44067898-44067898 on Build GRCh38
926940 c.83A>G p.H28R Chr.11: 44067710-44067710 on Build GRCh38
4574361 c.44G>C p.C15S Chr.11: 44067671-44067671 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003840853 Chr.11: 44067758-44067758 on Build GRCh38
rs761684714 Chr.11: 44067778-44067778 on Build GRCh38
rs748472039 Chr.11: 44067618-44067618 on Build GRCh38
rs748022584 Chr.11: 44067894-44067894 on Build GRCh38
rs759193149 Chr.11: 44067683-44067683 on Build GRCh38
rs751645360 Chr.11: 44067701-44067701 on Build GRCh38
rs533546001 Chr.11: 44067691-44067691 on Build GRCh38
rs964763415 Chr.11: 44067636-44067636 on Build GRCh38
rs761751860 Chr.11: 44067694-44067694 on Build GRCh38
rs200566554 Chr.11: 44067709-44067709 on Build GRCh38
rs773353064 Chr.11: 44067903-44067903 on Build GRCh38
rs138153442 Chr.11: 44067810-44067810 on Build GRCh38
rs955555448 Chr.11: 44067889-44067889 on Build GRCh38
rs756616330 Chr.11: 44067725-44067725 on Build GRCh38
rs757960019 Chr.11: 44067865-44067865 on Build GRCh38
rs768407099 Chr.11: 44067801-44067801 on Build GRCh38
rs775163954 Chr.11: 44067625-44067625 on Build GRCh38
rs908478538 Chr.11: 44067727-44067727 on Build GRCh38
rs767703599 Chr.11: 44067793-44067793 on Build GRCh38
rs373957500 Chr.11: 44067659-44067659 on Build GRCh38
rs755271658 Chr.11: 44067702-44067702 on Build GRCh38
rs920559150 Chr.11: 44067665-44067665 on Build GRCh38
rs892073600 Chr.11: 44067933-44067933 on Build GRCh38
rs752940426 Chr.11: 44067714-44067714 on Build GRCh38
rs905134277 Chr.11: 44067743-44067743 on Build GRCh38
rs201286589 Chr.11: 44067610-44067610 on Build GRCh38
rs532337429 Chr.11: 44067570-44067570 on Build GRCh38
rs756306112 Chr.11: 44067805-44067805 on Build GRCh38
rs374629229 Chr.11: 44067880-44067880 on Build GRCh38
rs367984824 Chr.11: 44067770-44067770 on Build GRCh38
rs779480692 Chr.11: 44067869-44067869 on Build GRCh38
rs151181120 Chr.11: 44067604-44067604 on Build GRCh38
rs376307381 Chr.11: 44067827-44067827 on Build GRCh38
rs771549464 Chr.11: 44067766-44067766 on Build GRCh38
rs773258771 Chr.11: 44067787-44067787 on Build GRCh38
rs946560129 Chr.11: 44067721-44067721 on Build GRCh38
rs757809899 Chr.11: 44067953-44067955 on Build GRCh38
rs757728408 Chr.11: 44067843-44067843 on Build GRCh38
rs140350489 Chr.11: 44067789-44067789 on Build GRCh38
rs934659912 Chr.11: 44067660-44067660 on Build GRCh38
rs751742842 Chr.11: 44067615-44067615 on Build GRCh38
rs778186993 Chr.11: 44067638-44067638 on Build GRCh38
rs1031907002 Chr.11: 44067931-44067931 on Build GRCh38
rs765169287 Chr.11: 44067784-44067784 on Build GRCh38
rs548480607 Chr.11: 44067616-44067616 on Build GRCh38
rs753776846 Chr.11: 44067693-44067693 on Build GRCh38
rs141016787 Chr.11: 44067685-44067685 on Build GRCh38
rs766370364 Chr.11: 44067700-44067700 on Build GRCh38
rs764328390 Chr.11: 44067957-44067957 on Build GRCh38
rs200759020 Chr.11: 44067879-44067879 on Build GRCh38
rs178514 Chr.11: 44067611-44067611 on Build GRCh38
rs749776484 Chr.11: 44067732-44067732 on Build GRCh38
rs386753060 Chr.11: 44067610-44067611 on Build GRCh38
rs762972719 Chr.11: 44067914-44067914 on Build GRCh38
rs749926984 Chr.11: 44067640-44067640 on Build GRCh38
rs775287741 Chr.11: 44067688-44067688 on Build GRCh38
rs975547159 Chr.11: 44067572-44067572 on Build GRCh38
rs769442179 Chr.11: 44067898-44067898 on Build GRCh38
rs749799703 Chr.11: 44067647-44067647 on Build GRCh38
rs776419680 Chr.11: 44067776-44067776 on Build GRCh38
rs775648801 Chr.11: 44067928-44067928 on Build GRCh38
rs770258735 Chr.11: 44067623-44067623 on Build GRCh38
rs369928247 Chr.11: 44067862-44067862 on Build GRCh38
rs865883341 Chr.11: 44067818-44067818 on Build GRCh38
rs151059930 Chr.11: 44067671-44067671 on Build GRCh38
rs778317904 Chr.11: 44067737-44067737 on Build GRCh38
rs150220771 Chr.11: 44067769-44067769 on Build GRCh38
rs771403133 Chr.11: 44067736-44067736 on Build GRCh38
rs776308998 Chr.11: 44067883-44067883 on Build GRCh38
rs778191564 Chr.11: 44067728-44067728 on Build GRCh38
rs1044133628 Chr.11: 44067830-44067830 on Build GRCh38
rs760502292 Chr.11: 44067690-44067690 on Build GRCh38
rs372910068 Chr.11: 44067929-44067929 on Build GRCh38
rs33952257 Chr.11: 44067802-44067802 on Build GRCh38
rs1048302440 Chr.11: 44067677-44067677 on Build GRCh38
rs374079881 Chr.11: 44067763-44067763 on Build GRCh38

View Full Product Details