Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 42816072-42816669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SVBP
Entrez Gene ID
374969
Gene Name
small vasohibin binding protein
Gene Aliases
CCDC23
UniGene
Hs.113919
Gene Chromosome Location
Chr.1: 42807052-42817388 on Build GRCh38
Transcripts
NM_199342.3 XM_017001226.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5745624 c.44C>A p.S15Y Chr.1: 42816501-42816501 on Build GRCh38
5832603 c.72_73delGA p.K25fs*30 Chr.1: 42816472-42816473 on Build GRCh38
6562425 c.60G>C p.E20D Chr.1: 42816485-42816485 on Build GRCh38
6997657 c.110C>T p.A37V Chr.1: 42816435-42816435 on Build GRCh38
1296453 c.99G>C p.Q33H Chr.1: 42816446-42816446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751883095 Chr.1: 42816582-42816582 on Build GRCh38
rs1019914873 Chr.1: 42816264-42816264 on Build GRCh38
rs775161924 Chr.1: 42816569-42816569 on Build GRCh38
rs12071025 Chr.1: 42816330-42816330 on Build GRCh38
rs72958986 Chr.1: 42816106-42816106 on Build GRCh38
rs933333247 Chr.1: 42816418-42816418 on Build GRCh38
rs568746320 Chr.1: 42816334-42816334 on Build GRCh38
rs997573284 Chr.1: 42816191-42816191 on Build GRCh38
rs1002526827 Chr.1: 42816534-42816534 on Build GRCh38
rs984614359 Chr.1: 42816335-42816335 on Build GRCh38
rs778418774 Chr.1: 42816435-42816435 on Build GRCh38
rs959900543 Chr.1: 42816331-42816331 on Build GRCh38
rs188917330 Chr.1: 42816575-42816575 on Build GRCh38
rs909077875 Chr.1: 42816338-42816338 on Build GRCh38
rs967295194 Chr.1: 42816268-42816268 on Build GRCh38
rs539580202 Chr.1: 42816174-42816174 on Build GRCh38
rs573089617 Chr.1: 42816296-42816296 on Build GRCh38
rs994410183 Chr.1: 42816107-42816107 on Build GRCh38
rs570145902 Chr.1: 42816593-42816594 on Build GRCh38
rs1050417028 Chr.1: 42816437-42816437 on Build GRCh38
rs763430740 Chr.1: 42816576-42816576 on Build GRCh38
rs909114657 Chr.1: 42816120-42816120 on Build GRCh38
rs760204977 Chr.1: 42816571-42816571 on Build GRCh38
rs971484396 Chr.1: 42816279-42816279 on Build GRCh38
rs752044972 Chr.1: 42816491-42816491 on Build GRCh38
rs202067679 Chr.1: 42816537-42816537 on Build GRCh38
rs776769690 Chr.1: 42816555-42816555 on Build GRCh38
rs770906506 Chr.1: 42816384-42816384 on Build GRCh38
rs563737990 Chr.1: 42816472-42816473 on Build GRCh38
rs1000977241 Chr.1: 42816218-42816218 on Build GRCh38
rs750809370 Chr.1: 42816466-42816466 on Build GRCh38
rs866925955 Chr.1: 42816635-42816635 on Build GRCh38
rs779602646 Chr.1: 42816127-42816127 on Build GRCh38
rs971027965 Chr.1: 42816529-42816529 on Build GRCh38
rs1011281617 Chr.1: 42816242-42816242 on Build GRCh38
rs767112660 Chr.1: 42816417-42816417 on Build GRCh38
rs770996496 Chr.1: 42816531-42816531 on Build GRCh38
rs771407593 Chr.1: 42816166-42816167 on Build GRCh38
rs199914473 Chr.1: 42816519-42816519 on Build GRCh38
rs749174274 Chr.1: 42816533-42816533 on Build GRCh38
rs752729476 Chr.1: 42816515-42816515 on Build GRCh38
rs763156879 Chr.1: 42816470-42816470 on Build GRCh38
rs376257181 Chr.1: 42816394-42816394 on Build GRCh38
rs1018032339 Chr.1: 42816164-42816167 on Build GRCh38
rs756375760 Chr.1: 42816517-42816517 on Build GRCh38
rs531595637 Chr.1: 42816322-42816322 on Build GRCh38
rs190973512 Chr.1: 42816131-42816131 on Build GRCh38
rs111428896 Chr.1: 42816425-42816425 on Build GRCh38
rs563727581 Chr.1: 42816424-42816424 on Build GRCh38
rs766628194 Chr.1: 42816578-42816578 on Build GRCh38
rs56361642 Chr.1: 42816309-42816309 on Build GRCh38
rs940577284 Chr.1: 42816340-42816340 on Build GRCh38
rs753269381 Chr.1: 42816412-42816413 on Build GRCh38
rs756747970 Chr.1: 42816424-42816425 on Build GRCh38
rs1047318275 Chr.1: 42816119-42816119 on Build GRCh38
rs553033776 Chr.1: 42816232-42816232 on Build GRCh38
rs1048947927 Chr.1: 42816111-42816111 on Build GRCh38
rs768354651 Chr.1: 42816573-42816573 on Build GRCh38
rs768377602 Chr.1: 42816241-42816241 on Build GRCh38
rs142728295 Chr.1: 42816584-42816584 on Build GRCh38
rs755093082 Chr.1: 42816492-42816492 on Build GRCh38
rs200991379 Chr.1: 42816552-42816552 on Build GRCh38
rs1032508025 Chr.1: 42816227-42816227 on Build GRCh38
rs756143342 Chr.1: 42816588-42816588 on Build GRCh38
rs1010239274 Chr.1: 42816246-42816246 on Build GRCh38
rs781283021 Chr.1: 42816497-42816497 on Build GRCh38
rs745766902 Chr.1: 42816562-42816562 on Build GRCh38
rs745400426 Chr.1: 42816547-42816551 on Build GRCh38
rs889257946 Chr.1: 42816215-42816215 on Build GRCh38
rs998435951 Chr.1: 42816605-42816605 on Build GRCh38
rs199812799 Chr.1: 42816454-42816454 on Build GRCh38
rs1050482869 Chr.1: 42816210-42816210 on Build GRCh38
rs201807646 Chr.1: 42816450-42816450 on Build GRCh38
rs775319979 Chr.1: 42816426-42816426 on Build GRCh38
rs759175572 Chr.1: 42816398-42816398 on Build GRCh38
rs766974181 Chr.1: 42816476-42816476 on Build GRCh38
rs901841360 Chr.1: 42816156-42816156 on Build GRCh38
rs370680427 Chr.1: 42816563-42816563 on Build GRCh38
rs1009792935 Chr.1: 42816513-42816513 on Build GRCh38
rs748155128 Chr.1: 42816319-42816319 on Build GRCh38
rs182751175 Chr.1: 42816365-42816365 on Build GRCh38
rs78373429 Chr.1: 42816583-42816583 on Build GRCh38

View Full Product Details