Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 10049068-10049596 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKEF1
Entrez Gene ID
63926
Gene Name
ankyrin repeat and EF-hand domain containing 1
Gene Aliases
ANKRD5
UniGene
Hs.70903
Gene Chromosome Location
Chr.20: 10034999-10056764 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Gene Symbol
SNAP25-AS1
Entrez Gene ID
100131208
Gene Name
SNAP25 antisense RNA 1
Gene Aliases
-
UniGene
Hs.658178
Gene Chromosome Location
Chr.20: 10023812-10219506 on Build GRCh38
Transcripts
NM_001303472.1 NM_022096.5 NM_198798.2 XM_005260792.2 NR_040710.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3544030 c.962C>T p.P321L Chr.20: 10049531-10049531 on Build GRCh38
5981663 c.852T>G p.H284Q Chr.20: 10049421-10049421 on Build GRCh38
5526683 c.877G>A p.E293K Chr.20: 10049446-10049446 on Build GRCh38
228730 c.961C>T p.P321S Chr.20: 10049530-10049530 on Build GRCh38
116305 c.924A>T p.R308S Chr.20: 10049493-10049493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905136648 Chr.20: 10049302-10049302 on Build GRCh38
rs540985162 Chr.20: 10049136-10049136 on Build GRCh38
rs184151014 Chr.20: 10049383-10049383 on Build GRCh38
rs908160855 Chr.20: 10049285-10049285 on Build GRCh38
rs937656932 Chr.20: 10049217-10049217 on Build GRCh38
rs1033463399 Chr.20: 10049547-10049547 on Build GRCh38
rs747069695 Chr.20: 10049576-10049576 on Build GRCh38
rs1041792855 Chr.20: 10049533-10049533 on Build GRCh38
rs545543499 Chr.20: 10049359-10049359 on Build GRCh38
rs1014309633 Chr.20: 10049332-10049332 on Build GRCh38
rs758659066 Chr.20: 10049564-10049564 on Build GRCh38
rs926212189 Chr.20: 10049146-10049146 on Build GRCh38
rs933779621 Chr.20: 10049181-10049181 on Build GRCh38
rs1033247909 Chr.20: 10049378-10049378 on Build GRCh38
rs748415063 Chr.20: 10049479-10049479 on Build GRCh38
rs186921087 Chr.20: 10049536-10049536 on Build GRCh38
rs773796694 Chr.20: 10049351-10049351 on Build GRCh38
rs972866021 Chr.20: 10049167-10049167 on Build GRCh38
rs972898520 Chr.20: 10049467-10049467 on Build GRCh38
rs199561328 Chr.20: 10049537-10049537 on Build GRCh38
rs766805289 Chr.20: 10049358-10049358 on Build GRCh38
rs1020806127 Chr.20: 10049333-10049333 on Build GRCh38
rs369257705 Chr.20: 10049452-10049452 on Build GRCh38
rs1038259970 Chr.20: 10049300-10049300 on Build GRCh38
rs373127767 Chr.20: 10049356-10049356 on Build GRCh38
rs926338740 Chr.20: 10049205-10049205 on Build GRCh38
rs757849696 Chr.20: 10049405-10049405 on Build GRCh38
rs777251278 Chr.20: 10049498-10049498 on Build GRCh38
rs757155070 Chr.20: 10049427-10049427 on Build GRCh38
rs750377351 Chr.20: 10049556-10049556 on Build GRCh38
rs747122699 Chr.20: 10049487-10049487 on Build GRCh38
rs148077085 Chr.20: 10049553-10049553 on Build GRCh38
rs770211371 Chr.20: 10049517-10049517 on Build GRCh38
rs772356859 Chr.20: 10049423-10049423 on Build GRCh38
rs756006935 Chr.20: 10049468-10049468 on Build GRCh38
rs778186200 Chr.20: 10049567-10049567 on Build GRCh38
rs1042166323 Chr.20: 10049323-10049323 on Build GRCh38
rs759755317 Chr.20: 10049505-10049505 on Build GRCh38
rs267606071 Chr.20: 10049496-10049496 on Build GRCh38
rs772229318 Chr.20: 10049480-10049480 on Build GRCh38
rs370078042 Chr.20: 10049530-10049530 on Build GRCh38
rs115583213 Chr.20: 10049526-10049526 on Build GRCh38
rs781241130 Chr.20: 10049451-10049451 on Build GRCh38
rs1000634569 Chr.20: 10049349-10049349 on Build GRCh38
rs528690112 Chr.20: 10049497-10049497 on Build GRCh38
rs975547885 Chr.20: 10049390-10049390 on Build GRCh38
rs1046803403 Chr.20: 10049262-10049262 on Build GRCh38
rs778933391 Chr.20: 10049469-10049469 on Build GRCh38
rs576406250 Chr.20: 10049255-10049255 on Build GRCh38
rs980342959 Chr.20: 10049204-10049205 on Build GRCh38
rs903155669 Chr.20: 10049341-10049341 on Build GRCh38
rs114138607 Chr.20: 10049094-10049094 on Build GRCh38
rs759991864 Chr.20: 10049367-10049367 on Build GRCh38
rs149418188 Chr.20: 10049559-10049559 on Build GRCh38
rs779921230 Chr.20: 10049560-10049560 on Build GRCh38
rs941040257 Chr.20: 10049299-10049299 on Build GRCh38
rs1020098763 Chr.20: 10049126-10049126 on Build GRCh38
rs767353145 Chr.20: 10049555-10049555 on Build GRCh38
rs941022745 Chr.20: 10049453-10049453 on Build GRCh38
rs752401938 Chr.20: 10049403-10049403 on Build GRCh38
rs118012679 Chr.20: 10049178-10049178 on Build GRCh38
rs1056398348 Chr.20: 10049511-10049511 on Build GRCh38
rs201956158 Chr.20: 10049285-10049285 on Build GRCh38
rs772282364 Chr.20: 10049345-10049345 on Build GRCh38
rs751463255 Chr.20: 10049421-10049421 on Build GRCh38
rs752926595 Chr.20: 10049562-10049562 on Build GRCh38
rs767077811 Chr.20: 10049567-10049567 on Build GRCh38
rs937808393 Chr.20: 10049304-10049304 on Build GRCh38
rs761863142 Chr.20: 10049473-10049482 on Build GRCh38
rs763619763 Chr.20: 10049411-10049411 on Build GRCh38
rs751048294 Chr.20: 10049539-10049539 on Build GRCh38
rs778026616 Chr.20: 10049482-10049482 on Build GRCh38
rs541316772 Chr.20: 10049329-10049329 on Build GRCh38
rs146891340 Chr.20: 10049493-10049493 on Build GRCh38
rs917670712 Chr.20: 10049561-10049561 on Build GRCh38
rs975577686 Chr.20: 10049177-10049177 on Build GRCh38
rs1056351224 Chr.20: 10049317-10049317 on Build GRCh38
rs1008620786 Chr.20: 10049370-10049370 on Build GRCh38
rs652633 Chr.20: 10049540-10049540 on Build GRCh38
rs964232713 Chr.20: 10049176-10049176 on Build GRCh38

View Full Product Details