Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 32031767-32032345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCM2L
Entrez Gene ID
140706
Gene Name
CCM2 like scaffolding protein
Gene Aliases
C20orf160, dJ310O13.5
UniGene
Hs.382151
Gene Chromosome Location
Chr.20: 32010379-32032182 on Build GRCh38
Transcripts
NM_080625.3 XM_011528566.1 XM_011528567.2 XM_011528568.2 XM_011528569.1

SNP Detail

SNP ID Chromosome Location
rs560376066 Chr.20: 32031914-32031914 on Build GRCh38
rs965090922 Chr.20: 32032295-32032295 on Build GRCh38
rs749130741 Chr.20: 32031797-32031797 on Build GRCh38
rs35570641 Chr.20: 32031872-32031873 on Build GRCh38
rs899526501 Chr.20: 32031793-32031793 on Build GRCh38
rs369522646 Chr.20: 32032190-32032190 on Build GRCh38
rs966955438 Chr.20: 32031963-32031963 on Build GRCh38
rs6119754 Chr.20: 32032120-32032120 on Build GRCh38
rs1048634012 Chr.20: 32031805-32031805 on Build GRCh38
rs909625478 Chr.20: 32032083-32032083 on Build GRCh38
rs912100049 Chr.20: 32031993-32031993 on Build GRCh38
rs192666423 Chr.20: 32031954-32031954 on Build GRCh38
rs1044304164 Chr.20: 32032237-32032237 on Build GRCh38
rs1000609883 Chr.20: 32032272-32032272 on Build GRCh38
rs905754728 Chr.20: 32032245-32032245 on Build GRCh38
rs1055262618 Chr.20: 32031898-32031898 on Build GRCh38
rs939824701 Chr.20: 32032110-32032110 on Build GRCh38
rs184465256 Chr.20: 32032002-32032002 on Build GRCh38
rs1019395004 Chr.20: 32031980-32031981 on Build GRCh38
rs773950174 Chr.20: 32031811-32031811 on Build GRCh38
rs777212559 Chr.20: 32032112-32032112 on Build GRCh38
rs562921464 Chr.20: 32031834-32031834 on Build GRCh38
rs1014158238 Chr.20: 32031877-32031877 on Build GRCh38
rs535463804 Chr.20: 32032155-32032155 on Build GRCh38
rs956112396 Chr.20: 32031986-32031986 on Build GRCh38
rs892918397 Chr.20: 32032230-32032230 on Build GRCh38
rs578158271 Chr.20: 32031848-32031848 on Build GRCh38
rs951871879 Chr.20: 32032010-32032010 on Build GRCh38
rs996246612 Chr.20: 32032300-32032300 on Build GRCh38
rs905605480 Chr.20: 32031887-32031887 on Build GRCh38
rs1045370270 Chr.20: 32032075-32032075 on Build GRCh38
rs1048025150 Chr.20: 32032073-32032073 on Build GRCh38
rs902763981 Chr.20: 32032251-32032251 on Build GRCh38
rs568313801 Chr.20: 32031933-32031934 on Build GRCh38
rs1008894796 Chr.20: 32031969-32031969 on Build GRCh38
rs975013660 Chr.20: 32032037-32032037 on Build GRCh38
rs765703161 Chr.20: 32032216-32032216 on Build GRCh38
rs759896874 Chr.20: 32032144-32032144 on Build GRCh38
rs975234571 Chr.20: 32031998-32031998 on Build GRCh38
rs922153384 Chr.20: 32032044-32032044 on Build GRCh38
rs964788582 Chr.20: 32031982-32031982 on Build GRCh38
rs983311934 Chr.20: 32032025-32032025 on Build GRCh38
rs1042571950 Chr.20: 32032250-32032250 on Build GRCh38
rs1004460485 Chr.20: 32031861-32031861 on Build GRCh38
rs1033520699 Chr.20: 32031956-32031956 on Build GRCh38
rs113297996 Chr.20: 32031833-32031833 on Build GRCh38
rs757020943 Chr.20: 32032284-32032284 on Build GRCh38
rs760710810 Chr.20: 32031992-32031992 on Build GRCh38
rs1001698908 Chr.20: 32031891-32031891 on Build GRCh38
rs528958121 Chr.20: 32032281-32032281 on Build GRCh38
rs547292075 Chr.20: 32031871-32031871 on Build GRCh38
rs141724773 Chr.20: 32031865-32031865 on Build GRCh38
rs926954548 Chr.20: 32032094-32032094 on Build GRCh38
rs752992355 Chr.20: 32032267-32032267 on Build GRCh38
rs768509571 Chr.20: 32031809-32031809 on Build GRCh38
rs6119753 Chr.20: 32031879-32031879 on Build GRCh38
rs772266566 Chr.20: 32031846-32031846 on Build GRCh38
rs150585237 Chr.20: 32032186-32032187 on Build GRCh38
rs6061114 Chr.20: 32032082-32032082 on Build GRCh38
rs907791672 Chr.20: 32032056-32032057 on Build GRCh38
rs970051383 Chr.20: 32031905-32031905 on Build GRCh38
rs895677726 Chr.20: 32032119-32032119 on Build GRCh38
rs978757756 Chr.20: 32031911-32031911 on Build GRCh38
rs114808344 Chr.20: 32032065-32032065 on Build GRCh38
rs1022820546 Chr.20: 32031890-32031890 on Build GRCh38

View Full Product Details