Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 54650778-54651303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOK5
Entrez Gene ID
55816
Gene Name
docking protein 5
Gene Aliases
C20orf180, IRS-6, IRS6
UniGene
Hs.473133
Gene Chromosome Location
Chr.20: 54475472-54651171 on Build GRCh38
Transcripts
NM_018431.4 NM_177959.2 XM_011528904.1

SNP Detail

SNP ID Chromosome Location
rs982880351 Chr.20: 54650958-54650958 on Build GRCh38
rs866061319 Chr.20: 54651125-54651125 on Build GRCh38
rs569425845 Chr.20: 54651046-54651046 on Build GRCh38
rs984155297 Chr.20: 54651058-54651058 on Build GRCh38
rs1034502638 Chr.20: 54651015-54651015 on Build GRCh38
rs553748970 Chr.20: 54651283-54651283 on Build GRCh38
rs1026469291 Chr.20: 54651121-54651121 on Build GRCh38
rs1052100242 Chr.20: 54650865-54650865 on Build GRCh38
rs913669962 Chr.20: 54651225-54651225 on Build GRCh38
rs747025404 Chr.20: 54650869-54650869 on Build GRCh38
rs371667416 Chr.20: 54650901-54650901 on Build GRCh38
rs2841 Chr.20: 54651054-54651054 on Build GRCh38
rs533982941 Chr.20: 54651122-54651122 on Build GRCh38
rs946406743 Chr.20: 54650909-54650909 on Build GRCh38
rs879565399 Chr.20: 54651227-54651227 on Build GRCh38
rs1053521118 Chr.20: 54651045-54651045 on Build GRCh38
rs1018222638 Chr.20: 54650846-54650846 on Build GRCh38
rs760213139 Chr.20: 54651260-54651260 on Build GRCh38
rs980823189 Chr.20: 54651233-54651233 on Build GRCh38
rs1001644275 Chr.20: 54650990-54650990 on Build GRCh38
rs1039117079 Chr.20: 54651095-54651095 on Build GRCh38
rs1007639388 Chr.20: 54650824-54650824 on Build GRCh38
rs183194169 Chr.20: 54650832-54650832 on Build GRCh38
rs895900972 Chr.20: 54651019-54651019 on Build GRCh38
rs1007936960 Chr.20: 54651094-54651094 on Build GRCh38
rs1016850894 Chr.20: 54651074-54651074 on Build GRCh38
rs764782317 Chr.20: 54651088-54651088 on Build GRCh38
rs150414359 Chr.20: 54650806-54650806 on Build GRCh38
rs950935073 Chr.20: 54650899-54650899 on Build GRCh38
rs771015717 Chr.20: 54650939-54650939 on Build GRCh38
rs995034918 Chr.20: 54651101-54651101 on Build GRCh38
rs994964906 Chr.20: 54650864-54650864 on Build GRCh38
rs988257645 Chr.20: 54651171-54651171 on Build GRCh38
rs761687987 Chr.20: 54651084-54651084 on Build GRCh38
rs560989605 Chr.20: 54650847-54650847 on Build GRCh38
rs1026582334 Chr.20: 54650884-54650884 on Build GRCh38
rs890558795 Chr.20: 54650815-54650815 on Build GRCh38
rs1043401201 Chr.20: 54651235-54651235 on Build GRCh38
rs112799548 Chr.20: 54650986-54650986 on Build GRCh38
rs201024891 Chr.20: 54650989-54650989 on Build GRCh38
rs1043678180 Chr.20: 54650919-54650919 on Build GRCh38
rs2842 Chr.20: 54651043-54651043 on Build GRCh38
rs775008631 Chr.20: 54651241-54651241 on Build GRCh38
rs1014502485 Chr.20: 54651157-54651157 on Build GRCh38
rs950985465 Chr.20: 54651146-54651146 on Build GRCh38
rs15899 Chr.20: 54651088-54651088 on Build GRCh38
rs529772477 Chr.20: 54650928-54650928 on Build GRCh38
rs970421866 Chr.20: 54651180-54651180 on Build GRCh38
rs2843 Chr.20: 54651096-54651096 on Build GRCh38
rs2840 Chr.20: 54651037-54651037 on Build GRCh38
rs776671747 Chr.20: 54651020-54651020 on Build GRCh38
rs1024944831 Chr.20: 54651169-54651169 on Build GRCh38

View Full Product Details