Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 6034247-6034713 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRLS1
Entrez Gene ID
54675
Gene Name
cardiolipin synthase 1
Gene Aliases
C20orf155, CLS, CLS1, GCD10, dJ967N21.6
UniGene
Hs.743956
Gene Chromosome Location
Chr.20: 6005938-6040053 on Build GRCh38
Transcripts
NM_001127458.1 NM_001323561.1 NM_001323562.1 NM_001323563.1 NM_001323564.1 NM_019095.5 NR_136617.1 XM_005260738.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4768940 c.814A>T p.I272L Chr.20: 6034548-6034548 on Build GRCh38
7138308 c.786C>T p.F262F Chr.20: 6034520-6034520 on Build GRCh38
2765874 c.759G>A p.V253V Chr.20: 6034493-6034493 on Build GRCh38
7138309 c.489C>T p.F163F Chr.20: 6034520-6034520 on Build GRCh38
6498945 c.465A>G p.A155A Chr.20: 6034496-6034496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780213898 Chr.20: 6034463-6034463 on Build GRCh38
rs748825471 Chr.20: 6034451-6034454 on Build GRCh38
rs772875563 Chr.20: 6034491-6034491 on Build GRCh38
rs6076915 Chr.20: 6034311-6034311 on Build GRCh38
rs950105654 Chr.20: 6034649-6034649 on Build GRCh38
rs763979185 Chr.20: 6034577-6034577 on Build GRCh38
rs1055965954 Chr.20: 6034601-6034601 on Build GRCh38
rs776326624 Chr.20: 6034532-6034532 on Build GRCh38
rs146060368 Chr.20: 6034543-6034543 on Build GRCh38
rs752375104 Chr.20: 6034548-6034548 on Build GRCh38
rs540034167 Chr.20: 6034576-6034577 on Build GRCh38
rs781262385 Chr.20: 6034415-6034415 on Build GRCh38
rs769049210 Chr.20: 6034468-6034468 on Build GRCh38
rs374478862 Chr.20: 6034431-6034431 on Build GRCh38
rs117802504 Chr.20: 6034271-6034271 on Build GRCh38
rs200256769 Chr.20: 6034464-6034464 on Build GRCh38
rs750537554 Chr.20: 6034578-6034578 on Build GRCh38
rs920837701 Chr.20: 6034545-6034545 on Build GRCh38
rs961719191 Chr.20: 6034400-6034400 on Build GRCh38
rs748695489 Chr.20: 6034480-6034480 on Build GRCh38
rs756160131 Chr.20: 6034605-6034605 on Build GRCh38
rs930919590 Chr.20: 6034592-6034592 on Build GRCh38
rs6139898 Chr.20: 6034308-6034308 on Build GRCh38
rs925185122 Chr.20: 6034487-6034487 on Build GRCh38
rs745672754 Chr.20: 6034268-6034268 on Build GRCh38
rs943713635 Chr.20: 6034576-6034576 on Build GRCh38
rs577275848 Chr.20: 6034362-6034362 on Build GRCh38
rs1010729081 Chr.20: 6034269-6034269 on Build GRCh38
rs969211190 Chr.20: 6034441-6034441 on Build GRCh38
rs962039156 Chr.20: 6034292-6034292 on Build GRCh38
rs201221373 Chr.20: 6034607-6034607 on Build GRCh38
rs745898147 Chr.20: 6034458-6034458 on Build GRCh38
rs778776613 Chr.20: 6034602-6034603 on Build GRCh38
rs754594687 Chr.20: 6034291-6034292 on Build GRCh38
rs199802039 Chr.20: 6034486-6034486 on Build GRCh38
rs987219708 Chr.20: 6034338-6034338 on Build GRCh38
rs957968396 Chr.20: 6034503-6034503 on Build GRCh38
rs765083673 Chr.20: 6034547-6034547 on Build GRCh38
rs1025490240 Chr.20: 6034371-6034371 on Build GRCh38
rs765029620 Chr.20: 6034444-6034449 on Build GRCh38
rs567026622 Chr.20: 6034656-6034656 on Build GRCh38
rs772670785 Chr.20: 6034286-6034286 on Build GRCh38
rs760223500 Chr.20: 6034508-6034508 on Build GRCh38
rs367739543 Chr.20: 6034522-6034522 on Build GRCh38
rs142267934 Chr.20: 6034620-6034620 on Build GRCh38
rs542392435 Chr.20: 6034679-6034679 on Build GRCh38
rs774789705 Chr.20: 6034471-6034471 on Build GRCh38
rs758352834 Chr.20: 6034554-6034554 on Build GRCh38

View Full Product Details