Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44290940-44291484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AIRE
Entrez Gene ID
326
Gene Name
autoimmune regulator
Gene Aliases
AIRE1, APECED, APS1, APSI, PGA1
UniGene
Hs.129829
Gene Chromosome Location
Chr.21: 44285838-44298219 on Build GRCh38
Transcripts
NM_000383.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4660679 c.346G>A p.G116S Chr.21: 44291152-44291152 on Build GRCh38
4101921 c.967C>A p.L323M Chr.21: 44291182-44291182 on Build GRCh38
6161630 c.990C>A p.I330I Chr.21: 44291205-44291205 on Build GRCh38
1592790 c.158G>A p.G53E Chr.21: 44290964-44290964 on Build GRCh38
6161632 c.922C>G p.L308V Chr.21: 44291137-44291137 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780919403 Chr.21: 44291050-44291050 on Build GRCh38
rs561479459 Chr.21: 44291418-44291418 on Build GRCh38
rs745665855 Chr.21: 44291056-44291056 on Build GRCh38
rs754932526 Chr.21: 44291131-44291131 on Build GRCh38
rs145274715 Chr.21: 44291163-44291163 on Build GRCh38
rs1078480 Chr.21: 44291319-44291319 on Build GRCh38
rs937101384 Chr.21: 44291337-44291337 on Build GRCh38
rs2073613 Chr.21: 44291320-44291320 on Build GRCh38
rs139659079 Chr.21: 44291077-44291077 on Build GRCh38
rs1800524 Chr.21: 44291261-44291261 on Build GRCh38
rs200741288 Chr.21: 44291248-44291248 on Build GRCh38
rs768040995 Chr.21: 44291000-44291000 on Build GRCh38
rs761114243 Chr.21: 44291084-44291084 on Build GRCh38
rs758528374 Chr.21: 44291240-44291240 on Build GRCh38
rs547823315 Chr.21: 44291249-44291249 on Build GRCh38
rs1030975173 Chr.21: 44291308-44291308 on Build GRCh38
rs766234328 Chr.21: 44291223-44291223 on Build GRCh38
rs139808903 Chr.21: 44291123-44291123 on Build GRCh38
rs914447650 Chr.21: 44291341-44291341 on Build GRCh38
rs74162062 Chr.21: 44291142-44291142 on Build GRCh38
rs556665590 Chr.21: 44291353-44291353 on Build GRCh38
rs1054395568 Chr.21: 44290977-44290977 on Build GRCh38
rs149769247 Chr.21: 44291127-44291127 on Build GRCh38
rs386833675 Chr.21: 44291182-44291194 on Build GRCh38
rs777217133 Chr.21: 44291017-44291017 on Build GRCh38
rs772333071 Chr.21: 44291079-44291079 on Build GRCh38
rs147889221 Chr.21: 44291151-44291151 on Build GRCh38
rs114771343 Chr.21: 44291349-44291349 on Build GRCh38
rs936714310 Chr.21: 44291140-44291140 on Build GRCh38
rs766114406 Chr.21: 44291260-44291260 on Build GRCh38
rs179363885 Chr.21: 44291192-44291192 on Build GRCh38
rs763636007 Chr.21: 44291107-44291107 on Build GRCh38
rs751066946 Chr.21: 44291112-44291112 on Build GRCh38
rs769768095 Chr.21: 44290962-44290962 on Build GRCh38
rs760103978 Chr.21: 44291181-44291181 on Build GRCh38
rs776013167 Chr.21: 44291251-44291251 on Build GRCh38
rs1036182929 Chr.21: 44291370-44291370 on Build GRCh38
rs780049983 Chr.21: 44291065-44291065 on Build GRCh38
rs773506800 Chr.21: 44291083-44291083 on Build GRCh38
rs141955483 Chr.21: 44291088-44291088 on Build GRCh38
rs542739016 Chr.21: 44291411-44291411 on Build GRCh38
rs567075906 Chr.21: 44291233-44291233 on Build GRCh38
rs970283322 Chr.21: 44291342-44291342 on Build GRCh38
rs757935716 Chr.21: 44291015-44291015 on Build GRCh38
rs1057517011 Chr.21: 44291192-44291192 on Build GRCh38
rs139874934 Chr.21: 44291161-44291161 on Build GRCh38
rs946284099 Chr.21: 44291282-44291282 on Build GRCh38
rs766901260 Chr.21: 44291209-44291209 on Build GRCh38
rs924001537 Chr.21: 44291139-44291139 on Build GRCh38
rs775495071 Chr.21: 44290965-44290965 on Build GRCh38
rs74162063 Chr.21: 44291197-44291197 on Build GRCh38
rs922714199 Chr.21: 44291053-44291053 on Build GRCh38
rs897758151 Chr.21: 44291386-44291386 on Build GRCh38
rs137910738 Chr.21: 44290985-44290985 on Build GRCh38
rs536338321 Chr.21: 44291115-44291115 on Build GRCh38
rs550679632 Chr.21: 44291060-44291060 on Build GRCh38
rs542469295 Chr.21: 44291394-44291394 on Build GRCh38
rs766896493 Chr.21: 44291011-44291011 on Build GRCh38
rs386833674 Chr.21: 44291147-44291147 on Build GRCh38
rs35271728 Chr.21: 44291448-44291449 on Build GRCh38
rs748426061 Chr.21: 44291345-44291345 on Build GRCh38
rs747297616 Chr.21: 44291245-44291245 on Build GRCh38
rs527944302 Chr.21: 44291106-44291106 on Build GRCh38
rs897353805 Chr.21: 44291081-44291081 on Build GRCh38
rs747034666 Chr.21: 44291232-44291232 on Build GRCh38
rs761394248 Chr.21: 44291208-44291208 on Build GRCh38
rs777218693 Chr.21: 44291089-44291089 on Build GRCh38
rs977998923 Chr.21: 44291290-44291290 on Build GRCh38
rs889987563 Chr.21: 44291419-44291419 on Build GRCh38
rs565602272 Chr.21: 44291465-44291465 on Build GRCh38
rs768876713 Chr.21: 44291076-44291076 on Build GRCh38
rs749304027 Chr.21: 44291248-44291249 on Build GRCh38
rs1036633160 Chr.21: 44291205-44291205 on Build GRCh38
rs372011129 Chr.21: 44291029-44291029 on Build GRCh38
rs142503647 Chr.21: 44291259-44291259 on Build GRCh38
rs59281354 Chr.21: 44291210-44291210 on Build GRCh38
rs202027254 Chr.21: 44291162-44291162 on Build GRCh38
rs1016946107 Chr.21: 44290971-44290971 on Build GRCh38
rs768447024 Chr.21: 44291381-44291381 on Build GRCh38
rs56393821 Chr.21: 44291117-44291117 on Build GRCh38
rs745835455 Chr.21: 44290960-44290960 on Build GRCh38
rs775921321 Chr.21: 44291198-44291198 on Build GRCh38
rs150634562 Chr.21: 44291116-44291116 on Build GRCh38
rs763315098 Chr.21: 44291255-44291255 on Build GRCh38
rs749193829 Chr.21: 44291075-44291075 on Build GRCh38
rs565153757 Chr.21: 44290979-44290979 on Build GRCh38
rs776951380 Chr.21: 44291171-44291171 on Build GRCh38
rs868756561 Chr.21: 44291114-44291114 on Build GRCh38
rs1006980686 Chr.21: 44291431-44291431 on Build GRCh38
rs372856026 Chr.21: 44291213-44291213 on Build GRCh38
rs777693599 Chr.21: 44291241-44291241 on Build GRCh38
rs750768166 Chr.21: 44291009-44291009 on Build GRCh38
rs756873062 Chr.21: 44291043-44291043 on Build GRCh38
rs1036662603 Chr.21: 44291364-44291364 on Build GRCh38
rs142297552 Chr.21: 44291010-44291010 on Build GRCh38
rs375855661 Chr.21: 44291215-44291215 on Build GRCh38
rs763110689 Chr.21: 44290967-44290967 on Build GRCh38
rs762165526 Chr.21: 44291078-44291078 on Build GRCh38
rs573444005 Chr.21: 44291429-44291429 on Build GRCh38
rs778929451 Chr.21: 44291122-44291122 on Build GRCh38
rs762075832 Chr.21: 44290990-44290990 on Build GRCh38
rs755022448 Chr.21: 44291225-44291225 on Build GRCh38
rs372997338 Chr.21: 44291256-44291256 on Build GRCh38
rs1041193627 Chr.21: 44291463-44291463 on Build GRCh38
rs375893942 Chr.21: 44291072-44291072 on Build GRCh38
rs972756453 Chr.21: 44291193-44291193 on Build GRCh38
rs753679257 Chr.21: 44291224-44291224 on Build GRCh38
rs1029648118 Chr.21: 44291194-44291194 on Build GRCh38
rs754240163 Chr.21: 44291012-44291012 on Build GRCh38
rs931103650 Chr.21: 44291391-44291391 on Build GRCh38
rs540594107 Chr.21: 44291456-44291456 on Build GRCh38
rs376422124 Chr.21: 44291390-44291390 on Build GRCh38
rs749284009 Chr.21: 44291130-44291130 on Build GRCh38

View Full Product Details