Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46308551-46309112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GTSE1
Entrez Gene ID
51512
Gene Name
G2 and S-phase expressed 1
Gene Aliases
B99
UniGene
Hs.386189
Gene Chromosome Location
Chr.22: 46295879-46333699 on Build GRCh38
Transcripts
NM_016426.6 XM_005261627.3 XM_011530211.2 XM_011530213.2 XM_017028815.1 XM_017028816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7208364 c.597C>T p.L199L Chr.22: 46308778-46308778 on Build GRCh38
7076789 c.463C>T p.L155L Chr.22: 46308701-46308701 on Build GRCh38
1190480 c.467G>A p.G156E Chr.22: 46308705-46308705 on Build GRCh38
6506286 c.556C>T p.P186S Chr.22: 46308794-46308794 on Build GRCh38
1416875 c.435_436insAACC p.L147fs*4 Chr.22: 46308616-46308617 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760751223 Chr.22: 46308799-46308799 on Build GRCh38
rs375274769 Chr.22: 46308678-46308678 on Build GRCh38
rs770026510 Chr.22: 46308665-46308665 on Build GRCh38
rs34660725 Chr.22: 46308658-46308658 on Build GRCh38
rs367919168 Chr.22: 46308694-46308694 on Build GRCh38
rs758457992 Chr.22: 46308651-46308651 on Build GRCh38
rs747735330 Chr.22: 46308750-46308751 on Build GRCh38
rs998599922 Chr.22: 46308935-46308935 on Build GRCh38
rs565932345 Chr.22: 46309057-46309057 on Build GRCh38
rs753402714 Chr.22: 46308962-46308962 on Build GRCh38
rs759546180 Chr.22: 46308848-46308848 on Build GRCh38
rs760065053 Chr.22: 46308741-46308741 on Build GRCh38
rs746454453 Chr.22: 46308655-46308655 on Build GRCh38
rs764918768 Chr.22: 46308695-46308695 on Build GRCh38
rs149905369 Chr.22: 46308730-46308730 on Build GRCh38
rs752974296 Chr.22: 46308616-46308616 on Build GRCh38
rs563452761 Chr.22: 46309008-46309008 on Build GRCh38
rs538178807 Chr.22: 46308952-46308952 on Build GRCh38
rs6008600 Chr.22: 46308779-46308779 on Build GRCh38
rs768217704 Chr.22: 46309029-46309029 on Build GRCh38
rs138493417 Chr.22: 46308673-46308673 on Build GRCh38
rs113335037 Chr.22: 46308745-46308745 on Build GRCh38
rs748370009 Chr.22: 46308914-46308914 on Build GRCh38
rs929204921 Chr.22: 46308771-46308771 on Build GRCh38
rs766428983 Chr.22: 46308580-46308580 on Build GRCh38
rs548602439 Chr.22: 46308901-46308901 on Build GRCh38
rs563937975 Chr.22: 46308915-46308915 on Build GRCh38
rs753240292 Chr.22: 46308729-46308729 on Build GRCh38
rs983305261 Chr.22: 46308775-46308775 on Build GRCh38
rs756897484 Chr.22: 46308963-46308963 on Build GRCh38
rs749583399 Chr.22: 46308726-46308726 on Build GRCh38
rs753807788 Chr.22: 46308675-46308675 on Build GRCh38
rs570881035 Chr.22: 46308782-46308782 on Build GRCh38
rs767426101 Chr.22: 46308858-46308858 on Build GRCh38
rs56268435 Chr.22: 46308787-46308787 on Build GRCh38
rs141545267 Chr.22: 46308570-46308570 on Build GRCh38
rs747868772 Chr.22: 46308660-46308660 on Build GRCh38
rs755597920 Chr.22: 46308867-46308867 on Build GRCh38
rs199893603 Chr.22: 46308795-46308795 on Build GRCh38
rs145330697 Chr.22: 46308874-46308874 on Build GRCh38
rs527323907 Chr.22: 46308631-46308631 on Build GRCh38
rs549293420 Chr.22: 46309039-46309039 on Build GRCh38
rs566751040 Chr.22: 46308896-46308896 on Build GRCh38
rs764310133 Chr.22: 46308750-46308750 on Build GRCh38
rs752322552 Chr.22: 46308786-46308786 on Build GRCh38
rs910087344 Chr.22: 46308993-46308993 on Build GRCh38
rs866124881 Chr.22: 46308705-46308705 on Build GRCh38
rs747502735 Chr.22: 46308897-46308897 on Build GRCh38
rs763541676 Chr.22: 46308873-46308873 on Build GRCh38
rs543176342 Chr.22: 46308984-46308984 on Build GRCh38
rs762788802 Chr.22: 46308762-46308762 on Build GRCh38
rs763635576 Chr.22: 46308961-46308961 on Build GRCh38
rs767657741 Chr.22: 46308615-46308615 on Build GRCh38
rs750098566 Chr.22: 46308981-46308981 on Build GRCh38
rs945990548 Chr.22: 46309070-46309070 on Build GRCh38
rs34404175 Chr.22: 46308837-46308837 on Build GRCh38
rs6007819 Chr.22: 46309019-46309019 on Build GRCh38
rs149616575 Chr.22: 46308789-46308789 on Build GRCh38
rs747530341 Chr.22: 46308991-46308991 on Build GRCh38
rs754021337 Chr.22: 46308754-46308754 on Build GRCh38
rs773577660 Chr.22: 46308922-46308922 on Build GRCh38
rs963660849 Chr.22: 46308960-46308960 on Build GRCh38
rs759313739 Chr.22: 46308934-46308934 on Build GRCh38
rs199691143 Chr.22: 46308725-46308725 on Build GRCh38
rs768751251 Chr.22: 46308900-46308900 on Build GRCh38
rs147669373 Chr.22: 46308936-46308936 on Build GRCh38
rs780797269 Chr.22: 46308717-46308717 on Build GRCh38
rs746351114 Chr.22: 46308794-46308794 on Build GRCh38
rs779235500 Chr.22: 46308875-46308875 on Build GRCh38
rs34499849 Chr.22: 46308970-46308971 on Build GRCh38
rs780557753 Chr.22: 46308783-46308783 on Build GRCh38
rs779706575 Chr.22: 46308653-46308653 on Build GRCh38
rs756952008 Chr.22: 46308619-46308619 on Build GRCh38
rs780082841 Chr.22: 46308708-46308708 on Build GRCh38
rs371420066 Chr.22: 46308948-46308948 on Build GRCh38
rs758749914 Chr.22: 46308983-46308983 on Build GRCh38
rs752585625 Chr.22: 46308859-46308859 on Build GRCh38
rs11913324 Chr.22: 46308790-46308790 on Build GRCh38
rs1024695842 Chr.22: 46308879-46308879 on Build GRCh38
rs201046742 Chr.22: 46308839-46308839 on Build GRCh38
rs772107261 Chr.22: 46308744-46308744 on Build GRCh38
rs746298166 Chr.22: 46308884-46308884 on Build GRCh38
rs754817124 Chr.22: 46308680-46308681 on Build GRCh38
rs940117756 Chr.22: 46308804-46308804 on Build GRCh38
rs1012683069 Chr.22: 46308894-46308894 on Build GRCh38
rs201133511 Chr.22: 46308774-46308774 on Build GRCh38
rs939322604 Chr.22: 46309054-46309054 on Build GRCh38
rs984278199 Chr.22: 46308985-46308985 on Build GRCh38
rs763360056 Chr.22: 46308676-46308676 on Build GRCh38
rs779875858 Chr.22: 46308978-46308978 on Build GRCh38
rs371896148 Chr.22: 46308657-46308657 on Build GRCh38
rs915798857 Chr.22: 46309068-46309068 on Build GRCh38
rs763557911 Chr.22: 46308577-46308577 on Build GRCh38
rs1054614 Chr.22: 46308621-46308621 on Build GRCh38
rs750280204 Chr.22: 46308581-46308581 on Build GRCh38
rs536511117 Chr.22: 46308798-46308798 on Build GRCh38
rs1042867086 Chr.22: 46309088-46309088 on Build GRCh38
rs745761570 Chr.22: 46308635-46308635 on Build GRCh38
rs35141277 Chr.22: 46308713-46308713 on Build GRCh38
rs778688311 Chr.22: 46308749-46308750 on Build GRCh38
rs928082083 Chr.22: 46309038-46309038 on Build GRCh38
rs200502529 Chr.22: 46308937-46308937 on Build GRCh38
rs371660161 Chr.22: 46308702-46308702 on Build GRCh38
rs774348655 Chr.22: 46308844-46308844 on Build GRCh38
rs375036882 Chr.22: 46308720-46308720 on Build GRCh38
rs1055153466 Chr.22: 46309059-46309059 on Build GRCh38
rs776901641 Chr.22: 46308904-46308904 on Build GRCh38
rs768641304 Chr.22: 46308802-46308802 on Build GRCh38
rs200181605 Chr.22: 46308603-46308603 on Build GRCh38
rs116016223 Chr.22: 46308793-46308793 on Build GRCh38
rs34725504 Chr.22: 46308683-46308683 on Build GRCh38
rs569884891 Chr.22: 46308690-46308690 on Build GRCh38
rs888545598 Chr.22: 46308709-46308709 on Build GRCh38
rs758703273 Chr.22: 46308892-46308892 on Build GRCh38
rs528383028 Chr.22: 46309071-46309071 on Build GRCh38
rs143271063 Chr.22: 46308977-46308977 on Build GRCh38
rs35225564 Chr.22: 46308821-46308822 on Build GRCh38
rs141501144 Chr.22: 46308680-46308680 on Build GRCh38
rs760795052 Chr.22: 46308747-46308747 on Build GRCh38
rs776875143 Chr.22: 46308830-46308830 on Build GRCh38
rs539781674 Chr.22: 46308693-46308693 on Build GRCh38
rs1010516706 Chr.22: 46308697-46308697 on Build GRCh38

View Full Product Details