Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 21001562-21002144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOB
Entrez Gene ID
338
Gene Name
apolipoprotein B
Gene Aliases
FLDB, LDLCQ4, apoB-100, apoB-48
UniGene
Hs.120759
Gene Chromosome Location
Chr.2: 21001429-21044073 on Build GRCh38
Transcripts
NM_000384.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015886 c.13399G>T p.A4467S Chr.2: 21002023-21002023 on Build GRCh38
1405049 c.13527C>A p.Y4509* Chr.2: 21001895-21001895 on Build GRCh38
1015882 c.13538T>G p.F4513C Chr.2: 21001884-21001884 on Build GRCh38
7324023 c.13487G>A p.R4496K Chr.2: 21001935-21001935 on Build GRCh38
1183248 c.13442C>T p.A4481V Chr.2: 21001980-21001980 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1000065533 Chr.2: 21002116-21002116 on Build GRCh38
rs758028252 Chr.2: 21001768-21001768 on Build GRCh38
rs377641527 Chr.2: 21001814-21001814 on Build GRCh38
rs779345915 Chr.2: 21002018-21002018 on Build GRCh38
rs72654428 Chr.2: 21001719-21001719 on Build GRCh38
rs900908225 Chr.2: 21002028-21002028 on Build GRCh38
rs1021009945 Chr.2: 21001864-21001864 on Build GRCh38
rs770231338 Chr.2: 21001841-21001841 on Build GRCh38
rs374044926 Chr.2: 21002115-21002115 on Build GRCh38
rs138421941 Chr.2: 21001973-21001973 on Build GRCh38
rs1048669409 Chr.2: 21001599-21001599 on Build GRCh38
rs771143412 Chr.2: 21001946-21001946 on Build GRCh38
rs72654429 Chr.2: 21001628-21001628 on Build GRCh38
rs374389311 Chr.2: 21001959-21001959 on Build GRCh38
rs771608572 Chr.2: 21002047-21002047 on Build GRCh38
rs374018447 Chr.2: 21001775-21001775 on Build GRCh38
rs755251454 Chr.2: 21001934-21001934 on Build GRCh38
rs748727595 Chr.2: 21001815-21001815 on Build GRCh38
rs749868276 Chr.2: 21001965-21001965 on Build GRCh38
rs750076573 Chr.2: 21002102-21002102 on Build GRCh38
rs762466056 Chr.2: 21001710-21001710 on Build GRCh38
rs777726812 Chr.2: 21001784-21001784 on Build GRCh38
rs763773989 Chr.2: 21001872-21001872 on Build GRCh38
rs771286073 Chr.2: 21001968-21001968 on Build GRCh38
rs756887607 Chr.2: 21001582-21001582 on Build GRCh38
rs886055571 Chr.2: 21001821-21001821 on Build GRCh38
rs886055569 Chr.2: 21001645-21001645 on Build GRCh38
rs754253964 Chr.2: 21002114-21002114 on Build GRCh38
rs986583835 Chr.2: 21002110-21002110 on Build GRCh38
rs938535592 Chr.2: 21001818-21001818 on Build GRCh38
rs773326388 Chr.2: 21002087-21002087 on Build GRCh38
rs763341676 Chr.2: 21001846-21001846 on Build GRCh38
rs1801695 Chr.2: 21001981-21001981 on Build GRCh38
rs767091894 Chr.2: 21001858-21001858 on Build GRCh38
rs72654427 Chr.2: 21001742-21001742 on Build GRCh38
rs756692121 Chr.2: 21001828-21001828 on Build GRCh38
rs749915303 Chr.2: 21001743-21001743 on Build GRCh38
rs370286693 Chr.2: 21001976-21001976 on Build GRCh38
rs142702699 Chr.2: 21001978-21001978 on Build GRCh38
rs769181415 Chr.2: 21001692-21001692 on Build GRCh38
rs776450015 Chr.2: 21002066-21002066 on Build GRCh38
rs1030430429 Chr.2: 21002082-21002082 on Build GRCh38
rs746353350 Chr.2: 21002045-21002045 on Build GRCh38
rs749634224 Chr.2: 21001937-21001937 on Build GRCh38
rs776276258 Chr.2: 21001977-21001977 on Build GRCh38
rs201587166 Chr.2: 21002006-21002006 on Build GRCh38
rs756545438 Chr.2: 21001939-21001939 on Build GRCh38
rs146661220 Chr.2: 21002000-21002000 on Build GRCh38
rs886055570 Chr.2: 21001763-21001763 on Build GRCh38
rs746732981 Chr.2: 21002061-21002061 on Build GRCh38
rs953483853 Chr.2: 21002103-21002103 on Build GRCh38
rs1021679810 Chr.2: 21001596-21001596 on Build GRCh38
rs938275256 Chr.2: 21001586-21001586 on Build GRCh38
rs770544459 Chr.2: 21001853-21001853 on Build GRCh38
rs200374122 Chr.2: 21001970-21001970 on Build GRCh38
rs574659380 Chr.2: 21002013-21002013 on Build GRCh38
rs760957637 Chr.2: 21002104-21002104 on Build GRCh38
rs778140846 Chr.2: 21001833-21001833 on Build GRCh38
rs757136007 Chr.2: 21001915-21001915 on Build GRCh38
rs756787301 Chr.2: 21001901-21001902 on Build GRCh38
rs756281873 Chr.2: 21002002-21002002 on Build GRCh38
rs781520074 Chr.2: 21001936-21001936 on Build GRCh38
rs200469887 Chr.2: 21002080-21002080 on Build GRCh38
rs12713450 Chr.2: 21001971-21001971 on Build GRCh38
rs146687572 Chr.2: 21001752-21001752 on Build GRCh38
rs762982664 Chr.2: 21002088-21002088 on Build GRCh38
rs746534965 Chr.2: 21001639-21001639 on Build GRCh38
rs747210676 Chr.2: 21001691-21001691 on Build GRCh38
rs758554859 Chr.2: 21001797-21001797 on Build GRCh38
rs886055568 Chr.2: 21001613-21001613 on Build GRCh38
rs897737982 Chr.2: 21001817-21001817 on Build GRCh38
rs747300780 Chr.2: 21001806-21001806 on Build GRCh38
rs141477444 Chr.2: 21001922-21001922 on Build GRCh38
rs753852142 Chr.2: 21001892-21001892 on Build GRCh38
rs562574661 Chr.2: 21001940-21001942 on Build GRCh38
rs956901460 Chr.2: 21001714-21001714 on Build GRCh38
rs749549578 Chr.2: 21002010-21002010 on Build GRCh38
rs546806987 Chr.2: 21001867-21001867 on Build GRCh38
rs748453503 Chr.2: 21001839-21001839 on Build GRCh38
rs183812948 Chr.2: 21002053-21002053 on Build GRCh38
rs776485634 Chr.2: 21002079-21002079 on Build GRCh38
rs773822083 Chr.2: 21001842-21001842 on Build GRCh38
rs752665378 Chr.2: 21002091-21002091 on Build GRCh38
rs764183555 Chr.2: 21002112-21002112 on Build GRCh38
rs371437581 Chr.2: 21001856-21001856 on Build GRCh38
rs769871675 Chr.2: 21002085-21002085 on Build GRCh38
rs759187968 Chr.2: 21001761-21001761 on Build GRCh38
rs888095957 Chr.2: 21001600-21001600 on Build GRCh38
rs188019153 Chr.2: 21001699-21001699 on Build GRCh38
rs768132227 Chr.2: 21002064-21002064 on Build GRCh38
rs866063436 Chr.2: 21002074-21002074 on Build GRCh38
rs774487085 Chr.2: 21001966-21001966 on Build GRCh38
rs751629013 Chr.2: 21001933-21001933 on Build GRCh38
rs779964877 Chr.2: 21001801-21001801 on Build GRCh38
rs566760356 Chr.2: 21001919-21001919 on Build GRCh38
rs1003873446 Chr.2: 21001616-21001616 on Build GRCh38
rs770273010 Chr.2: 21001712-21001712 on Build GRCh38
rs753345874 Chr.2: 21001698-21001700 on Build GRCh38
rs755057102 Chr.2: 21001982-21001982 on Build GRCh38
rs112667751 Chr.2: 21001905-21001905 on Build GRCh38
rs764014607 Chr.2: 21001975-21001975 on Build GRCh38
rs956933186 Chr.2: 21002063-21002063 on Build GRCh38
rs767676919 Chr.2: 21001995-21001995 on Build GRCh38
rs1049527523 Chr.2: 21002055-21002055 on Build GRCh38
rs773841094 Chr.2: 21001855-21001855 on Build GRCh38
rs958014928 Chr.2: 21001836-21001836 on Build GRCh38
rs145832414 Chr.2: 21001771-21001771 on Build GRCh38
rs147416761 Chr.2: 21001974-21001974 on Build GRCh38
rs767768628 Chr.2: 21002089-21002089 on Build GRCh38
rs760551634 Chr.2: 21002011-21002011 on Build GRCh38

View Full Product Details