Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 62331182-62331731 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf14
Entrez Gene ID
57415
Gene Name
chromosome 3 open reading frame 14
Gene Aliases
HT021
UniGene
Hs.47166
Gene Chromosome Location
Chr.3: 62318973-62336213 on Build GRCh38
Transcripts
NM_001291941.1 NM_001291942.1 NM_020685.4 XM_017006931.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4119724 c.204A>G p.L68L Chr.3: 62331351-62331351 on Build GRCh38
1047914 c.179A>G p.D60G Chr.3: 62331326-62331326 on Build GRCh38
584715 c.229C>T p.R77W Chr.3: 62331376-62331376 on Build GRCh38
3596927 c.200C>T p.S67L Chr.3: 62331347-62331347 on Build GRCh38
7094889 c.245C>A p.S82Y Chr.3: 62331392-62331392 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033253326 Chr.3: 62331706-62331706 on Build GRCh38
rs761938086 Chr.3: 62331225-62331226 on Build GRCh38
rs757007909 Chr.3: 62331369-62331369 on Build GRCh38
rs558094271 Chr.3: 62331599-62331599 on Build GRCh38
rs767067224 Chr.3: 62331446-62331447 on Build GRCh38
rs761806675 Chr.3: 62331245-62331245 on Build GRCh38
rs780990044 Chr.3: 62331376-62331376 on Build GRCh38
rs537526827 Chr.3: 62331678-62331678 on Build GRCh38
rs766341593 Chr.3: 62331317-62331317 on Build GRCh38
rs753858875 Chr.3: 62331338-62331338 on Build GRCh38
rs774204763 Chr.3: 62331299-62331299 on Build GRCh38
rs545276471 Chr.3: 62331629-62331629 on Build GRCh38
rs377492209 Chr.3: 62331420-62331420 on Build GRCh38
rs774080294 Chr.3: 62331406-62331406 on Build GRCh38
rs377684604 Chr.3: 62331348-62331348 on Build GRCh38
rs1004457653 Chr.3: 62331648-62331648 on Build GRCh38
rs926378032 Chr.3: 62331549-62331549 on Build GRCh38
rs368619330 Chr.3: 62331321-62331321 on Build GRCh38
rs766783517 Chr.3: 62331428-62331428 on Build GRCh38
rs967665238 Chr.3: 62331677-62331677 on Build GRCh38
rs908507333 Chr.3: 62331436-62331437 on Build GRCh38
rs376111558 Chr.3: 62331320-62331320 on Build GRCh38
rs899626246 Chr.3: 62331248-62331248 on Build GRCh38
rs567089108 Chr.3: 62331340-62331340 on Build GRCh38
rs1029856889 Chr.3: 62331254-62331254 on Build GRCh38
rs765336051 Chr.3: 62331437-62331437 on Build GRCh38
rs1000417449 Chr.3: 62331702-62331702 on Build GRCh38
rs565782937 Chr.3: 62331415-62331415 on Build GRCh38
rs191007223 Chr.3: 62331440-62331440 on Build GRCh38
rs988252976 Chr.3: 62331271-62331271 on Build GRCh38
rs965400777 Chr.3: 62331250-62331250 on Build GRCh38
rs769584152 Chr.3: 62331382-62331382 on Build GRCh38
rs759034843 Chr.3: 62331437-62331440 on Build GRCh38
rs764337435 Chr.3: 62331451-62331451 on Build GRCh38
rs768185451 Chr.3: 62331298-62331298 on Build GRCh38
rs953364153 Chr.3: 62331313-62331313 on Build GRCh38
rs777208778 Chr.3: 62331435-62331435 on Build GRCh38
rs180755007 Chr.3: 62331310-62331310 on Build GRCh38
rs937741021 Chr.3: 62331567-62331567 on Build GRCh38
rs748731086 Chr.3: 62331401-62331401 on Build GRCh38
rs753040845 Chr.3: 62331688-62331688 on Build GRCh38
rs200467315 Chr.3: 62331403-62331403 on Build GRCh38
rs186089632 Chr.3: 62331347-62331347 on Build GRCh38
rs138007949 Chr.3: 62331211-62331211 on Build GRCh38
rs1048548624 Chr.3: 62331579-62331579 on Build GRCh38
rs545127392 Chr.3: 62331209-62331209 on Build GRCh38
rs1015558714 Chr.3: 62331221-62331221 on Build GRCh38
rs767996396 Chr.3: 62331311-62331311 on Build GRCh38
rs998717190 Chr.3: 62331268-62331268 on Build GRCh38
rs747831876 Chr.3: 62331412-62331412 on Build GRCh38
rs539441604 Chr.3: 62331502-62331502 on Build GRCh38
rs762560688 Chr.3: 62331283-62331283 on Build GRCh38
rs1009286028 Chr.3: 62331574-62331574 on Build GRCh38
rs1050947164 Chr.3: 62331524-62331524 on Build GRCh38
rs772515832 Chr.3: 62331413-62331413 on Build GRCh38
rs35261777 Chr.3: 62331330-62331330 on Build GRCh38
rs890905664 Chr.3: 62331531-62331531 on Build GRCh38
rs774057339 Chr.3: 62331426-62331429 on Build GRCh38
rs745708122 Chr.3: 62331377-62331377 on Build GRCh38
rs374112102 Chr.3: 62331443-62331443 on Build GRCh38
rs761065667 Chr.3: 62331325-62331325 on Build GRCh38
rs1046592770 Chr.3: 62331507-62331507 on Build GRCh38
rs780102330 Chr.3: 62331395-62331395 on Build GRCh38
rs113682843 Chr.3: 62331318-62331318 on Build GRCh38
rs371543865 Chr.3: 62331353-62331353 on Build GRCh38

View Full Product Details