Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47161478-47162077 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CALM2
Entrez Gene ID
805
Gene Name
calmodulin 2
Gene Aliases
CAMII, LQT15, PHKD, PHKD2, caM
UniGene
Hs.468442
Gene Chromosome Location
Chr.2: 47160082-47176936 on Build GRCh38
Transcripts
NM_001305624.1 NM_001305625.1 NM_001305626.1 NM_001743.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1283910 c.290G>C p.G97A Chr.2: 47161854-47161854 on Build GRCh38
4545645 c.382G>A p.E128K Chr.2: 47161762-47161762 on Build GRCh38
6232984 c.360A>G p.E120E Chr.2: 47161784-47161784 on Build GRCh38
3581871 c.319C>T p.R107C Chr.2: 47161825-47161825 on Build GRCh38
4612613 c.417_418insCT p.E140fs*7 Chr.2: 47161726-47161727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760234446 Chr.2: 47161536-47161536 on Build GRCh38
rs570786521 Chr.2: 47161996-47161996 on Build GRCh38
rs113446790 Chr.2: 47161627-47161627 on Build GRCh38
rs1023423013 Chr.2: 47161939-47161939 on Build GRCh38
rs554708053 Chr.2: 47161578-47161578 on Build GRCh38
rs751334551 Chr.2: 47161808-47161808 on Build GRCh38
rs200594401 Chr.2: 47161740-47161740 on Build GRCh38
rs546507069 Chr.2: 47161796-47161796 on Build GRCh38
rs761547095 Chr.2: 47161823-47161823 on Build GRCh38
rs755216773 Chr.2: 47161895-47161895 on Build GRCh38
rs111287024 Chr.2: 47162009-47162009 on Build GRCh38
rs145787488 Chr.2: 47161577-47161577 on Build GRCh38
rs148957461 Chr.2: 47161582-47161582 on Build GRCh38
rs532281556 Chr.2: 47161542-47161542 on Build GRCh38
rs530280284 Chr.2: 47161618-47161618 on Build GRCh38
rs750307914 Chr.2: 47161863-47161863 on Build GRCh38
rs2454084 Chr.2: 47161626-47161626 on Build GRCh38
rs3729956 Chr.2: 47161648-47161648 on Build GRCh38
rs957191434 Chr.2: 47161538-47161538 on Build GRCh38
rs548132869 Chr.2: 47162014-47162014 on Build GRCh38
rs11551437 Chr.2: 47161833-47161833 on Build GRCh38
rs201444040 Chr.2: 47161724-47161724 on Build GRCh38
rs916716582 Chr.2: 47162043-47162043 on Build GRCh38
rs999872183 Chr.2: 47161892-47161892 on Build GRCh38
rs17850324 Chr.2: 47161774-47161774 on Build GRCh38
rs143704343 Chr.2: 47161910-47161910 on Build GRCh38
rs1010697319 Chr.2: 47161921-47161921 on Build GRCh38
rs756159706 Chr.2: 47161879-47161879 on Build GRCh38
rs34169936 Chr.2: 47161830-47161831 on Build GRCh38
rs372947059 Chr.2: 47161720-47161720 on Build GRCh38
rs1011415595 Chr.2: 47161853-47161853 on Build GRCh38
rs1011653901 Chr.2: 47161551-47161551 on Build GRCh38
rs574833849 Chr.2: 47161664-47161664 on Build GRCh38
rs576033867 Chr.2: 47161706-47161706 on Build GRCh38
rs909420359 Chr.2: 47161585-47161587 on Build GRCh38
rs942178620 Chr.2: 47161598-47161598 on Build GRCh38
rs989250211 Chr.2: 47161576-47161576 on Build GRCh38
rs111887282 Chr.2: 47161585-47161585 on Build GRCh38
rs2454085 Chr.2: 47161627-47161627 on Build GRCh38
rs571239564 Chr.2: 47161537-47161537 on Build GRCh38
rs138521350 Chr.2: 47161625-47161626 on Build GRCh38
rs730882254 Chr.2: 47161857-47161857 on Build GRCh38
rs879596925 Chr.2: 47161626-47161627 on Build GRCh38
rs976129290 Chr.2: 47161571-47161571 on Build GRCh38
rs182560859 Chr.2: 47162026-47162026 on Build GRCh38
rs552074188 Chr.2: 47162035-47162035 on Build GRCh38
rs780167668 Chr.2: 47161885-47161885 on Build GRCh38
rs398124650 Chr.2: 47161744-47161744 on Build GRCh38
rs999440442 Chr.2: 47161518-47161518 on Build GRCh38
rs374968950 Chr.2: 47161716-47161716 on Build GRCh38
rs199921656 Chr.2: 47161690-47161690 on Build GRCh38
rs398124648 Chr.2: 47161748-47161748 on Build GRCh38
rs113557417 Chr.2: 47161607-47161607 on Build GRCh38
rs547194410 Chr.2: 47161543-47161543 on Build GRCh38
rs758625359 Chr.2: 47161642-47161642 on Build GRCh38
rs186427740 Chr.2: 47162028-47162028 on Build GRCh38
rs1055668652 Chr.2: 47161602-47161602 on Build GRCh38
rs775065505 Chr.2: 47161765-47161765 on Build GRCh38
rs965121733 Chr.2: 47161561-47161561 on Build GRCh38
rs542095802 Chr.2: 47161702-47161702 on Build GRCh38
rs983412031 Chr.2: 47162017-47162017 on Build GRCh38
rs72877072 Chr.2: 47161573-47161573 on Build GRCh38
rs751754595 Chr.2: 47161712-47161713 on Build GRCh38
rs2016682 Chr.2: 47161705-47161705 on Build GRCh38
rs370698055 Chr.2: 47161893-47161893 on Build GRCh38
rs749848022 Chr.2: 47161623-47161624 on Build GRCh38
rs960088136 Chr.2: 47162022-47162022 on Build GRCh38
rs1030819981 Chr.2: 47161557-47161557 on Build GRCh38
rs779466443 Chr.2: 47161896-47161896 on Build GRCh38
rs773732456 Chr.2: 47161710-47161710 on Build GRCh38
rs549847269 Chr.2: 47161512-47161512 on Build GRCh38
rs547325050 Chr.2: 47161961-47161961 on Build GRCh38
rs398124649 Chr.2: 47161737-47161737 on Build GRCh38
rs779412690 Chr.2: 47161704-47161704 on Build GRCh38
rs1048558484 Chr.2: 47161666-47161666 on Build GRCh38
rs398124647 Chr.2: 47161851-47161851 on Build GRCh38
rs1030369451 Chr.2: 47161989-47161989 on Build GRCh38
rs3729955 Chr.2: 47161844-47161844 on Build GRCh38
rs928652203 Chr.2: 47162020-47162020 on Build GRCh38
rs112882205 Chr.2: 47161877-47161878 on Build GRCh38
rs561747169 Chr.2: 47161875-47161876 on Build GRCh38
rs564738016 Chr.2: 47161841-47161841 on Build GRCh38
rs748541560 Chr.2: 47161897-47161897 on Build GRCh38
rs1017689503 Chr.2: 47161558-47161558 on Build GRCh38
rs752901781 Chr.2: 47161979-47161979 on Build GRCh38
rs772418940 Chr.2: 47161903-47161903 on Build GRCh38
rs755983446 Chr.2: 47161958-47161958 on Build GRCh38
rs997997305 Chr.2: 47161957-47161957 on Build GRCh38
rs201028012 Chr.2: 47161625-47161625 on Build GRCh38
rs559193755 Chr.2: 47161984-47161984 on Build GRCh38
rs762591090 Chr.2: 47161790-47161790 on Build GRCh38
rs915677379 Chr.2: 47161607-47161608 on Build GRCh38
rs11551460 Chr.2: 47161843-47161843 on Build GRCh38
rs367611069 Chr.2: 47161701-47161701 on Build GRCh38
rs113581500 Chr.2: 47161626-47161627 on Build GRCh38
rs757451233 Chr.2: 47161948-47161948 on Build GRCh38

View Full Product Details