Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89273169-89273696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FANCI
Entrez Gene ID
55215
Gene Name
Fanconi anemia complementation group I
Gene Aliases
KIAA1794
UniGene
Hs.513126
Gene Chromosome Location
Chr.15: 89243954-89317132 on Build GRCh38
Transcripts
NM_001113378.1 NM_018193.2 XM_011521756.2 XM_011521757.2 XM_011521764.2 XM_011521767.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5367864 c.975+3_975+4insT p.? Chr.15: 89273472-89273473 on Build GRCh38
6660498 c.969G>A p.Q323Q Chr.15: 89273463-89273463 on Build GRCh38
5155051 c.975+4delT p.? Chr.15: 89273473-89273473 on Build GRCh38
3936895 c.898G>A p.D300N Chr.15: 89273392-89273392 on Build GRCh38
5977681 c.958C>T p.Q320* Chr.15: 89273452-89273452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771315184 Chr.15: 89273506-89273506 on Build GRCh38
rs750839825 Chr.15: 89273354-89273354 on Build GRCh38
rs542389647 Chr.15: 89273420-89273420 on Build GRCh38
rs777033124 Chr.15: 89273509-89273509 on Build GRCh38
rs539753367 Chr.15: 89273529-89273535 on Build GRCh38
rs867727416 Chr.15: 89273219-89273219 on Build GRCh38
rs1017405664 Chr.15: 89273665-89273665 on Build GRCh38
rs59945953 Chr.15: 89273326-89273326 on Build GRCh38
rs752942375 Chr.15: 89273513-89273513 on Build GRCh38
rs760217615 Chr.15: 89273311-89273311 on Build GRCh38
rs960217181 Chr.15: 89273606-89273606 on Build GRCh38
rs754088170 Chr.15: 89273479-89273479 on Build GRCh38
rs765354483 Chr.15: 89273328-89273331 on Build GRCh38
rs756685742 Chr.15: 89273511-89273517 on Build GRCh38
rs765028410 Chr.15: 89273511-89273512 on Build GRCh38
rs753646184 Chr.15: 89273516-89273516 on Build GRCh38
rs1034296739 Chr.15: 89273591-89273591 on Build GRCh38
rs909889244 Chr.15: 89273277-89273277 on Build GRCh38
rs376369914 Chr.15: 89273326-89273327 on Build GRCh38
rs766563407 Chr.15: 89273427-89273429 on Build GRCh38
rs756884560 Chr.15: 89273512-89273514 on Build GRCh38
rs16942922 Chr.15: 89273511-89273511 on Build GRCh38
rs77107320 Chr.15: 89273297-89273297 on Build GRCh38
rs759815521 Chr.15: 89273480-89273480 on Build GRCh38
rs761782799 Chr.15: 89273466-89273466 on Build GRCh38
rs919072141 Chr.15: 89273612-89273612 on Build GRCh38
rs749013382 Chr.15: 89273428-89273428 on Build GRCh38
rs752817738 Chr.15: 89273484-89273484 on Build GRCh38
rs918702019 Chr.15: 89273234-89273234 on Build GRCh38
rs771609410 Chr.15: 89273518-89273518 on Build GRCh38
rs74442202 Chr.15: 89273310-89273310 on Build GRCh38
rs762462713 Chr.15: 89273336-89273336 on Build GRCh38
rs942662158 Chr.15: 89273281-89273281 on Build GRCh38
rs59956491 Chr.15: 89273535-89273536 on Build GRCh38
rs796945711 Chr.15: 89273326-89273327 on Build GRCh38
rs769485443 Chr.15: 89273413-89273413 on Build GRCh38
rs1039253043 Chr.15: 89273318-89273318 on Build GRCh38
rs766659063 Chr.15: 89273476-89273476 on Build GRCh38
rs763087620 Chr.15: 89273361-89273362 on Build GRCh38
rs745489708 Chr.15: 89273330-89273330 on Build GRCh38
rs775024411 Chr.15: 89273334-89273334 on Build GRCh38
rs141073698 Chr.15: 89273440-89273440 on Build GRCh38
rs745623288 Chr.15: 89273412-89273412 on Build GRCh38
rs773701774 Chr.15: 89273503-89273503 on Build GRCh38
rs778739892 Chr.15: 89273501-89273501 on Build GRCh38
rs755931346 Chr.15: 89273493-89273493 on Build GRCh38
rs60761593 Chr.15: 89273327-89273327 on Build GRCh38
rs866916992 Chr.15: 89273469-89273469 on Build GRCh38
rs777154321 Chr.15: 89273511-89273514 on Build GRCh38
rs537221499 Chr.15: 89273194-89273194 on Build GRCh38
rs559708641 Chr.15: 89273619-89273619 on Build GRCh38
rs769529791 Chr.15: 89273332-89273332 on Build GRCh38
rs574372732 Chr.15: 89273263-89273263 on Build GRCh38
rs911553744 Chr.15: 89273307-89273307 on Build GRCh38
rs771496697 Chr.15: 89273502-89273502 on Build GRCh38
rs762466296 Chr.15: 89273345-89273345 on Build GRCh38
rs1014077986 Chr.15: 89273617-89273617 on Build GRCh38
rs558814083 Chr.15: 89273512-89273512 on Build GRCh38
rs754986558 Chr.15: 89273380-89273380 on Build GRCh38
rs763699645 Chr.15: 89273341-89273341 on Build GRCh38
rs565632197 Chr.15: 89273321-89273322 on Build GRCh38
rs528357302 Chr.15: 89273547-89273547 on Build GRCh38
rs753333895 Chr.15: 89273511-89273511 on Build GRCh38
rs993053635 Chr.15: 89273230-89273230 on Build GRCh38
rs751890595 Chr.15: 89273473-89273473 on Build GRCh38
rs753571742 Chr.15: 89273640-89273640 on Build GRCh38
rs778590467 Chr.15: 89273512-89273513 on Build GRCh38
rs761042580 Chr.15: 89273473-89273473 on Build GRCh38
rs779806221 Chr.15: 89273512-89273516 on Build GRCh38
rs980706697 Chr.15: 89273198-89273198 on Build GRCh38
rs1024430096 Chr.15: 89273450-89273450 on Build GRCh38
rs963640007 Chr.15: 89273583-89273583 on Build GRCh38
rs763860993 Chr.15: 89273512-89273512 on Build GRCh38
rs545096970 Chr.15: 89273350-89273350 on Build GRCh38
rs747532056 Chr.15: 89273519-89273519 on Build GRCh38
rs773615217 Chr.15: 89273472-89273472 on Build GRCh38
rs563201817 Chr.15: 89273368-89273368 on Build GRCh38
rs984126550 Chr.15: 89273272-89273272 on Build GRCh38
rs377561215 Chr.15: 89273408-89273408 on Build GRCh38
rs773333718 Chr.15: 89273334-89273334 on Build GRCh38
rs748925759 Chr.15: 89273497-89273497 on Build GRCh38
rs575416245 Chr.15: 89273399-89273399 on Build GRCh38
rs35436382 Chr.15: 89273451-89273451 on Build GRCh38
rs904194707 Chr.15: 89273549-89273549 on Build GRCh38
rs1033579219 Chr.15: 89273563-89273563 on Build GRCh38
rs922821639 Chr.15: 89273362-89273362 on Build GRCh38
rs72300014 Chr.15: 89273311-89273311 on Build GRCh38
rs556124953 Chr.15: 89273260-89273260 on Build GRCh38
rs796280343 Chr.15: 89273511-89273512 on Build GRCh38
rs758508365 Chr.15: 89273485-89273485 on Build GRCh38
rs984178681 Chr.15: 89273644-89273644 on Build GRCh38
rs746939739 Chr.15: 89273385-89273385 on Build GRCh38
rs10707260 Chr.15: 89273310-89273310 on Build GRCh38
rs774051668 Chr.15: 89273444-89273444 on Build GRCh38
rs34985075 Chr.15: 89273535-89273535 on Build GRCh38
rs12591472 Chr.15: 89273207-89273207 on Build GRCh38
rs767961759 Chr.15: 89273511-89273518 on Build GRCh38
rs375180333 Chr.15: 89273468-89273468 on Build GRCh38
rs779768567 Chr.15: 89273494-89273494 on Build GRCh38
rs35764601 Chr.15: 89273538-89273538 on Build GRCh38
rs771907104 Chr.15: 89273512-89273515 on Build GRCh38
rs756052900 Chr.15: 89273515-89273515 on Build GRCh38
rs113970806 Chr.15: 89273327-89273327 on Build GRCh38
rs573813614 Chr.15: 89273327-89273331 on Build GRCh38
rs374558367 Chr.15: 89273310-89273311 on Build GRCh38
rs764144724 Chr.15: 89273514-89273514 on Build GRCh38
rs750328805 Chr.15: 89273488-89273488 on Build GRCh38
rs778651951 Chr.15: 89273517-89273517 on Build GRCh38
rs553508408 Chr.15: 89273226-89273226 on Build GRCh38
rs374951861 Chr.15: 89273566-89273566 on Build GRCh38
rs1053685062 Chr.15: 89273388-89273388 on Build GRCh38
rs547430360 Chr.15: 89273562-89273562 on Build GRCh38
rs1025430169 Chr.15: 89273638-89273638 on Build GRCh38
rs577519841 Chr.15: 89273312-89273312 on Build GRCh38
rs761155261 Chr.15: 89273505-89273505 on Build GRCh38
rs904774714 Chr.15: 89273321-89273327 on Build GRCh38
rs777158028 Chr.15: 89273520-89273520 on Build GRCh38
rs759939218 Chr.15: 89273511-89273513 on Build GRCh38
rs757236694 Chr.15: 89273396-89273396 on Build GRCh38
rs374523772 Chr.15: 89273364-89273364 on Build GRCh38
rs765280843 Chr.15: 89273481-89273481 on Build GRCh38

View Full Product Details