Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75231239-75231742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGA3
Entrez Gene ID
23163
Gene Name
golgi associated, gamma adaptin ear containing, ARF binding protein 3
Gene Aliases
-
UniGene
Hs.87726
Gene Chromosome Location
Chr.17: 75225464-75262393 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Gene Symbol
NUP85
Entrez Gene ID
79902
Gene Name
nucleoporin 85
Gene Aliases
FROUNT, Nup75
UniGene
Hs.362817
Gene Chromosome Location
Chr.17: 75205450-75235759 on Build GRCh38
Transcripts
XM_011524563.2 NM_001303276.1 NM_024844.4 XM_005257692.3 XM_006722094.1 XM_011525267.1 XM_017025108.1 XM_017025109.1 XM_017025110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4069509 c.613T>C p.F205L Chr.17: 75231362-75231362 on Build GRCh38
3692029 c.595G>A p.A199T Chr.17: 75231344-75231344 on Build GRCh38
3522064 c.1176C>T p.L392L Chr.17: 75231421-75231421 on Build GRCh38
6021666 c.694G>C p.E232Q Chr.17: 75231592-75231592 on Build GRCh38
3522065 c.672C>T p.L224L Chr.17: 75231421-75231421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746937292 Chr.17: 75231394-75231394 on Build GRCh38
rs751376070 Chr.17: 75231604-75231604 on Build GRCh38
rs4486943 Chr.17: 75231666-75231666 on Build GRCh38
rs773329765 Chr.17: 75231397-75231397 on Build GRCh38
rs376815821 Chr.17: 75231659-75231659 on Build GRCh38
rs1005460276 Chr.17: 75231631-75231631 on Build GRCh38
rs746979086 Chr.17: 75231455-75231455 on Build GRCh38
rs754689741 Chr.17: 75231609-75231609 on Build GRCh38
rs373954540 Chr.17: 75231292-75231292 on Build GRCh38
rs368696821 Chr.17: 75231444-75231444 on Build GRCh38
rs772863492 Chr.17: 75231669-75231669 on Build GRCh38
rs766106181 Chr.17: 75231704-75231704 on Build GRCh38
rs766593108 Chr.17: 75231403-75231403 on Build GRCh38
rs760887411 Chr.17: 75231541-75231541 on Build GRCh38
rs768851467 Chr.17: 75231654-75231654 on Build GRCh38
rs761130285 Chr.17: 75231650-75231650 on Build GRCh38
rs746145105 Chr.17: 75231642-75231642 on Build GRCh38
rs778646112 Chr.17: 75231337-75231337 on Build GRCh38
rs751954996 Chr.17: 75231354-75231354 on Build GRCh38
rs536741412 Chr.17: 75231531-75231531 on Build GRCh38
rs761461241 Chr.17: 75231557-75231557 on Build GRCh38
rs1007667 Chr.17: 75231687-75231687 on Build GRCh38
rs755626871 Chr.17: 75231312-75231312 on Build GRCh38
rs371897302 Chr.17: 75231398-75231398 on Build GRCh38
rs374077496 Chr.17: 75231383-75231383 on Build GRCh38
rs917027828 Chr.17: 75231453-75231453 on Build GRCh38
rs373981140 Chr.17: 75231688-75231688 on Build GRCh38
rs373359585 Chr.17: 75231560-75231560 on Build GRCh38
rs528938704 Chr.17: 75231277-75231277 on Build GRCh38
rs767940065 Chr.17: 75231303-75231303 on Build GRCh38
rs958398239 Chr.17: 75231670-75231670 on Build GRCh38
rs753410107 Chr.17: 75231443-75231443 on Build GRCh38
rs771037987 Chr.17: 75231629-75231629 on Build GRCh38
rs372369598 Chr.17: 75231472-75231472 on Build GRCh38
rs202174050 Chr.17: 75231471-75231471 on Build GRCh38
rs967064166 Chr.17: 75231689-75231689 on Build GRCh38
rs563260338 Chr.17: 75231307-75231307 on Build GRCh38
rs900986946 Chr.17: 75231278-75231278 on Build GRCh38
rs760120045 Chr.17: 75231307-75231307 on Build GRCh38
rs34266504 Chr.17: 75231568-75231569 on Build GRCh38
rs778959266 Chr.17: 75231635-75231635 on Build GRCh38
rs552086683 Chr.17: 75231436-75231436 on Build GRCh38
rs772349233 Chr.17: 75231644-75231644 on Build GRCh38
rs774518410 Chr.17: 75231409-75231409 on Build GRCh38
rs1006696914 Chr.17: 75231525-75231525 on Build GRCh38
rs769835709 Chr.17: 75231395-75231395 on Build GRCh38
rs116387801 Chr.17: 75231473-75231473 on Build GRCh38
rs375572023 Chr.17: 75231433-75231433 on Build GRCh38
rs768653087 Chr.17: 75231456-75231456 on Build GRCh38
rs773775576 Chr.17: 75231433-75231434 on Build GRCh38
rs781131003 Chr.17: 75231612-75231612 on Build GRCh38
rs201069857 Chr.17: 75231533-75231533 on Build GRCh38
rs750170347 Chr.17: 75231569-75231569 on Build GRCh38
rs750082457 Chr.17: 75231448-75231448 on Build GRCh38
rs758169835 Chr.17: 75231450-75231450 on Build GRCh38
rs749910869 Chr.17: 75231584-75231584 on Build GRCh38
rs977156396 Chr.17: 75231696-75231696 on Build GRCh38
rs759915823 Chr.17: 75231418-75231418 on Build GRCh38
rs533839113 Chr.17: 75231710-75231710 on Build GRCh38
rs867960400 Chr.17: 75231333-75231333 on Build GRCh38
rs759836494 Chr.17: 75231665-75231666 on Build GRCh38
rs368928199 Chr.17: 75231522-75231522 on Build GRCh38
rs752840719 Chr.17: 75231682-75231688 on Build GRCh38
rs199959682 Chr.17: 75231517-75231517 on Build GRCh38
rs775002839 Chr.17: 75231303-75231304 on Build GRCh38
rs984056921 Chr.17: 75231326-75231326 on Build GRCh38
rs758059695 Chr.17: 75231578-75231578 on Build GRCh38
rs764663392 Chr.17: 75231544-75231544 on Build GRCh38
rs748054022 Chr.17: 75231617-75231617 on Build GRCh38
rs868100581 Chr.17: 75231655-75231655 on Build GRCh38
rs766816960 Chr.17: 75231649-75231662 on Build GRCh38
rs754353175 Chr.17: 75231545-75231545 on Build GRCh38
rs370767968 Chr.17: 75231677-75231677 on Build GRCh38
rs763438088 Chr.17: 75231646-75231646 on Build GRCh38
rs775907528 Chr.17: 75231539-75231539 on Build GRCh38
rs775343744 Chr.17: 75231351-75231351 on Build GRCh38
rs936898451 Chr.17: 75231476-75231476 on Build GRCh38
rs901286852 Chr.17: 75231581-75231581 on Build GRCh38
rs763158301 Chr.17: 75231290-75231290 on Build GRCh38
rs543455821 Chr.17: 75231295-75231295 on Build GRCh38
rs368248317 Chr.17: 75231475-75231475 on Build GRCh38
rs745658584 Chr.17: 75231343-75231343 on Build GRCh38
rs767588202 Chr.17: 75231669-75231679 on Build GRCh38
rs577488194 Chr.17: 75231690-75231690 on Build GRCh38
rs1053976355 Chr.17: 75231510-75231510 on Build GRCh38
rs146201831 Chr.17: 75231666-75231683 on Build GRCh38
rs114077696 Chr.17: 75231315-75231315 on Build GRCh38
rs888293829 Chr.17: 75231622-75231622 on Build GRCh38
rs756953676 Chr.17: 75231334-75231334 on Build GRCh38
rs756199535 Chr.17: 75231686-75231686 on Build GRCh38
rs142625251 Chr.17: 75231590-75231590 on Build GRCh38
rs532095899 Chr.17: 75231415-75231415 on Build GRCh38
rs78010571 Chr.17: 75231434-75231434 on Build GRCh38
rs755270581 Chr.17: 75231293-75231293 on Build GRCh38
rs537685059 Chr.17: 75231671-75231671 on Build GRCh38
rs771947376 Chr.17: 75231344-75231344 on Build GRCh38
rs779897099 Chr.17: 75231454-75231454 on Build GRCh38
rs765737247 Chr.17: 75231432-75231449 on Build GRCh38
rs762304079 Chr.17: 75231678-75231678 on Build GRCh38
rs766789350 Chr.17: 75231291-75231291 on Build GRCh38

View Full Product Details