Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 7765105-7765646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4M
Entrez Gene ID
10332
Gene Name
C-type lectin domain family 4 member M
Gene Aliases
CD209L, CD299, DC-SIGN2, DC-SIGNR, DCSIGNR, HP10347, L-SIGN, LSIGN
UniGene
Hs.421437
Gene Chromosome Location
Chr.19: 7763052-7770559 on Build GRCh38
Transcripts
NM_001144904.1 NM_001144905.1 NM_001144906.1 NM_001144907.1 NM_001144908.1 NM_001144909.1 NM_001144910.1 NM_001144911.1 NM_014257.4 NR_026707.1 NR_026708.1 NR_026709.1 XM_006722611.2 XM_006722612.3 XM_006722613.3 XM_006722614.3 XM_006722615.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1248429 c.215-3C>T p.? Chr.19: 7765635-7765635 on Build GRCh38
3893919 c.171C>T p.S57S Chr.19: 7765225-7765225 on Build GRCh38
713875 c.148G>C p.A50P Chr.19: 7765202-7765202 on Build GRCh38
5583389 c.168C>T p.L56L Chr.19: 7765222-7765222 on Build GRCh38
6873519 c.166G>A p.V56I Chr.19: 7765244-7765244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888141250 Chr.19: 7765443-7765444 on Build GRCh38
rs376211903 Chr.19: 7765624-7765624 on Build GRCh38
rs760325881 Chr.19: 7765371-7765371 on Build GRCh38
rs750582492 Chr.19: 7765225-7765227 on Build GRCh38
rs760344022 Chr.19: 7765269-7765269 on Build GRCh38
rs370560578 Chr.19: 7765202-7765202 on Build GRCh38
rs999398089 Chr.19: 7765529-7765529 on Build GRCh38
rs186047913 Chr.19: 7765458-7765458 on Build GRCh38
rs750326441 Chr.19: 7765305-7765305 on Build GRCh38
rs768828883 Chr.19: 7765213-7765213 on Build GRCh38
rs752772572 Chr.19: 7765420-7765420 on Build GRCh38
rs767009217 Chr.19: 7765252-7765252 on Build GRCh38
rs777641342 Chr.19: 7765199-7765199 on Build GRCh38
rs755858867 Chr.19: 7765197-7765197 on Build GRCh38
rs988057265 Chr.19: 7765317-7765317 on Build GRCh38
rs921081418 Chr.19: 7765426-7765426 on Build GRCh38
rs773048909 Chr.19: 7765596-7765596 on Build GRCh38
rs1826308 Chr.19: 7765429-7765429 on Build GRCh38
rs377116159 Chr.19: 7765166-7765166 on Build GRCh38
rs199688006 Chr.19: 7765154-7765154 on Build GRCh38
rs751022549 Chr.19: 7765155-7765155 on Build GRCh38
rs767050381 Chr.19: 7765167-7765167 on Build GRCh38
rs748331766 Chr.19: 7765230-7765230 on Build GRCh38
rs778939763 Chr.19: 7765520-7765520 on Build GRCh38
rs117140487 Chr.19: 7765303-7765303 on Build GRCh38
rs1012384541 Chr.19: 7765465-7765465 on Build GRCh38
rs763807427 Chr.19: 7765377-7765377 on Build GRCh38
rs772626777 Chr.19: 7765582-7765582 on Build GRCh38
rs749645849 Chr.19: 7765310-7765310 on Build GRCh38
rs1047755286 Chr.19: 7765389-7765389 on Build GRCh38
rs771426798 Chr.19: 7765239-7765239 on Build GRCh38
rs1826306 Chr.19: 7765564-7765564 on Build GRCh38
rs267605766 Chr.19: 7765228-7765228 on Build GRCh38
rs749624448 Chr.19: 7765336-7765336 on Build GRCh38
rs201606704 Chr.19: 7765257-7765257 on Build GRCh38
rs894825095 Chr.19: 7765463-7765463 on Build GRCh38
rs994804988 Chr.19: 7765144-7765144 on Build GRCh38
rs776951272 Chr.19: 7765143-7765143 on Build GRCh38
rs535347455 Chr.19: 7765275-7765275 on Build GRCh38
rs930189127 Chr.19: 7765375-7765375 on Build GRCh38
rs555004163 Chr.19: 7765234-7765234 on Build GRCh38
rs748242020 Chr.19: 7765325-7765325 on Build GRCh38
rs764089786 Chr.19: 7765620-7765620 on Build GRCh38
rs112951388 Chr.19: 7765536-7765536 on Build GRCh38
rs368381502 Chr.19: 7765595-7765595 on Build GRCh38
rs942761278 Chr.19: 7765321-7765321 on Build GRCh38
rs765042577 Chr.19: 7765396-7765396 on Build GRCh38
rs766312087 Chr.19: 7765600-7765600 on Build GRCh38
rs866432975 Chr.19: 7765223-7765223 on Build GRCh38
rs770009726 Chr.19: 7765231-7765231 on Build GRCh38
rs112431723 Chr.19: 7765587-7765587 on Build GRCh38
rs763369162 Chr.19: 7765238-7765238 on Build GRCh38
rs761579304 Chr.19: 7765390-7765390 on Build GRCh38
rs372985078 Chr.19: 7765585-7765585 on Build GRCh38
rs781267858 Chr.19: 7765322-7765322 on Build GRCh38
rs1013822695 Chr.19: 7765497-7765497 on Build GRCh38
rs775124977 Chr.19: 7765361-7765361 on Build GRCh38
rs776471982 Chr.19: 7765385-7765385 on Build GRCh38
rs752150138 Chr.19: 7765268-7765268 on Build GRCh38
rs756317522 Chr.19: 7765330-7765330 on Build GRCh38
rs777953243 Chr.19: 7765567-7765567 on Build GRCh38
rs539558959 Chr.19: 7765201-7765201 on Build GRCh38
rs747028313 Chr.19: 7765207-7765207 on Build GRCh38
rs761463467 Chr.19: 7765594-7765594 on Build GRCh38
rs368204062 Chr.19: 7765376-7765376 on Build GRCh38
rs759978815 Chr.19: 7765250-7765250 on Build GRCh38
rs373961931 Chr.19: 7765150-7765150 on Build GRCh38
rs570966645 Chr.19: 7765193-7765193 on Build GRCh38
rs978394605 Chr.19: 7765253-7765253 on Build GRCh38
rs986694801 Chr.19: 7765537-7765537 on Build GRCh38
rs748004723 Chr.19: 7765547-7765547 on Build GRCh38
rs967924634 Chr.19: 7765245-7765245 on Build GRCh38
rs756470864 Chr.19: 7765224-7765225 on Build GRCh38
rs573568167 Chr.19: 7765614-7765614 on Build GRCh38
rs1040869089 Chr.19: 7765421-7765421 on Build GRCh38
rs780236752 Chr.19: 7765262-7765262 on Build GRCh38
rs751551142 Chr.19: 7765606-7765606 on Build GRCh38
rs757485066 Chr.19: 7765625-7765625 on Build GRCh38
rs772467195 Chr.19: 7765353-7765353 on Build GRCh38
rs1003129408 Chr.19: 7765441-7765441 on Build GRCh38
rs181106015 Chr.19: 7765443-7765443 on Build GRCh38
rs898678593 Chr.19: 7765130-7765130 on Build GRCh38
rs1826307 Chr.19: 7765514-7765514 on Build GRCh38
rs749467138 Chr.19: 7765569-7765569 on Build GRCh38
rs768755596 Chr.19: 7765134-7765134 on Build GRCh38
rs913875310 Chr.19: 7765570-7765570 on Build GRCh38
rs774966793 Chr.19: 7765244-7765244 on Build GRCh38
rs112788979 Chr.19: 7765436-7765436 on Build GRCh38
rs771380457 Chr.19: 7765344-7765344 on Build GRCh38
rs778693946 Chr.19: 7765304-7765304 on Build GRCh38
rs571702383 Chr.19: 7765348-7765348 on Build GRCh38
rs753680993 Chr.19: 7765289-7765289 on Build GRCh38
rs541060001 Chr.19: 7765400-7765400 on Build GRCh38
rs766288287 Chr.19: 7765412-7765412 on Build GRCh38
rs762726617 Chr.19: 7765598-7765598 on Build GRCh38
rs750191741 Chr.19: 7765398-7765398 on Build GRCh38
rs756229634 Chr.19: 7765422-7765422 on Build GRCh38
rs754935461 Chr.19: 7765612-7765612 on Build GRCh38
rs542520111 Chr.19: 7765572-7765572 on Build GRCh38
rs1027082599 Chr.19: 7765156-7765156 on Build GRCh38
rs923885692 Chr.19: 7765254-7765254 on Build GRCh38
rs746316121 Chr.19: 7765349-7765349 on Build GRCh38
rs781154601 Chr.19: 7765222-7765222 on Build GRCh38
rs758300329 Chr.19: 7765311-7765311 on Build GRCh38
rs969549771 Chr.19: 7765517-7765517 on Build GRCh38
rs766887426 Chr.19: 7765151-7765151 on Build GRCh38
rs113778282 Chr.19: 7765145-7765145 on Build GRCh38
rs374998358 Chr.19: 7765591-7765591 on Build GRCh38
rs1030797003 Chr.19: 7765531-7765531 on Build GRCh38
rs200921491 Chr.19: 7765169-7765169 on Build GRCh38

View Full Product Details