Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 69505636-69506134 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM155B
Entrez Gene ID
27112
Gene Name
family with sequence similarity 155 member B
Gene Aliases
CXorf63, TED, TMEM28, bB57D9.1
UniGene
Hs.87619
Gene Chromosome Location
Chr.X: 69505204-69532508 on Build GRCh38
Transcripts
NM_015686.2 XM_011530908.2 XM_011530909.2

SNP Detail

SNP ID Chromosome Location
rs148811454 Chr.X: 69506095-69506095 on Build GRCh38
rs374491730 Chr.X: 69505942-69505942 on Build GRCh38
rs780007252 Chr.X: 69505828-69505829 on Build GRCh38
rs141000146 Chr.X: 69505952-69505952 on Build GRCh38
rs766987707 Chr.X: 69505772-69505772 on Build GRCh38
rs754805211 Chr.X: 69505728-69505728 on Build GRCh38
rs756307961 Chr.X: 69505909-69505909 on Build GRCh38
rs201151491 Chr.X: 69505943-69505943 on Build GRCh38
rs772089289 Chr.X: 69505784-69505784 on Build GRCh38
rs753540595 Chr.X: 69505781-69505781 on Build GRCh38
rs749254240 Chr.X: 69505829-69505829 on Build GRCh38
rs867076028 Chr.X: 69505911-69505911 on Build GRCh38
rs201872871 Chr.X: 69505810-69505810 on Build GRCh38
rs758459917 Chr.X: 69505786-69505786 on Build GRCh38
rs748720224 Chr.X: 69505951-69505951 on Build GRCh38
rs749883388 Chr.X: 69505774-69505774 on Build GRCh38
rs773625287 Chr.X: 69505996-69505996 on Build GRCh38
rs759359643 Chr.X: 69505975-69505975 on Build GRCh38
rs1171942 Chr.X: 69505797-69505797 on Build GRCh38
rs770053557 Chr.X: 69505805-69505805 on Build GRCh38
rs767373488 Chr.X: 69505974-69505974 on Build GRCh38
rs775289057 Chr.X: 69505961-69505961 on Build GRCh38
rs774234254 Chr.X: 69506043-69506043 on Build GRCh38
rs763487166 Chr.X: 69505770-69505770 on Build GRCh38
rs769524017 Chr.X: 69505769-69505770 on Build GRCh38
rs749585287 Chr.X: 69505941-69505941 on Build GRCh38
rs775968689 Chr.X: 69505806-69505806 on Build GRCh38
rs752650528 Chr.X: 69505785-69505785 on Build GRCh38
rs769078970 Chr.X: 69505685-69505685 on Build GRCh38
rs149821108 Chr.X: 69505958-69505958 on Build GRCh38
rs777876500 Chr.X: 69505791-69505791 on Build GRCh38
rs146856746 Chr.X: 69505973-69505973 on Build GRCh38
rs752396362 Chr.X: 69506105-69506105 on Build GRCh38
rs772582182 Chr.X: 69505995-69505995 on Build GRCh38
rs917671957 Chr.X: 69506026-69506026 on Build GRCh38
rs760206282 Chr.X: 69505775-69505775 on Build GRCh38
rs759755485 Chr.X: 69505768-69505768 on Build GRCh38
rs867468316 Chr.X: 69506075-69506075 on Build GRCh38
rs760118397 Chr.X: 69505819-69505819 on Build GRCh38
rs780282770 Chr.X: 69505939-69505939 on Build GRCh38
rs758991107 Chr.X: 69505957-69505957 on Build GRCh38
rs755330269 Chr.X: 69505993-69505993 on Build GRCh38
rs759273141 Chr.X: 69505856-69505856 on Build GRCh38
rs753254642 Chr.X: 69505655-69505655 on Build GRCh38
rs769444908 Chr.X: 69506016-69506016 on Build GRCh38
rs142803076 Chr.X: 69505813-69505813 on Build GRCh38
rs746162821 Chr.X: 69505801-69505801 on Build GRCh38
rs764041624 Chr.X: 69505970-69505970 on Build GRCh38
rs905115266 Chr.X: 69505867-69505867 on Build GRCh38
rs764919201 Chr.X: 69505864-69505864 on Build GRCh38
rs754714563 Chr.X: 69505783-69505783 on Build GRCh38
rs765971562 Chr.X: 69505826-69505826 on Build GRCh38
rs750042647 Chr.X: 69505677-69505677 on Build GRCh38
rs775270963 Chr.X: 69506023-69506023 on Build GRCh38
rs774841719 Chr.X: 69505814-69505814 on Build GRCh38
rs138484665 Chr.X: 69506087-69506087 on Build GRCh38
rs144542226 Chr.X: 69505753-69505753 on Build GRCh38
rs140715552 Chr.X: 69505981-69505981 on Build GRCh38
rs763953914 Chr.X: 69506039-69506039 on Build GRCh38
rs771231753 Chr.X: 69505754-69505754 on Build GRCh38
rs770211531 Chr.X: 69505769-69505769 on Build GRCh38
rs776253229 Chr.X: 69505955-69505955 on Build GRCh38
rs753535740 Chr.X: 69505726-69505726 on Build GRCh38
rs1045301116 Chr.X: 69506104-69506104 on Build GRCh38
rs781216420 Chr.X: 69505798-69505798 on Build GRCh38
rs772274449 Chr.X: 69505737-69505737 on Build GRCh38
rs778657309 Chr.X: 69505730-69505730 on Build GRCh38
rs761541566 Chr.X: 69506065-69506065 on Build GRCh38
rs756149484 Chr.X: 69505978-69505978 on Build GRCh38
rs1005425404 Chr.X: 69506017-69506017 on Build GRCh38
rs1052211839 Chr.X: 69505773-69505773 on Build GRCh38
rs758302010 Chr.X: 69505880-69505880 on Build GRCh38
rs199640674 Chr.X: 69505924-69505924 on Build GRCh38
rs200504139 Chr.X: 69505787-69505787 on Build GRCh38
rs767075576 Chr.X: 69505667-69505667 on Build GRCh38
rs145917665 Chr.X: 69506080-69506080 on Build GRCh38
rs147875150 Chr.X: 69505758-69505758 on Build GRCh38
rs753427791 Chr.X: 69505836-69505836 on Build GRCh38
rs755446501 Chr.X: 69505719-69505719 on Build GRCh38
rs371709320 Chr.X: 69506028-69506028 on Build GRCh38
rs201772031 Chr.X: 69505997-69505997 on Build GRCh38
rs1013855143 Chr.X: 69505792-69505792 on Build GRCh38
rs766511300 Chr.X: 69506099-69506099 on Build GRCh38
rs756703590 Chr.X: 69505656-69505656 on Build GRCh38
rs200299714 Chr.X: 69505994-69505994 on Build GRCh38
rs774742857 Chr.X: 69505708-69505708 on Build GRCh38
rs752493079 Chr.X: 69505866-69505866 on Build GRCh38
rs748174015 Chr.X: 69505735-69505735 on Build GRCh38
rs371432784 Chr.X: 69505894-69505894 on Build GRCh38
rs995425500 Chr.X: 69505870-69505870 on Build GRCh38
rs1044959512 Chr.X: 69505808-69505808 on Build GRCh38
rs778039174 Chr.X: 69505747-69505747 on Build GRCh38
rs1055959173 Chr.X: 69505953-69505953 on Build GRCh38
rs751542420 Chr.X: 69505972-69505972 on Build GRCh38
rs368057024 Chr.X: 69505963-69505963 on Build GRCh38
rs935067122 Chr.X: 69505764-69505764 on Build GRCh38
rs190801973 Chr.X: 69505987-69505987 on Build GRCh38
rs764059847 Chr.X: 69505885-69505885 on Build GRCh38
rs151056099 Chr.X: 69505904-69505904 on Build GRCh38

View Full Product Details