Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70656229-70656665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JCHAIN
Entrez Gene ID
3512
Gene Name
joining chain of multimeric IgA and IgM
Gene Aliases
IGCJ, IGJ, JCH
UniGene
Hs.643431
Gene Chromosome Location
Chr.4: 70655541-70676901 on Build GRCh38
Transcripts
NM_144646.3 XM_006714211.2 XM_006714212.2 XM_011531925.1 XM_011531926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4125762 c.393C>A p.C131* Chr.4: 70656416-70656416 on Build GRCh38
5768821 c.381C>T p.D127D Chr.4: 70656428-70656428 on Build GRCh38
4168481 c.427G>T p.E143* Chr.4: 70656382-70656382 on Build GRCh38
3975011 c.443A>G p.E148G Chr.4: 70656366-70656366 on Build GRCh38
1633759 c.270-7T>C p.? Chr.4: 70656546-70656546 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746559683 Chr.4: 70656311-70656311 on Build GRCh38
rs1038756692 Chr.4: 70656629-70656629 on Build GRCh38
rs202169418 Chr.4: 70656371-70656371 on Build GRCh38
rs748096958 Chr.4: 70656523-70656523 on Build GRCh38
rs1016259668 Chr.4: 70656256-70656256 on Build GRCh38
rs373594444 Chr.4: 70656291-70656291 on Build GRCh38
rs774401488 Chr.4: 70656418-70656418 on Build GRCh38
rs371133971 Chr.4: 70656288-70656288 on Build GRCh38
rs749150009 Chr.4: 70656509-70656509 on Build GRCh38
rs752044808 Chr.4: 70656483-70656483 on Build GRCh38
rs969875708 Chr.4: 70656411-70656411 on Build GRCh38
rs754448731 Chr.4: 70656607-70656607 on Build GRCh38
rs762320270 Chr.4: 70656568-70656568 on Build GRCh38
rs758728357 Chr.4: 70656462-70656462 on Build GRCh38
rs755812286 Chr.4: 70656497-70656497 on Build GRCh38
rs970794545 Chr.4: 70656264-70656264 on Build GRCh38
rs377331102 Chr.4: 70656501-70656501 on Build GRCh38
rs759488146 Chr.4: 70656534-70656534 on Build GRCh38
rs770092875 Chr.4: 70656539-70656539 on Build GRCh38
rs772071970 Chr.4: 70656529-70656529 on Build GRCh38
rs776788987 Chr.4: 70656531-70656531 on Build GRCh38
rs758959759 Chr.4: 70656289-70656289 on Build GRCh38
rs201356678 Chr.4: 70656381-70656381 on Build GRCh38
rs765894756 Chr.4: 70656514-70656514 on Build GRCh38
rs575017166 Chr.4: 70656385-70656385 on Build GRCh38
rs926494462 Chr.4: 70656366-70656366 on Build GRCh38
rs535962036 Chr.4: 70656395-70656395 on Build GRCh38
rs554652827 Chr.4: 70656419-70656419 on Build GRCh38
rs981687958 Chr.4: 70656416-70656416 on Build GRCh38
rs374362054 Chr.4: 70656287-70656287 on Build GRCh38
rs764579156 Chr.4: 70656252-70656252 on Build GRCh38
rs147967696 Chr.4: 70656391-70656391 on Build GRCh38
rs145025435 Chr.4: 70656386-70656386 on Build GRCh38
rs765322118 Chr.4: 70656445-70656445 on Build GRCh38
rs982093158 Chr.4: 70656266-70656266 on Build GRCh38
rs185001891 Chr.4: 70656487-70656487 on Build GRCh38
rs775472343 Chr.4: 70656566-70656566 on Build GRCh38
rs754357230 Chr.4: 70656344-70656344 on Build GRCh38
rs748984740 Chr.4: 70656394-70656394 on Build GRCh38
rs751854443 Chr.4: 70656567-70656567 on Build GRCh38
rs12331718 Chr.4: 70656254-70656254 on Build GRCh38
rs756823556 Chr.4: 70656484-70656484 on Build GRCh38
rs763362976 Chr.4: 70656551-70656551 on Build GRCh38
rs572710543 Chr.4: 70656471-70656471 on Build GRCh38
rs267600240 Chr.4: 70656526-70656526 on Build GRCh38
rs577191197 Chr.4: 70656545-70656545 on Build GRCh38
rs370641440 Chr.4: 70656431-70656431 on Build GRCh38
rs79589978 Chr.4: 70656566-70656566 on Build GRCh38
rs376801317 Chr.4: 70656308-70656308 on Build GRCh38
rs1042180308 Chr.4: 70656463-70656463 on Build GRCh38
rs759812216 Chr.4: 70656432-70656432 on Build GRCh38
rs766553791 Chr.4: 70656421-70656421 on Build GRCh38
rs947497900 Chr.4: 70656410-70656410 on Build GRCh38
rs374027021 Chr.4: 70656495-70656495 on Build GRCh38
rs771866781 Chr.4: 70656301-70656302 on Build GRCh38
rs939784403 Chr.4: 70656642-70656642 on Build GRCh38
rs753129856 Chr.4: 70656456-70656456 on Build GRCh38
rs756734000 Chr.4: 70656367-70656367 on Build GRCh38
rs558865433 Chr.4: 70656517-70656517 on Build GRCh38
rs78599402 Chr.4: 70656594-70656594 on Build GRCh38
rs963414124 Chr.4: 70656323-70656323 on Build GRCh38

View Full Product Details