Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 47031727-47032204 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GABRB1
Entrez Gene ID
2560
Gene Name
gamma-aminobutyric acid type A receptor beta1 subunit
Gene Aliases
-
UniGene
Hs.254117
Gene Chromosome Location
Chr.4: 47031278-47438409 on Build GRCh38
Transcripts
NM_000812.3 XM_017007986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6668574 c.172G>A p.G58R Chr.4: 47032005-47032005 on Build GRCh38
5496491 c.81-1G>T p.? Chr.4: 47031913-47031913 on Build GRCh38
5709253 c.137A>G p.K46R Chr.4: 47031970-47031970 on Build GRCh38
6512175 c.165C>A p.D55E Chr.4: 47031998-47031998 on Build GRCh38
481300 c.97A>C p.N33H Chr.4: 47031930-47031930 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45460902 Chr.4: 47032058-47032058 on Build GRCh38
rs369105252 Chr.4: 47032086-47032087 on Build GRCh38
rs11735112 Chr.4: 47031755-47031755 on Build GRCh38
rs149501911 Chr.4: 47032138-47032139 on Build GRCh38
rs745790497 Chr.4: 47031951-47031951 on Build GRCh38
rs376403161 Chr.4: 47032085-47032087 on Build GRCh38
rs763791394 Chr.4: 47031772-47031772 on Build GRCh38
rs766473031 Chr.4: 47031751-47031758 on Build GRCh38
rs78252491 Chr.4: 47031775-47031775 on Build GRCh38
rs71654859 Chr.4: 47032073-47032074 on Build GRCh38
rs948286541 Chr.4: 47031852-47031852 on Build GRCh38
rs758592647 Chr.4: 47032045-47032045 on Build GRCh38
rs553300116 Chr.4: 47031950-47031950 on Build GRCh38
rs747856985 Chr.4: 47031867-47031868 on Build GRCh38
rs200686643 Chr.4: 47031751-47031756 on Build GRCh38
rs538091356 Chr.4: 47032078-47032079 on Build GRCh38
rs780532729 Chr.4: 47031878-47031878 on Build GRCh38
rs952559323 Chr.4: 47032106-47032106 on Build GRCh38
rs776645242 Chr.4: 47031759-47031759 on Build GRCh38
rs1007236807 Chr.4: 47031961-47031961 on Build GRCh38
rs767850083 Chr.4: 47031873-47031873 on Build GRCh38
rs754189722 Chr.4: 47032024-47032024 on Build GRCh38
rs755247546 Chr.4: 47031904-47031904 on Build GRCh38
rs889731175 Chr.4: 47031926-47031926 on Build GRCh38
rs112951462 Chr.4: 47032130-47032130 on Build GRCh38
rs758413999 Chr.4: 47031775-47031776 on Build GRCh38
rs774566867 Chr.4: 47031751-47031752 on Build GRCh38
rs754419160 Chr.4: 47031776-47031777 on Build GRCh38
rs571541868 Chr.4: 47032098-47032098 on Build GRCh38
rs765142853 Chr.4: 47031756-47031761 on Build GRCh38
rs878915564 Chr.4: 47032064-47032064 on Build GRCh38
rs865820336 Chr.4: 47032131-47032131 on Build GRCh38
rs767061726 Chr.4: 47032023-47032023 on Build GRCh38
rs768226870 Chr.4: 47032009-47032009 on Build GRCh38
rs28564672 Chr.4: 47031774-47031774 on Build GRCh38
rs72416056 Chr.4: 47031754-47031755 on Build GRCh38
rs774062328 Chr.4: 47032010-47032010 on Build GRCh38
rs67779259 Chr.4: 47032087-47032087 on Build GRCh38
rs779730898 Chr.4: 47031753-47031754 on Build GRCh38
rs769774963 Chr.4: 47031958-47031958 on Build GRCh38
rs921569079 Chr.4: 47032180-47032180 on Build GRCh38
rs73247629 Chr.4: 47032128-47032128 on Build GRCh38
rs796838260 Chr.4: 47031755-47031756 on Build GRCh38
rs756039260 Chr.4: 47032052-47032052 on Build GRCh38
rs901017404 Chr.4: 47032016-47032016 on Build GRCh38
rs546534250 Chr.4: 47032094-47032094 on Build GRCh38
rs754052722 Chr.4: 47031756-47031757 on Build GRCh38
rs369463882 Chr.4: 47031875-47031875 on Build GRCh38
rs772220562 Chr.4: 47031924-47031924 on Build GRCh38
rs796145608 Chr.4: 47032128-47032129 on Build GRCh38
rs574120897 Chr.4: 47032162-47032162 on Build GRCh38
rs377115389 Chr.4: 47031765-47031765 on Build GRCh38
rs932858940 Chr.4: 47031783-47031783 on Build GRCh38
rs761387895 Chr.4: 47031778-47031778 on Build GRCh38
rs1057075830 Chr.4: 47031974-47031974 on Build GRCh38
rs954432883 Chr.4: 47032185-47032185 on Build GRCh38
rs374503142 Chr.4: 47032018-47032018 on Build GRCh38
rs142160221 Chr.4: 47031996-47031996 on Build GRCh38
rs778057446 Chr.4: 47031867-47031867 on Build GRCh38
rs1019459592 Chr.4: 47031966-47031966 on Build GRCh38
rs916868016 Chr.4: 47031844-47031844 on Build GRCh38
rs200020355 Chr.4: 47031760-47031760 on Build GRCh38
rs371613712 Chr.4: 47031776-47031776 on Build GRCh38
rs779283546 Chr.4: 47031915-47031915 on Build GRCh38
rs375911533 Chr.4: 47032127-47032128 on Build GRCh38
rs750157260 Chr.4: 47031774-47031775 on Build GRCh38
rs796353971 Chr.4: 47032086-47032087 on Build GRCh38
rs771540640 Chr.4: 47031750-47031751 on Build GRCh38
rs754314249 Chr.4: 47031880-47031880 on Build GRCh38
rs567442181 Chr.4: 47031752-47031755 on Build GRCh38
rs113036607 Chr.4: 47031772-47031772 on Build GRCh38
rs72109821 Chr.4: 47032128-47032129 on Build GRCh38
rs71611989 Chr.4: 47031787-47031787 on Build GRCh38
rs749940839 Chr.4: 47032017-47032017 on Build GRCh38
rs757105864 Chr.4: 47031753-47031758 on Build GRCh38
rs765507674 Chr.4: 47032039-47032039 on Build GRCh38
rs928131362 Chr.4: 47031857-47031857 on Build GRCh38
rs71193897 Chr.4: 47032161-47032162 on Build GRCh38
rs894366963 Chr.4: 47031995-47031995 on Build GRCh38
rs747118464 Chr.4: 47032047-47032047 on Build GRCh38
rs796531806 Chr.4: 47031754-47031755 on Build GRCh38
rs769612012 Chr.4: 47031763-47031763 on Build GRCh38
rs866583531 Chr.4: 47031756-47031756 on Build GRCh38
rs6847977 Chr.4: 47032087-47032087 on Build GRCh38
rs757535545 Chr.4: 47031756-47031759 on Build GRCh38
rs761179801 Chr.4: 47032011-47032011 on Build GRCh38
rs1053878198 Chr.4: 47031785-47031785 on Build GRCh38
rs796391349 Chr.4: 47031775-47031776 on Build GRCh38
rs769433153 Chr.4: 47032027-47032028 on Build GRCh38
rs946771997 Chr.4: 47031906-47031906 on Build GRCh38
rs756392686 Chr.4: 47031877-47031877 on Build GRCh38
rs982363006 Chr.4: 47031856-47031856 on Build GRCh38
rs767084680 Chr.4: 47032012-47032012 on Build GRCh38
rs148675961 Chr.4: 47031919-47031919 on Build GRCh38
rs372458693 Chr.4: 47032033-47032033 on Build GRCh38
rs775250287 Chr.4: 47031969-47031969 on Build GRCh38
rs879085231 Chr.4: 47032066-47032066 on Build GRCh38
rs28475494 Chr.4: 47032129-47032129 on Build GRCh38
rs965046844 Chr.4: 47032148-47032148 on Build GRCh38
rs33980898 Chr.4: 47031755-47031756 on Build GRCh38
rs145810264 Chr.4: 47031770-47031770 on Build GRCh38
rs779809527 Chr.4: 47031775-47031775 on Build GRCh38
rs752999029 Chr.4: 47032041-47032041 on Build GRCh38
rs755408640 Chr.4: 47032028-47032028 on Build GRCh38
rs771228324 Chr.4: 47031751-47031760 on Build GRCh38
rs869194929 Chr.4: 47032083-47032087 on Build GRCh38
rs573910395 Chr.4: 47032170-47032170 on Build GRCh38
rs772121094 Chr.4: 47031782-47031782 on Build GRCh38
rs555934812 Chr.4: 47032168-47032168 on Build GRCh38
rs77472678 Chr.4: 47032119-47032119 on Build GRCh38
rs534624395 Chr.4: 47032089-47032089 on Build GRCh38

View Full Product Details