Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33988647-33989240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMACR
Entrez Gene ID
23600
Gene Name
alpha-methylacyl-CoA racemase
Gene Aliases
AMACRD, CBAS4, RACE, RM
UniGene
Hs.508343
Gene Chromosome Location
Chr.5: 33986986-34008115 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Gene Symbol
C1QTNF3-AMACR
Entrez Gene ID
100534612
Gene Name
C1QTNF3-AMACR readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.171929
Gene Chromosome Location
Chr.5: 33986986-34124528 on Build GRCh38
Transcripts
NM_001167595.1 NM_014324.5 NM_203382.2 NR_037951.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5934937 c.1087G>A p.E363K Chr.5: 33989155-33989155 on Build GRCh38
1067410 c.1084G>A p.E362K Chr.5: 33989158-33989158 on Build GRCh38
1067411 c.1042G>T p.E348* Chr.5: 33989200-33989200 on Build GRCh38
334652 c.1132G>A p.V378I Chr.5: 33989110-33989110 on Build GRCh38
5950437 c.1081C>G p.R361G Chr.5: 33989161-33989161 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537506468 Chr.5: 33988818-33988818 on Build GRCh38
rs989523253 Chr.5: 33988889-33988889 on Build GRCh38
rs1012952543 Chr.5: 33988800-33988800 on Build GRCh38
rs779669491 Chr.5: 33989122-33989122 on Build GRCh38
rs774406448 Chr.5: 33989158-33989158 on Build GRCh38
rs749703699 Chr.5: 33989217-33989217 on Build GRCh38
rs971158950 Chr.5: 33988856-33988856 on Build GRCh38
rs181895748 Chr.5: 33988705-33988705 on Build GRCh38
rs956784061 Chr.5: 33988883-33988883 on Build GRCh38
rs76132071 Chr.5: 33988788-33988788 on Build GRCh38
rs911613039 Chr.5: 33988921-33988921 on Build GRCh38
rs376816220 Chr.5: 33989070-33989070 on Build GRCh38
rs778402751 Chr.5: 33989211-33989211 on Build GRCh38
rs200270822 Chr.5: 33989119-33989119 on Build GRCh38
rs751045891 Chr.5: 33989202-33989202 on Build GRCh38
rs764977396 Chr.5: 33989193-33989193 on Build GRCh38
rs756743141 Chr.5: 33989205-33989205 on Build GRCh38
rs752566971 Chr.5: 33988720-33988720 on Build GRCh38
rs188193261 Chr.5: 33988948-33988948 on Build GRCh38
rs778457164 Chr.5: 33989038-33989038 on Build GRCh38
rs1021068621 Chr.5: 33988796-33988796 on Build GRCh38
rs553247803 Chr.5: 33988932-33988932 on Build GRCh38
rs541432932 Chr.5: 33988975-33988975 on Build GRCh38
rs748561928 Chr.5: 33989055-33989055 on Build GRCh38
rs770367046 Chr.5: 33989140-33989140 on Build GRCh38
rs756997856 Chr.5: 33989101-33989101 on Build GRCh38
rs759514531 Chr.5: 33989075-33989075 on Build GRCh38
rs764011770 Chr.5: 33989173-33989173 on Build GRCh38
rs566109692 Chr.5: 33988676-33988676 on Build GRCh38
rs753686325 Chr.5: 33989176-33989176 on Build GRCh38
rs745361517 Chr.5: 33989045-33989045 on Build GRCh38
rs561675407 Chr.5: 33989159-33989159 on Build GRCh38
rs748686701 Chr.5: 33989135-33989135 on Build GRCh38
rs770311997 Chr.5: 33989059-33989059 on Build GRCh38
rs749602984 Chr.5: 33989148-33989148 on Build GRCh38
rs150765689 Chr.5: 33989113-33989113 on Build GRCh38
rs763893617 Chr.5: 33989099-33989099 on Build GRCh38
rs773780297 Chr.5: 33989145-33989145 on Build GRCh38
rs750073270 Chr.5: 33989194-33989194 on Build GRCh38
rs771235608 Chr.5: 33989149-33989149 on Build GRCh38
rs374668256 Chr.5: 33989160-33989160 on Build GRCh38
rs777252887 Chr.5: 33989049-33989049 on Build GRCh38
rs763792102 Chr.5: 33989044-33989044 on Build GRCh38
rs986288438 Chr.5: 33988920-33988920 on Build GRCh38
rs752666056 Chr.5: 33989080-33989080 on Build GRCh38
rs185441763 Chr.5: 33988903-33988903 on Build GRCh38
rs138052540 Chr.5: 33989172-33989172 on Build GRCh38
rs114145560 Chr.5: 33988756-33988756 on Build GRCh38
rs776149094 Chr.5: 33989088-33989088 on Build GRCh38
rs774547866 Chr.5: 33989073-33989073 on Build GRCh38
rs769261872 Chr.5: 33989048-33989048 on Build GRCh38
rs947231666 Chr.5: 33988967-33988967 on Build GRCh38
rs6863560 Chr.5: 33988866-33988866 on Build GRCh38
rs767532180 Chr.5: 33989079-33989079 on Build GRCh38
rs557294515 Chr.5: 33988860-33988860 on Build GRCh38
rs763947586 Chr.5: 33988760-33988760 on Build GRCh38
rs1023882219 Chr.5: 33988843-33988843 on Build GRCh38
rs761488582 Chr.5: 33989191-33989191 on Build GRCh38
rs543993257 Chr.5: 33988757-33988757 on Build GRCh38
rs546903502 Chr.5: 33988670-33988670 on Build GRCh38
rs1035086458 Chr.5: 33988732-33988732 on Build GRCh38
rs779722935 Chr.5: 33989197-33989197 on Build GRCh38
rs755998638 Chr.5: 33989094-33989094 on Build GRCh38
rs778414134 Chr.5: 33989112-33989112 on Build GRCh38
rs773661915 Chr.5: 33989064-33989064 on Build GRCh38
rs763280369 Chr.5: 33989065-33989065 on Build GRCh38
rs775494356 Chr.5: 33989165-33989165 on Build GRCh38

View Full Product Details