Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47680194-47680720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRF2
Entrez Gene ID
222611
Gene Name
adhesion G protein-coupled receptor F2
Gene Aliases
GPR111, PGR20, hGPCR35
UniGene
Hs.724860
Gene Chromosome Location
Chr.6: 47656590-47697797 on Build GRCh38
Transcripts
NM_153839.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6583722 c.699G>A p.K233K Chr.6: 47680298-47680298 on Build GRCh38
7369894 c.494A>G p.K165R Chr.6: 47680297-47680297 on Build GRCh38
5870849 c.688G>A p.D230N Chr.6: 47680287-47680287 on Build GRCh38
7369898 c.437T>C p.I146T Chr.6: 47680240-47680240 on Build GRCh38
6899990 c.660C>T p.N220N Chr.6: 47680259-47680259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983637646 Chr.6: 47680697-47680697 on Build GRCh38
rs371953014 Chr.6: 47680314-47680314 on Build GRCh38
rs974459946 Chr.6: 47680380-47680380 on Build GRCh38
rs781542542 Chr.6: 47680318-47680318 on Build GRCh38
rs867070954 Chr.6: 47680410-47680410 on Build GRCh38
rs920377534 Chr.6: 47680390-47680390 on Build GRCh38
rs115587849 Chr.6: 47680269-47680269 on Build GRCh38
rs752279135 Chr.6: 47680221-47680221 on Build GRCh38
rs535703880 Chr.6: 47680238-47680238 on Build GRCh38
rs568302060 Chr.6: 47680257-47680257 on Build GRCh38
rs997950195 Chr.6: 47680551-47680551 on Build GRCh38
rs371181710 Chr.6: 47680240-47680240 on Build GRCh38
rs778742677 Chr.6: 47680309-47680309 on Build GRCh38
rs563194149 Chr.6: 47680437-47680437 on Build GRCh38
rs983345463 Chr.6: 47680695-47680695 on Build GRCh38
rs565994868 Chr.6: 47680566-47680566 on Build GRCh38
rs1031422039 Chr.6: 47680289-47680289 on Build GRCh38
rs1000175783 Chr.6: 47680589-47680589 on Build GRCh38
rs1056810084 Chr.6: 47680500-47680500 on Build GRCh38
rs930400859 Chr.6: 47680438-47680438 on Build GRCh38
rs1009813244 Chr.6: 47680568-47680568 on Build GRCh38
rs747222612 Chr.6: 47680559-47680559 on Build GRCh38
rs895002234 Chr.6: 47680244-47680244 on Build GRCh38
rs115905625 Chr.6: 47680297-47680297 on Build GRCh38
rs1008459492 Chr.6: 47680591-47680591 on Build GRCh38
rs1018978742 Chr.6: 47680601-47680601 on Build GRCh38
rs1003891620 Chr.6: 47680221-47680221 on Build GRCh38
rs6941294 Chr.6: 47680581-47680581 on Build GRCh38
rs758382892 Chr.6: 47680471-47680471 on Build GRCh38
rs534042943 Chr.6: 47680281-47680281 on Build GRCh38
rs368504564 Chr.6: 47680261-47680261 on Build GRCh38
rs773653307 Chr.6: 47680332-47680332 on Build GRCh38
rs770269182 Chr.6: 47680329-47680329 on Build GRCh38
rs1039371159 Chr.6: 47680538-47680538 on Build GRCh38
rs748447603 Chr.6: 47680323-47680323 on Build GRCh38
rs574883399 Chr.6: 47680359-47680359 on Build GRCh38
rs1034330813 Chr.6: 47680625-47680625 on Build GRCh38
rs758085964 Chr.6: 47680582-47680582 on Build GRCh38
rs757541870 Chr.6: 47680350-47680350 on Build GRCh38
rs141145040 Chr.6: 47680259-47680259 on Build GRCh38
rs899478383 Chr.6: 47680541-47680541 on Build GRCh38
rs544233157 Chr.6: 47680381-47680381 on Build GRCh38
rs143648591 Chr.6: 47680470-47680470 on Build GRCh38
rs142915340 Chr.6: 47680694-47680695 on Build GRCh38
rs6918351 Chr.6: 47680683-47680683 on Build GRCh38
rs757069234 Chr.6: 47680236-47680236 on Build GRCh38
rs951615449 Chr.6: 47680685-47680685 on Build GRCh38
rs542369563 Chr.6: 47680565-47680565 on Build GRCh38
rs527355224 Chr.6: 47680643-47680643 on Build GRCh38
rs927410842 Chr.6: 47680366-47680366 on Build GRCh38
rs992848059 Chr.6: 47680411-47680411 on Build GRCh38
rs894976657 Chr.6: 47680517-47680517 on Build GRCh38
rs910120298 Chr.6: 47680495-47680495 on Build GRCh38
rs573246198 Chr.6: 47680275-47680275 on Build GRCh38
rs768641325 Chr.6: 47680317-47680317 on Build GRCh38
rs777297561 Chr.6: 47680233-47680233 on Build GRCh38
rs964171043 Chr.6: 47680609-47680609 on Build GRCh38
rs771425814 Chr.6: 47680338-47680338 on Build GRCh38
rs958763562 Chr.6: 47680408-47680408 on Build GRCh38
rs570431855 Chr.6: 47680603-47680603 on Build GRCh38
rs978724432 Chr.6: 47680365-47680365 on Build GRCh38
rs552283127 Chr.6: 47680657-47680657 on Build GRCh38
rs368033292 Chr.6: 47680313-47680313 on Build GRCh38
rs755736619 Chr.6: 47680222-47680222 on Build GRCh38
rs550085988 Chr.6: 47680235-47680235 on Build GRCh38
rs576803769 Chr.6: 47680333-47680333 on Build GRCh38
rs992879110 Chr.6: 47680658-47680658 on Build GRCh38
rs575036811 Chr.6: 47680475-47680475 on Build GRCh38
rs375026503 Chr.6: 47680345-47680345 on Build GRCh38
rs1043588094 Chr.6: 47680558-47680558 on Build GRCh38
rs771225684 Chr.6: 47680578-47680578 on Build GRCh38
rs747072908 Chr.6: 47680315-47680315 on Build GRCh38
rs780235264 Chr.6: 47680247-47680247 on Build GRCh38
rs757019872 Chr.6: 47680308-47680308 on Build GRCh38
rs763629880 Chr.6: 47680356-47680356 on Build GRCh38
rs753541668 Chr.6: 47680305-47680305 on Build GRCh38
rs531325204 Chr.6: 47680686-47680686 on Build GRCh38
rs1012122437 Chr.6: 47680533-47680533 on Build GRCh38
rs778093772 Chr.6: 47680498-47680498 on Build GRCh38
rs752187354 Chr.6: 47680285-47680285 on Build GRCh38
rs773851787 Chr.6: 47680271-47680271 on Build GRCh38
rs745591083 Chr.6: 47680629-47680629 on Build GRCh38

View Full Product Details