Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 43396335-43396921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECW1
Entrez Gene ID
23072
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases
NEDL1
UniGene
Hs.164453
Gene Chromosome Location
Chr.7: 43112580-43566001 on Build GRCh38
Transcripts
NM_001287059.1 NM_015052.4 XM_005249665.3 XM_006715670.3 XM_006715671.3 XM_006715673.3 XM_011515220.2 XM_011515222.2 XM_011515223.2 XM_011515224.2 XM_011515225.2 XM_011515226.2 XM_017011882.1 XM_017011883.1 XM_017011884.1 XM_017011885.1 XM_017011886.1 XM_017011887.1 XM_017011888.1 XM_017011889.1 XM_017011890.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3230199 c.584_587delCCGT p.V196fs*8 Chr.7: 43396842-43396845 on Build GRCh38
746877 c.600A>G p.G200G Chr.7: 43396858-43396858 on Build GRCh38
4689916 c.598G>T p.G200* Chr.7: 43396856-43396856 on Build GRCh38
1313088 c.631+1G>C p.? Chr.7: 43396890-43396890 on Build GRCh38
1755274 c.630A>T p.S210S Chr.7: 43396888-43396888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs547939909 Chr.7: 43396791-43396791 on Build GRCh38
rs547206222 Chr.7: 43396701-43396701 on Build GRCh38
rs934714501 Chr.7: 43396548-43396548 on Build GRCh38
rs997936128 Chr.7: 43396642-43396642 on Build GRCh38
rs974671074 Chr.7: 43396457-43396457 on Build GRCh38
rs1054881260 Chr.7: 43396581-43396581 on Build GRCh38
rs1026066455 Chr.7: 43396382-43396382 on Build GRCh38
rs1003044361 Chr.7: 43396679-43396679 on Build GRCh38
rs148671210 Chr.7: 43396390-43396390 on Build GRCh38
rs889514118 Chr.7: 43396668-43396668 on Build GRCh38
rs1042315565 Chr.7: 43396530-43396530 on Build GRCh38
rs762506249 Chr.7: 43396827-43396827 on Build GRCh38
rs950408124 Chr.7: 43396393-43396393 on Build GRCh38
rs908849357 Chr.7: 43396413-43396413 on Build GRCh38
rs750980231 Chr.7: 43396842-43396842 on Build GRCh38
rs893116602 Chr.7: 43396590-43396591 on Build GRCh38
rs1005730884 Chr.7: 43396706-43396706 on Build GRCh38
rs1028581165 Chr.7: 43396665-43396665 on Build GRCh38
rs71540531 Chr.7: 43396440-43396440 on Build GRCh38
rs73101371 Chr.7: 43396716-43396716 on Build GRCh38
rs1013613916 Chr.7: 43396623-43396623 on Build GRCh38
rs544553242 Chr.7: 43396614-43396614 on Build GRCh38
rs926690395 Chr.7: 43396461-43396461 on Build GRCh38
rs540179273 Chr.7: 43396389-43396389 on Build GRCh38
rs1051164348 Chr.7: 43396502-43396502 on Build GRCh38
rs893828509 Chr.7: 43396503-43396503 on Build GRCh38
rs73101370 Chr.7: 43396708-43396708 on Build GRCh38
rs759315657 Chr.7: 43396784-43396784 on Build GRCh38
rs375753925 Chr.7: 43396870-43396870 on Build GRCh38
rs1026608534 Chr.7: 43396789-43396789 on Build GRCh38
rs903281174 Chr.7: 43396537-43396537 on Build GRCh38
rs752341096 Chr.7: 43396825-43396825 on Build GRCh38
rs763572809 Chr.7: 43396830-43396830 on Build GRCh38
rs374450219 Chr.7: 43396705-43396705 on Build GRCh38
rs374959981 Chr.7: 43396843-43396843 on Build GRCh38
rs760493961 Chr.7: 43396772-43396772 on Build GRCh38
rs945277853 Chr.7: 43396525-43396525 on Build GRCh38
rs562723261 Chr.7: 43396653-43396653 on Build GRCh38
rs776309267 Chr.7: 43396777-43396777 on Build GRCh38
rs764872831 Chr.7: 43396824-43396824 on Build GRCh38
rs773344957 Chr.7: 43396766-43396766 on Build GRCh38
rs938103835 Chr.7: 43396498-43396498 on Build GRCh38
rs901010621 Chr.7: 43396558-43396558 on Build GRCh38
rs770859134 Chr.7: 43396776-43396776 on Build GRCh38
rs772331590 Chr.7: 43396867-43396867 on Build GRCh38
rs907403385 Chr.7: 43396678-43396678 on Build GRCh38
rs923128796 Chr.7: 43396480-43396480 on Build GRCh38
rs763762233 Chr.7: 43396710-43396710 on Build GRCh38
rs933149057 Chr.7: 43396487-43396487 on Build GRCh38
rs2302446 Chr.7: 43396452-43396452 on Build GRCh38
rs755329283 Chr.7: 43396847-43396847 on Build GRCh38
rs984464190 Chr.7: 43396408-43396408 on Build GRCh38
rs761332667 Chr.7: 43396745-43396745 on Build GRCh38
rs1053765147 Chr.7: 43396501-43396501 on Build GRCh38
rs201771274 Chr.7: 43396844-43396844 on Build GRCh38
rs989995451 Chr.7: 43396875-43396875 on Build GRCh38
rs779347290 Chr.7: 43396863-43396863 on Build GRCh38
rs748371744 Chr.7: 43396866-43396866 on Build GRCh38

View Full Product Details