Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 67745849-67746435 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPA6
Entrez Gene ID
57094
Gene Name
carboxypeptidase A6
Gene Aliases
CPAH, ETL5, FEB11
UniGene
Hs.658850
Gene Chromosome Location
Chr.8: 67422170-67747114 on Build GRCh38
Transcripts
NM_020361.4 XM_011517570.2 XM_017013646.1 XM_017013647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3669916 c.99T>C p.Y33Y Chr.8: 67746031-67746031 on Build GRCh38
3745651 c.116+4A>C p.? Chr.8: 67746010-67746010 on Build GRCh38
5503382 c.24G>T p.R8S Chr.8: 67746106-67746106 on Build GRCh38
6113543 c.53G>T p.C18F Chr.8: 67746077-67746077 on Build GRCh38
4756910 c.94C>A p.L32I Chr.8: 67746036-67746036 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs188334231 Chr.8: 67746237-67746237 on Build GRCh38
rs767794744 Chr.8: 67746174-67746174 on Build GRCh38
rs1018666393 Chr.8: 67746036-67746036 on Build GRCh38
rs377180135 Chr.8: 67746081-67746081 on Build GRCh38
rs748998712 Chr.8: 67746142-67746142 on Build GRCh38
rs568876691 Chr.8: 67746145-67746145 on Build GRCh38
rs746533204 Chr.8: 67745989-67745989 on Build GRCh38
rs910910735 Chr.8: 67745889-67745889 on Build GRCh38
rs983914438 Chr.8: 67746302-67746302 on Build GRCh38
rs966289015 Chr.8: 67746245-67746245 on Build GRCh38
rs751585743 Chr.8: 67745887-67745887 on Build GRCh38
rs776577090 Chr.8: 67745992-67745992 on Build GRCh38
rs1007724929 Chr.8: 67746234-67746234 on Build GRCh38
rs946670441 Chr.8: 67746399-67746399 on Build GRCh38
rs909741872 Chr.8: 67746147-67746147 on Build GRCh38
rs556771570 Chr.8: 67746031-67746031 on Build GRCh38
rs530829401 Chr.8: 67746025-67746025 on Build GRCh38
rs144013159 Chr.8: 67746349-67746351 on Build GRCh38
rs1044018641 Chr.8: 67746400-67746400 on Build GRCh38
rs1041726289 Chr.8: 67746205-67746205 on Build GRCh38
rs886063082 Chr.8: 67746021-67746021 on Build GRCh38
rs1048228849 Chr.8: 67745986-67745986 on Build GRCh38
rs183899632 Chr.8: 67746023-67746023 on Build GRCh38
rs753639641 Chr.8: 67746215-67746215 on Build GRCh38
rs886063083 Chr.8: 67746205-67746205 on Build GRCh38
rs768509343 Chr.8: 67745991-67745991 on Build GRCh38
rs751208073 Chr.8: 67746178-67746178 on Build GRCh38
rs758819365 Chr.8: 67746113-67746113 on Build GRCh38
rs796925267 Chr.8: 67746289-67746289 on Build GRCh38
rs768856269 Chr.8: 67746161-67746161 on Build GRCh38
rs778807405 Chr.8: 67746076-67746076 on Build GRCh38
rs895364237 Chr.8: 67745981-67745981 on Build GRCh38
rs564422781 Chr.8: 67746411-67746411 on Build GRCh38
rs376304846 Chr.8: 67746229-67746229 on Build GRCh38
rs996321063 Chr.8: 67746249-67746249 on Build GRCh38
rs780746823 Chr.8: 67746127-67746127 on Build GRCh38
rs537166140 Chr.8: 67746348-67746349 on Build GRCh38
rs765895994 Chr.8: 67746117-67746117 on Build GRCh38
rs62511445 Chr.8: 67746286-67746286 on Build GRCh38
rs757678635 Chr.8: 67746040-67746040 on Build GRCh38
rs779485838 Chr.8: 67745985-67745985 on Build GRCh38
rs536119354 Chr.8: 67745962-67745962 on Build GRCh38
rs539443552 Chr.8: 67745877-67745877 on Build GRCh38
rs772119815 Chr.8: 67746087-67746087 on Build GRCh38
rs754502832 Chr.8: 67746063-67746063 on Build GRCh38
rs772940006 Chr.8: 67746082-67746082 on Build GRCh38
rs771582944 Chr.8: 67745978-67745978 on Build GRCh38
rs752233230 Chr.8: 67746024-67746024 on Build GRCh38
rs180803318 Chr.8: 67746274-67746274 on Build GRCh38
rs371447072 Chr.8: 67745964-67745964 on Build GRCh38
rs779189009 Chr.8: 67746041-67746041 on Build GRCh38
rs755948332 Chr.8: 67746130-67746130 on Build GRCh38
rs368736137 Chr.8: 67746032-67746032 on Build GRCh38
rs999670106 Chr.8: 67746222-67746222 on Build GRCh38
rs1019111380 Chr.8: 67746231-67746231 on Build GRCh38
rs376559739 Chr.8: 67746137-67746137 on Build GRCh38
rs973242531 Chr.8: 67746141-67746141 on Build GRCh38
rs16933590 Chr.8: 67745892-67745892 on Build GRCh38
rs141229869 Chr.8: 67746003-67746003 on Build GRCh38
rs545461667 Chr.8: 67746190-67746190 on Build GRCh38
rs889316279 Chr.8: 67746232-67746232 on Build GRCh38
rs373565150 Chr.8: 67746110-67746110 on Build GRCh38
rs761546850 Chr.8: 67745999-67745999 on Build GRCh38
rs775886277 Chr.8: 67746095-67746095 on Build GRCh38
rs928048959 Chr.8: 67746363-67746363 on Build GRCh38
rs765133780 Chr.8: 67746145-67746145 on Build GRCh38
rs777248963 Chr.8: 67746106-67746106 on Build GRCh38
rs754451669 Chr.8: 67746118-67746118 on Build GRCh38
rs951098557 Chr.8: 67746289-67746289 on Build GRCh38
rs773617395 Chr.8: 67746002-67746002 on Build GRCh38
rs761953994 Chr.8: 67746168-67746168 on Build GRCh38
rs2553663 Chr.8: 67746398-67746398 on Build GRCh38
rs748121520 Chr.8: 67746073-67746073 on Build GRCh38
rs559634854 Chr.8: 67746216-67746216 on Build GRCh38
rs879136810 Chr.8: 67745895-67745896 on Build GRCh38
rs746478158 Chr.8: 67746059-67746059 on Build GRCh38
rs766768786 Chr.8: 67746176-67746176 on Build GRCh38
rs991291083 Chr.8: 67746377-67746377 on Build GRCh38
rs759742958 Chr.8: 67746008-67746008 on Build GRCh38
rs1019195069 Chr.8: 67746104-67746104 on Build GRCh38
rs745559095 Chr.8: 67745972-67745972 on Build GRCh38
rs200644843 Chr.8: 67746084-67746084 on Build GRCh38
rs764323458 Chr.8: 67746105-67746105 on Build GRCh38
rs201169482 Chr.8: 67746101-67746101 on Build GRCh38
rs939353626 Chr.8: 67746158-67746158 on Build GRCh38
rs774502318 Chr.8: 67746129-67746129 on Build GRCh38
rs770783757 Chr.8: 67746143-67746143 on Build GRCh38
rs530580560 Chr.8: 67746376-67746376 on Build GRCh38
rs765617953 Chr.8: 67746171-67746171 on Build GRCh38
rs961872522 Chr.8: 67746331-67746331 on Build GRCh38
rs759742005 Chr.8: 67746165-67746165 on Build GRCh38
rs567035404 Chr.8: 67745905-67745905 on Build GRCh38
rs773789263 Chr.8: 67746173-67746173 on Build GRCh38
rs958569542 Chr.8: 67746336-67746336 on Build GRCh38
rs747276469 Chr.8: 67746149-67746149 on Build GRCh38
rs981260294 Chr.8: 67746348-67746348 on Build GRCh38
rs147067921 Chr.8: 67746069-67746069 on Build GRCh38
rs770301336 Chr.8: 67745945-67745945 on Build GRCh38
rs917047013 Chr.8: 67746415-67746415 on Build GRCh38
rs1014766923 Chr.8: 67746320-67746320 on Build GRCh38
rs757926740 Chr.8: 67746050-67746050 on Build GRCh38
rs752718665 Chr.8: 67746176-67746176 on Build GRCh38
rs531326049 Chr.8: 67746139-67746140 on Build GRCh38
rs138360068 Chr.8: 67746111-67746111 on Build GRCh38
rs116871526 Chr.8: 67746308-67746308 on Build GRCh38
rs758163267 Chr.8: 67746180-67746180 on Build GRCh38
rs779759787 Chr.8: 67746051-67746051 on Build GRCh38
rs976937892 Chr.8: 67745950-67745950 on Build GRCh38
rs956165753 Chr.8: 67745943-67745943 on Build GRCh38
rs369835154 Chr.8: 67746049-67746049 on Build GRCh38
rs1028698258 Chr.8: 67746262-67746262 on Build GRCh38

View Full Product Details