Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 40015184-40015769 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDO2
Entrez Gene ID
169355
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
INDOL1
UniGene
Hs.676257
Gene Chromosome Location
Chr.8: 39934955-40016391 on Build GRCh38
Transcripts
NM_194294.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
454555 c.905C>A p.P302H Chr.8: 40015283-40015283 on Build GRCh38
1099807 c.1129T>G p.L377V Chr.8: 40015507-40015507 on Build GRCh38
5874688 c.974G>A p.G325E Chr.8: 40015352-40015352 on Build GRCh38
3649125 c.963C>T p.I321I Chr.8: 40015341-40015341 on Build GRCh38
1099803 c.881A>G p.Y294C Chr.8: 40015259-40015259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199709416 Chr.8: 40015598-40015598 on Build GRCh38
rs767275058 Chr.8: 40015343-40015343 on Build GRCh38
rs781690088 Chr.8: 40015454-40015454 on Build GRCh38
rs1048647583 Chr.8: 40015384-40015384 on Build GRCh38
rs750983808 Chr.8: 40015379-40015379 on Build GRCh38
rs769451642 Chr.8: 40015603-40015603 on Build GRCh38
rs779333801 Chr.8: 40015260-40015260 on Build GRCh38
rs770066631 Chr.8: 40015399-40015399 on Build GRCh38
rs956227202 Chr.8: 40015622-40015622 on Build GRCh38
rs536506056 Chr.8: 40015288-40015288 on Build GRCh38
rs753081734 Chr.8: 40015563-40015563 on Build GRCh38
rs1052663089 Chr.8: 40015691-40015691 on Build GRCh38
rs748888650 Chr.8: 40015609-40015609 on Build GRCh38
rs373052122 Chr.8: 40015328-40015328 on Build GRCh38
rs776608062 Chr.8: 40015301-40015301 on Build GRCh38
rs761051665 Chr.8: 40015632-40015632 on Build GRCh38
rs552327315 Chr.8: 40015618-40015618 on Build GRCh38
rs907094758 Chr.8: 40015535-40015535 on Build GRCh38
rs151299328 Chr.8: 40015637-40015637 on Build GRCh38
rs759542246 Chr.8: 40015338-40015338 on Build GRCh38
rs778081807 Chr.8: 40015573-40015573 on Build GRCh38
rs558816457 Chr.8: 40015380-40015380 on Build GRCh38
rs372601729 Chr.8: 40015626-40015626 on Build GRCh38
rs760274958 Chr.8: 40015349-40015349 on Build GRCh38
rs368029995 Chr.8: 40015648-40015648 on Build GRCh38
rs753303778 Chr.8: 40015247-40015247 on Build GRCh38
rs760459934 Chr.8: 40015413-40015413 on Build GRCh38
rs370047903 Chr.8: 40015290-40015290 on Build GRCh38
rs373073789 Chr.8: 40015216-40015216 on Build GRCh38
rs778845272 Chr.8: 40015589-40015589 on Build GRCh38
rs530047396 Chr.8: 40015446-40015446 on Build GRCh38
rs753866882 Chr.8: 40015580-40015580 on Build GRCh38
rs746915988 Chr.8: 40015519-40015519 on Build GRCh38
rs1009281565 Chr.8: 40015543-40015543 on Build GRCh38
rs773715519 Chr.8: 40015617-40015617 on Build GRCh38
rs370060883 Chr.8: 40015450-40015450 on Build GRCh38
rs964795429 Chr.8: 40015672-40015672 on Build GRCh38
rs1046060364 Chr.8: 40015412-40015412 on Build GRCh38
rs756438349 Chr.8: 40015564-40015564 on Build GRCh38
rs747011736 Chr.8: 40015590-40015590 on Build GRCh38
rs781509860 Chr.8: 40015396-40015396 on Build GRCh38
rs772016439 Chr.8: 40015409-40015409 on Build GRCh38
rs748192172 Chr.8: 40015312-40015312 on Build GRCh38
rs4503083 Chr.8: 40015416-40015416 on Build GRCh38
rs368981034 Chr.8: 40015212-40015212 on Build GRCh38
rs764772490 Chr.8: 40015433-40015433 on Build GRCh38
rs768492657 Chr.8: 40015520-40015520 on Build GRCh38
rs768546107 Chr.8: 40015594-40015594 on Build GRCh38
rs774313775 Chr.8: 40015223-40015223 on Build GRCh38
rs200984760 Chr.8: 40015631-40015631 on Build GRCh38
rs374890915 Chr.8: 40015494-40015494 on Build GRCh38
rs72632016 Chr.8: 40015534-40015534 on Build GRCh38
rs532698228 Chr.8: 40015610-40015610 on Build GRCh38
rs981410825 Chr.8: 40015306-40015306 on Build GRCh38
rs112415348 Chr.8: 40015265-40015265 on Build GRCh38
rs143841632 Chr.8: 40015424-40015424 on Build GRCh38
rs183730312 Chr.8: 40015733-40015733 on Build GRCh38
rs368255551 Chr.8: 40015555-40015555 on Build GRCh38
rs758425715 Chr.8: 40015700-40015700 on Build GRCh38
rs768043479 Chr.8: 40015559-40015559 on Build GRCh38
rs377700687 Chr.8: 40015533-40015533 on Build GRCh38
rs763027626 Chr.8: 40015221-40015221 on Build GRCh38
rs545208551 Chr.8: 40015352-40015352 on Build GRCh38
rs752360511 Chr.8: 40015235-40015235 on Build GRCh38
rs748247079 Chr.8: 40015398-40015398 on Build GRCh38
rs757782327 Chr.8: 40015257-40015257 on Build GRCh38
rs1012793230 Chr.8: 40015228-40015228 on Build GRCh38
rs35446289 Chr.8: 40015494-40015494 on Build GRCh38
rs369887911 Chr.8: 40015326-40015326 on Build GRCh38
rs34602909 Chr.8: 40015745-40015746 on Build GRCh38
rs947836797 Chr.8: 40015653-40015653 on Build GRCh38
rs527368826 Chr.8: 40015646-40015646 on Build GRCh38
rs935575836 Chr.8: 40015677-40015677 on Build GRCh38
rs558073584 Chr.8: 40015686-40015686 on Build GRCh38
rs768100520 Chr.8: 40015620-40015620 on Build GRCh38
rs60581047 Chr.8: 40015222-40015222 on Build GRCh38
rs762470687 Chr.8: 40015541-40015541 on Build GRCh38
rs775505208 Chr.8: 40015215-40015215 on Build GRCh38
rs567190911 Chr.8: 40015239-40015239 on Build GRCh38
rs763751985 Chr.8: 40015350-40015350 on Build GRCh38
rs771915562 Chr.8: 40015502-40015502 on Build GRCh38
rs780320507 Chr.8: 40015387-40015387 on Build GRCh38
rs763564115 Chr.8: 40015241-40015241 on Build GRCh38
rs776333086 Chr.8: 40015420-40015420 on Build GRCh38
rs749350659 Chr.8: 40015500-40015500 on Build GRCh38
rs2981162 Chr.8: 40015669-40015669 on Build GRCh38
rs894077073 Chr.8: 40015708-40015708 on Build GRCh38
rs543682768 Chr.8: 40015532-40015532 on Build GRCh38
rs2981161 Chr.8: 40015347-40015347 on Build GRCh38
rs756689363 Chr.8: 40015355-40015355 on Build GRCh38
rs768889839 Chr.8: 40015295-40015295 on Build GRCh38
rs775167877 Chr.8: 40015411-40015411 on Build GRCh38
rs952676631 Chr.8: 40015630-40015630 on Build GRCh38
rs759351934 Chr.8: 40015226-40015226 on Build GRCh38
rs983678276 Chr.8: 40015713-40015713 on Build GRCh38
rs751886484 Chr.8: 40015451-40015451 on Build GRCh38
rs749901630 Chr.8: 40015440-40015440 on Build GRCh38
rs745850654 Chr.8: 40015408-40015408 on Build GRCh38
rs757423775 Chr.8: 40015582-40015582 on Build GRCh38
rs567204916 Chr.8: 40015679-40015679 on Build GRCh38
rs745426946 Chr.8: 40015232-40015232 on Build GRCh38
rs746234998 Chr.8: 40015282-40015282 on Build GRCh38
rs369897046 Chr.8: 40015381-40015381 on Build GRCh38
rs753026543 Chr.8: 40015471-40015471 on Build GRCh38
rs987509951 Chr.8: 40015270-40015270 on Build GRCh38
rs772855871 Chr.8: 40015605-40015605 on Build GRCh38
rs767203401 Chr.8: 40015230-40015230 on Build GRCh38
rs766804092 Chr.8: 40015553-40015553 on Build GRCh38
rs747887808 Chr.8: 40015595-40015595 on Build GRCh38
rs779974479 Chr.8: 40015510-40015510 on Build GRCh38
rs956109249 Chr.8: 40015321-40015321 on Build GRCh38
rs1046004626 Chr.8: 40015665-40015665 on Build GRCh38
rs1032107204 Chr.8: 40015319-40015319 on Build GRCh38
rs755181173 Chr.8: 40015394-40015394 on Build GRCh38
rs376274283 Chr.8: 40015554-40015554 on Build GRCh38
rs199835214 Chr.8: 40015453-40015453 on Build GRCh38
rs776748042 Chr.8: 40015219-40015219 on Build GRCh38
rs773412552 Chr.8: 40015400-40015400 on Build GRCh38
rs762382412 Chr.8: 40015455-40015465 on Build GRCh38
rs764718999 Chr.8: 40015365-40015365 on Build GRCh38
rs747361203 Chr.8: 40015391-40015391 on Build GRCh38
rs756383360 Chr.8: 40015489-40015489 on Build GRCh38
rs768187167 Chr.8: 40015524-40015529 on Build GRCh38

View Full Product Details