Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 193096557-193097130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLRX2
Entrez Gene ID
51022
Gene Name
glutaredoxin 2
Gene Aliases
CGI-133, GRX2
UniGene
Hs.458283
Gene Chromosome Location
Chr.1: 193096465-193106535 on Build GRCh38
Transcripts
NM_001243399.1 NM_001319291.1 NM_016066.4 NM_197962.2 XM_017001397.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4815704 c.494G>C p.*165S Chr.1: 193096626-193096626 on Build GRCh38
6671766 c.475_476insA p.S159fs*2 Chr.1: 193096647-193096648 on Build GRCh38
6562437 c.487G>C p.E163Q Chr.1: 193096636-193096636 on Build GRCh38
6671767 c.472_473insA p.S158fs*2 Chr.1: 193096647-193096648 on Build GRCh38
6562438 c.484G>C p.E162Q Chr.1: 193096636-193096636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780399378 Chr.1: 193096726-193096726 on Build GRCh38
rs184415012 Chr.1: 193096592-193096592 on Build GRCh38
rs556097790 Chr.1: 193097093-193097093 on Build GRCh38
rs774500615 Chr.1: 193096593-193096593 on Build GRCh38
rs761746383 Chr.1: 193096602-193096602 on Build GRCh38
rs950186449 Chr.1: 193096883-193096883 on Build GRCh38
rs548168934 Chr.1: 193096813-193096813 on Build GRCh38
rs938714035 Chr.1: 193096703-193096703 on Build GRCh38
rs775829297 Chr.1: 193096586-193096586 on Build GRCh38
rs141929600 Chr.1: 193096669-193096669 on Build GRCh38
rs1042180259 Chr.1: 193096907-193096907 on Build GRCh38
rs186724362 Chr.1: 193096994-193096994 on Build GRCh38
rs996047897 Chr.1: 193096933-193096933 on Build GRCh38
rs892282759 Chr.1: 193096873-193096873 on Build GRCh38
rs755025298 Chr.1: 193096876-193096876 on Build GRCh38
rs1030660519 Chr.1: 193096959-193096959 on Build GRCh38
rs560139641 Chr.1: 193096706-193096706 on Build GRCh38
rs767808174 Chr.1: 193096604-193096604 on Build GRCh38
rs780731719 Chr.1: 193096696-193096696 on Build GRCh38
rs1017255970 Chr.1: 193097048-193097048 on Build GRCh38
rs546142805 Chr.1: 193096801-193096801 on Build GRCh38
rs540518012 Chr.1: 193096697-193096697 on Build GRCh38
rs752476632 Chr.1: 193096652-193096652 on Build GRCh38
rs751423249 Chr.1: 193096730-193096733 on Build GRCh38
rs112938279 Chr.1: 193096635-193096635 on Build GRCh38
rs893759905 Chr.1: 193096932-193096932 on Build GRCh38
rs61823210 Chr.1: 193096658-193096658 on Build GRCh38
rs937540116 Chr.1: 193096865-193096865 on Build GRCh38
rs772105735 Chr.1: 193096773-193096773 on Build GRCh38
rs773424695 Chr.1: 193096788-193096788 on Build GRCh38
rs372259057 Chr.1: 193096682-193096682 on Build GRCh38
rs150124803 Chr.1: 193096825-193096825 on Build GRCh38
rs963146415 Chr.1: 193097055-193097055 on Build GRCh38
rs370646100 Chr.1: 193096868-193096868 on Build GRCh38
rs982930148 Chr.1: 193097090-193097090 on Build GRCh38
rs771221874 Chr.1: 193096802-193096802 on Build GRCh38
rs918169851 Chr.1: 193097091-193097091 on Build GRCh38
rs962029456 Chr.1: 193096800-193096800 on Build GRCh38
rs148511977 Chr.1: 193096647-193096647 on Build GRCh38
rs903238812 Chr.1: 193096918-193096918 on Build GRCh38
rs766360974 Chr.1: 193096682-193096682 on Build GRCh38
rs917961065 Chr.1: 193096659-193096659 on Build GRCh38
rs35090051 Chr.1: 193096637-193096637 on Build GRCh38
rs376497379 Chr.1: 193096753-193096753 on Build GRCh38
rs1009521881 Chr.1: 193096924-193096924 on Build GRCh38
rs1007242148 Chr.1: 193097033-193097033 on Build GRCh38
rs756757914 Chr.1: 193096679-193096679 on Build GRCh38
rs548092191 Chr.1: 193096830-193096830 on Build GRCh38
rs138465476 Chr.1: 193096856-193096856 on Build GRCh38
rs763728884 Chr.1: 193096630-193096630 on Build GRCh38
rs751214984 Chr.1: 193096919-193096920 on Build GRCh38
rs769225274 Chr.1: 193096750-193096750 on Build GRCh38
rs974394242 Chr.1: 193096926-193096938 on Build GRCh38
rs759444633 Chr.1: 193096773-193096775 on Build GRCh38
rs527631966 Chr.1: 193096969-193096969 on Build GRCh38
rs34721216 Chr.1: 193097082-193097082 on Build GRCh38
rs34014687 Chr.1: 193097076-193097076 on Build GRCh38
rs766474479 Chr.1: 193096829-193096829 on Build GRCh38
rs758425419 Chr.1: 193096655-193096655 on Build GRCh38
rs781548865 Chr.1: 193096980-193096980 on Build GRCh38
rs181312956 Chr.1: 193096803-193096803 on Build GRCh38
rs569656900 Chr.1: 193097100-193097100 on Build GRCh38
rs772713285 Chr.1: 193096623-193096623 on Build GRCh38
rs773341502 Chr.1: 193096609-193096609 on Build GRCh38
rs890339930 Chr.1: 193097021-193097021 on Build GRCh38
rs1027188457 Chr.1: 193097078-193097078 on Build GRCh38
rs748654176 Chr.1: 193096756-193096756 on Build GRCh38
rs201987831 Chr.1: 193096743-193096743 on Build GRCh38
rs375313987 Chr.1: 193096763-193096763 on Build GRCh38

View Full Product Details