Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241857242-241857732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXO1
Entrez Gene ID
9156
Gene Name
exonuclease 1
Gene Aliases
HEX1, hExoI
UniGene
Hs.498248
Gene Chromosome Location
Chr.1: 241847967-241889939 on Build GRCh38
Transcripts
NM_001319224.1 NM_003686.4 NM_006027.4 NM_130398.3 XM_006711840.2 XM_011544321.1 XM_011544322.1 XM_011544323.2 XM_011544324.2 XM_017002793.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
906515 c.413G>A p.R138Q Chr.1: 241857352-241857352 on Build GRCh38
2050262 c.517G>C p.D173H Chr.1: 241857456-241857456 on Build GRCh38
4143581 c.493C>A p.Q165K Chr.1: 241857432-241857432 on Build GRCh38
6460274 c.413G>T p.R138L Chr.1: 241857352-241857352 on Build GRCh38
4030191 c.433C>A p.L145I Chr.1: 241857372-241857372 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777068778 Chr.1: 241857353-241857353 on Build GRCh38
rs149474050 Chr.1: 241857368-241857368 on Build GRCh38
rs750417274 Chr.1: 241857509-241857509 on Build GRCh38
rs78172944 Chr.1: 241857432-241857432 on Build GRCh38
rs374996599 Chr.1: 241857312-241857312 on Build GRCh38
rs777175940 Chr.1: 241857409-241857409 on Build GRCh38
rs771721157 Chr.1: 241857392-241857392 on Build GRCh38
rs369374399 Chr.1: 241857336-241857336 on Build GRCh38
rs764663715 Chr.1: 241857352-241857352 on Build GRCh38
rs553223057 Chr.1: 241857648-241857648 on Build GRCh38
rs751185774 Chr.1: 241857518-241857518 on Build GRCh38
rs762527393 Chr.1: 241857287-241857287 on Build GRCh38
rs1025375857 Chr.1: 241857385-241857385 on Build GRCh38
rs781563058 Chr.1: 241857524-241857524 on Build GRCh38
rs756758949 Chr.1: 241857378-241857378 on Build GRCh38
rs978417135 Chr.1: 241857262-241857262 on Build GRCh38
rs372814475 Chr.1: 241857337-241857337 on Build GRCh38
rs769882119 Chr.1: 241857419-241857419 on Build GRCh38
rs368973370 Chr.1: 241857655-241857655 on Build GRCh38
rs763121883 Chr.1: 241857446-241857446 on Build GRCh38
rs1057146808 Chr.1: 241857602-241857602 on Build GRCh38
rs904462238 Chr.1: 241857636-241857636 on Build GRCh38
rs753767623 Chr.1: 241857424-241857424 on Build GRCh38
rs777763548 Chr.1: 241857372-241857372 on Build GRCh38
rs775151146 Chr.1: 241857517-241857517 on Build GRCh38
rs925462040 Chr.1: 241857580-241857580 on Build GRCh38
rs992659532 Chr.1: 241857269-241857269 on Build GRCh38
rs908140818 Chr.1: 241857576-241857576 on Build GRCh38
rs192018075 Chr.1: 241857618-241857618 on Build GRCh38
rs904005495 Chr.1: 241857361-241857361 on Build GRCh38
rs376077603 Chr.1: 241857347-241857347 on Build GRCh38
rs779580828 Chr.1: 241857514-241857514 on Build GRCh38
rs778424706 Chr.1: 241857426-241857426 on Build GRCh38
rs911176364 Chr.1: 241857462-241857462 on Build GRCh38
rs749936394 Chr.1: 241857428-241857428 on Build GRCh38
rs535914355 Chr.1: 241857643-241857643 on Build GRCh38
rs749231655 Chr.1: 241857374-241857374 on Build GRCh38
rs945834542 Chr.1: 241857571-241857571 on Build GRCh38
rs765490234 Chr.1: 241857309-241857309 on Build GRCh38
rs774932337 Chr.1: 241857391-241857392 on Build GRCh38
rs936874455 Chr.1: 241857586-241857586 on Build GRCh38
rs760308371 Chr.1: 241857428-241857429 on Build GRCh38
rs780736011 Chr.1: 241857384-241857384 on Build GRCh38
rs773359403 Chr.1: 241857445-241857445 on Build GRCh38
rs1030545485 Chr.1: 241857538-241857540 on Build GRCh38
rs868747470 Chr.1: 241857481-241857481 on Build GRCh38
rs373134694 Chr.1: 241857360-241857360 on Build GRCh38
rs200953385 Chr.1: 241857443-241857443 on Build GRCh38
rs369513542 Chr.1: 241857358-241857358 on Build GRCh38
rs4149896 Chr.1: 241857502-241857502 on Build GRCh38
rs1043158379 Chr.1: 241857573-241857573 on Build GRCh38
rs753589406 Chr.1: 241857472-241857472 on Build GRCh38
rs141531261 Chr.1: 241857398-241857398 on Build GRCh38
rs371681142 Chr.1: 241857305-241857305 on Build GRCh38
rs748644107 Chr.1: 241857438-241857438 on Build GRCh38
rs764107885 Chr.1: 241857294-241857294 on Build GRCh38
rs937460831 Chr.1: 241857590-241857590 on Build GRCh38
rs774681741 Chr.1: 241857455-241857455 on Build GRCh38
rs756143900 Chr.1: 241857321-241857321 on Build GRCh38
rs201750121 Chr.1: 241857311-241857311 on Build GRCh38
rs756571866 Chr.1: 241857530-241857530 on Build GRCh38
rs754610089 Chr.1: 241857521-241857521 on Build GRCh38
rs964165014 Chr.1: 241857522-241857522 on Build GRCh38
rs746715238 Chr.1: 241857431-241857431 on Build GRCh38
rs201705780 Chr.1: 241857283-241857283 on Build GRCh38
rs767372227 Chr.1: 241857362-241857362 on Build GRCh38
rs775851634 Chr.1: 241857351-241857351 on Build GRCh38
rs757880338 Chr.1: 241857513-241857513 on Build GRCh38
rs761739538 Chr.1: 241857473-241857473 on Build GRCh38
rs552496936 Chr.1: 241857406-241857406 on Build GRCh38
rs1000287789 Chr.1: 241857440-241857440 on Build GRCh38
rs1046404939 Chr.1: 241857701-241857701 on Build GRCh38
rs751898352 Chr.1: 241857422-241857422 on Build GRCh38
rs1041508407 Chr.1: 241857628-241857632 on Build GRCh38
rs12037063 Chr.1: 241857515-241857515 on Build GRCh38
rs187618110 Chr.1: 241857555-241857555 on Build GRCh38
rs756050889 Chr.1: 241857371-241857371 on Build GRCh38
rs747366705 Chr.1: 241857400-241857400 on Build GRCh38
rs142450308 Chr.1: 241857350-241857350 on Build GRCh38
rs912878555 Chr.1: 241857562-241857562 on Build GRCh38
rs757966518 Chr.1: 241857430-241857430 on Build GRCh38
rs771195080 Chr.1: 241857454-241857454 on Build GRCh38
rs145375932 Chr.1: 241857297-241857297 on Build GRCh38
rs143955774 Chr.1: 241857397-241857397 on Build GRCh38
rs748063100 Chr.1: 241857316-241857316 on Build GRCh38
rs945896332 Chr.1: 241857622-241857622 on Build GRCh38
rs745578850 Chr.1: 241857421-241857421 on Build GRCh38
rs140218725 Chr.1: 241857411-241857411 on Build GRCh38
rs778317733 Chr.1: 241857381-241857381 on Build GRCh38
rs975341736 Chr.1: 241857291-241857291 on Build GRCh38
rs765285348 Chr.1: 241857359-241857359 on Build GRCh38
rs147663824 Chr.1: 241857348-241857348 on Build GRCh38
rs759706932 Chr.1: 241857460-241857460 on Build GRCh38
rs376344276 Chr.1: 241857471-241857471 on Build GRCh38
rs369069198 Chr.1: 241857432-241857433 on Build GRCh38
rs771582822 Chr.1: 241857296-241857297 on Build GRCh38
rs375914126 Chr.1: 241857526-241857526 on Build GRCh38

View Full Product Details