Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 92465372-92465921 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDE
Entrez Gene ID
3416
Gene Name
insulin degrading enzyme
Gene Aliases
INSULYSIN
UniGene
Hs.500546
Gene Chromosome Location
Chr.10: 92451684-92574484 on Build GRCh38
Transcripts
NM_001165946.1 NM_001322793.1 NM_001322794.1 NM_001322795.1 NM_001322796.1 NM_001322797.1 NM_004969.3 NR_136399.1 XM_017016187.1 XM_017016188.1 XM_017016189.1 XM_017016190.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1254581 c.2356C>T p.H786Y Chr.10: 92465808-92465808 on Build GRCh38
6683602 c.2470C>T p.R824C Chr.10: 92465694-92465694 on Build GRCh38
1745970 c.2433C>G p.F811L Chr.10: 92465731-92465731 on Build GRCh38
1210231 c.2471G>A p.R824H Chr.10: 92465693-92465693 on Build GRCh38
7351824 c.2321-2A>C p.? Chr.10: 92465845-92465845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs943391767 Chr.10: 92465603-92465603 on Build GRCh38
rs79133297 Chr.10: 92465436-92465436 on Build GRCh38
rs531759698 Chr.10: 92465804-92465804 on Build GRCh38
rs371158499 Chr.10: 92465798-92465798 on Build GRCh38
rs1011900990 Chr.10: 92465849-92465849 on Build GRCh38
rs966883190 Chr.10: 92465415-92465415 on Build GRCh38
rs143583824 Chr.10: 92465790-92465790 on Build GRCh38
rs374090123 Chr.10: 92465707-92465707 on Build GRCh38
rs377755807 Chr.10: 92465767-92465767 on Build GRCh38
rs919452156 Chr.10: 92465499-92465499 on Build GRCh38
rs756355043 Chr.10: 92465881-92465881 on Build GRCh38
rs1013145938 Chr.10: 92465618-92465618 on Build GRCh38
rs993188264 Chr.10: 92465546-92465546 on Build GRCh38
rs181441187 Chr.10: 92465410-92465410 on Build GRCh38
rs760116716 Chr.10: 92465840-92465840 on Build GRCh38
rs754067205 Chr.10: 92465605-92465605 on Build GRCh38
rs747993929 Chr.10: 92465693-92465693 on Build GRCh38
rs944528173 Chr.10: 92465512-92465512 on Build GRCh38
rs762034616 Chr.10: 92465634-92465634 on Build GRCh38
rs1020775201 Chr.10: 92465409-92465409 on Build GRCh38
rs959112665 Chr.10: 92465848-92465848 on Build GRCh38
rs760345406 Chr.10: 92465794-92465794 on Build GRCh38
rs990406671 Chr.10: 92465466-92465466 on Build GRCh38
rs755229563 Chr.10: 92465668-92465668 on Build GRCh38
rs893745525 Chr.10: 92465746-92465746 on Build GRCh38
rs775155984 Chr.10: 92465653-92465653 on Build GRCh38
rs549786749 Chr.10: 92465590-92465590 on Build GRCh38
rs915170149 Chr.10: 92465441-92465441 on Build GRCh38
rs773863452 Chr.10: 92465724-92465724 on Build GRCh38
rs745330969 Chr.10: 92465764-92465764 on Build GRCh38
rs2275218 Chr.10: 92465540-92465540 on Build GRCh38
rs577720253 Chr.10: 92465483-92465483 on Build GRCh38
rs61736444 Chr.10: 92465739-92465739 on Build GRCh38
rs766627099 Chr.10: 92465650-92465650 on Build GRCh38
rs765768051 Chr.10: 92465797-92465797 on Build GRCh38
rs560790879 Chr.10: 92465513-92465513 on Build GRCh38
rs138149617 Chr.10: 92465716-92465716 on Build GRCh38
rs770839965 Chr.10: 92465717-92465717 on Build GRCh38
rs981842304 Chr.10: 92465419-92465419 on Build GRCh38
rs774235358 Chr.10: 92465401-92465401 on Build GRCh38
rs781228645 Chr.10: 92465677-92465677 on Build GRCh38
rs34934289 Chr.10: 92465475-92465475 on Build GRCh38
rs750302921 Chr.10: 92465888-92465888 on Build GRCh38
rs777512890 Chr.10: 92465701-92465701 on Build GRCh38
rs755907583 Chr.10: 92465696-92465696 on Build GRCh38
rs773303572 Chr.10: 92465460-92465460 on Build GRCh38
rs538469 Chr.10: 92465636-92465636 on Build GRCh38
rs959039121 Chr.10: 92465443-92465443 on Build GRCh38
rs759893273 Chr.10: 92465606-92465606 on Build GRCh38
rs35640611 Chr.10: 92465791-92465791 on Build GRCh38
rs575269757 Chr.10: 92465425-92465425 on Build GRCh38
rs758973226 Chr.10: 92465641-92465641 on Build GRCh38
rs929432026 Chr.10: 92465501-92465501 on Build GRCh38
rs771781086 Chr.10: 92465769-92465769 on Build GRCh38
rs888048312 Chr.10: 92465604-92465604 on Build GRCh38
rs1027697044 Chr.10: 92465557-92465557 on Build GRCh38
rs971581027 Chr.10: 92465659-92465659 on Build GRCh38
rs369455056 Chr.10: 92465649-92465649 on Build GRCh38
rs754960451 Chr.10: 92465823-92465823 on Build GRCh38
rs765613544 Chr.10: 92465635-92465635 on Build GRCh38
rs902356171 Chr.10: 92465815-92465815 on Build GRCh38
rs751716971 Chr.10: 92465816-92465816 on Build GRCh38
rs12242504 Chr.10: 92465498-92465498 on Build GRCh38
rs777600939 Chr.10: 92465886-92465886 on Build GRCh38
rs375805971 Chr.10: 92465504-92465504 on Build GRCh38
rs751635118 Chr.10: 92465665-92465665 on Build GRCh38
rs1040164487 Chr.10: 92465524-92465524 on Build GRCh38
rs752902215 Chr.10: 92465847-92465847 on Build GRCh38
rs766928899 Chr.10: 92465811-92465811 on Build GRCh38

View Full Product Details