Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 112154439-112154810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPAM
Entrez Gene ID
57678
Gene Name
glycerol-3-phosphate acyltransferase, mitochondrial
Gene Aliases
GPAT, GPAT1
UniGene
Hs.42586
Gene Chromosome Location
Chr.10: 112149864-112215395 on Build GRCh38
Transcripts
NM_001244949.1 NM_020918.5 XM_005269998.1

SNP Detail

SNP ID Chromosome Location
rs897911521 Chr.10: 112154575-112154575 on Build GRCh38
rs932294995 Chr.10: 112154497-112154497 on Build GRCh38
rs748854463 Chr.10: 112154606-112154606 on Build GRCh38
rs922236893 Chr.10: 112154486-112154486 on Build GRCh38
rs41313471 Chr.10: 112154776-112154776 on Build GRCh38
rs983393955 Chr.10: 112154634-112154634 on Build GRCh38
rs761061631 Chr.10: 112154662-112154662 on Build GRCh38
rs867986988 Chr.10: 112154537-112154537 on Build GRCh38
rs975617399 Chr.10: 112154540-112154540 on Build GRCh38
rs773749498 Chr.10: 112154656-112154656 on Build GRCh38
rs909795338 Chr.10: 112154531-112154531 on Build GRCh38
rs910180331 Chr.10: 112154499-112154499 on Build GRCh38
rs3740149 Chr.10: 112154608-112154608 on Build GRCh38
rs975820290 Chr.10: 112154547-112154547 on Build GRCh38
rs765318307 Chr.10: 112154715-112154715 on Build GRCh38
rs546061694 Chr.10: 112154627-112154627 on Build GRCh38
rs923136082 Chr.10: 112154587-112154587 on Build GRCh38
rs368636066 Chr.10: 112154600-112154600 on Build GRCh38
rs371344866 Chr.10: 112154725-112154725 on Build GRCh38
rs776530332 Chr.10: 112154708-112154708 on Build GRCh38
rs766927356 Chr.10: 112154699-112154699 on Build GRCh38
rs796400704 Chr.10: 112154469-112154469 on Build GRCh38
rs145094728 Chr.10: 112154561-112154564 on Build GRCh38
rs141651151 Chr.10: 112154644-112154644 on Build GRCh38
rs745802183 Chr.10: 112154589-112154589 on Build GRCh38
rs1044801512 Chr.10: 112154784-112154784 on Build GRCh38
rs752901309 Chr.10: 112154723-112154723 on Build GRCh38
rs201875657 Chr.10: 112154714-112154714 on Build GRCh38
rs371600789 Chr.10: 112154620-112154620 on Build GRCh38
rs762568693 Chr.10: 112154692-112154693 on Build GRCh38
rs374846845 Chr.10: 112154688-112154688 on Build GRCh38
rs375579044 Chr.10: 112154597-112154597 on Build GRCh38
rs367548184 Chr.10: 112154724-112154724 on Build GRCh38
rs985401501 Chr.10: 112154529-112154529 on Build GRCh38
rs906571226 Chr.10: 112154658-112154658 on Build GRCh38
rs976528846 Chr.10: 112154787-112154787 on Build GRCh38
rs4918718 Chr.10: 112154534-112154534 on Build GRCh38
rs763494624 Chr.10: 112154731-112154731 on Build GRCh38

View Full Product Details