Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 114295419-114296006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AFAP1L2
Entrez Gene ID
84632
Gene Name
actin filament associated protein 1 like 2
Gene Aliases
CTB-1144G6.4, KIAA1914, XB130
UniGene
Hs.501106
Gene Chromosome Location
Chr.10: 114280730-114405143 on Build GRCh38
Transcripts
NM_001001936.2 NM_001287824.1 NM_032550.3 XM_005270233.4 XM_005270237.4 XM_005270241.4 XM_005270242.4 XM_011540286.2 XM_011540289.1 XM_011540290.1 XM_017016792.1 XM_017016793.1 XM_017016794.1 XM_017016795.1 XM_017016796.1 XM_017016797.1 XM_017016798.1 XM_017016800.1 XM_017016801.1 XM_017016802.1 XM_017016803.1 XM_017016804.1 XM_017016805.1 XM_017016806.1 XM_017016807.1 XM_017016808.1 XM_017016809.1 XM_017016810.1 XM_017016811.1 XM_017016812.1 XM_017016813.1 XM_017016814.1 XM_017016815.1 XM_017016816.1 XM_017016817.1 XM_017016818.1 XM_017016819.1 XM_017016820.1 XM_017016821.1 XM_017016822.1 XM_017016823.1 XM_017016824.1 XM_017016825.1 XM_017016826.1 XM_017016827.1 XM_017016828.1 XM_017016829.1 XM_017016830.1 XM_017016831.1 XM_017016832.1

SNP Detail

SNP ID Chromosome Location
rs371286212 Chr.10: 114295963-114295963 on Build GRCh38
rs564070494 Chr.10: 114295962-114295962 on Build GRCh38
rs756409439 Chr.10: 114295908-114295908 on Build GRCh38
rs10633998 Chr.10: 114295860-114295861 on Build GRCh38
rs774742812 Chr.10: 114295859-114295859 on Build GRCh38
rs1005291597 Chr.10: 114295594-114295594 on Build GRCh38
rs1017645799 Chr.10: 114295702-114295702 on Build GRCh38
rs993799212 Chr.10: 114295621-114295621 on Build GRCh38
rs976072282 Chr.10: 114295725-114295725 on Build GRCh38
rs1048072552 Chr.10: 114295496-114295496 on Build GRCh38
rs757928986 Chr.10: 114295523-114295523 on Build GRCh38
rs1009743358 Chr.10: 114295678-114295678 on Build GRCh38
rs576433315 Chr.10: 114295566-114295566 on Build GRCh38
rs957785112 Chr.10: 114295662-114295663 on Build GRCh38
rs557135747 Chr.10: 114295565-114295565 on Build GRCh38
rs370672702 Chr.10: 114295671-114295671 on Build GRCh38
rs537579400 Chr.10: 114295572-114295572 on Build GRCh38
rs1035031784 Chr.10: 114295932-114295932 on Build GRCh38
rs559570321 Chr.10: 114295735-114295735 on Build GRCh38
rs922550720 Chr.10: 114295515-114295515 on Build GRCh38
rs750004558 Chr.10: 114295521-114295521 on Build GRCh38
rs746022777 Chr.10: 114295889-114295889 on Build GRCh38
rs1000857953 Chr.10: 114295696-114295696 on Build GRCh38
rs764893565 Chr.10: 114295510-114295510 on Build GRCh38
rs574799243 Chr.10: 114295765-114295765 on Build GRCh38
rs369081437 Chr.10: 114295497-114295511 on Build GRCh38
rs563926876 Chr.10: 114295812-114295812 on Build GRCh38
rs541323088 Chr.10: 114295708-114295708 on Build GRCh38
rs1021584248 Chr.10: 114295685-114295685 on Build GRCh38
rs993913653 Chr.10: 114295828-114295828 on Build GRCh38
rs976548813 Chr.10: 114295732-114295732 on Build GRCh38
rs372525605 Chr.10: 114295543-114295543 on Build GRCh38
rs145394167 Chr.10: 114295647-114295647 on Build GRCh38
rs923302492 Chr.10: 114295732-114295733 on Build GRCh38
rs1044024311 Chr.10: 114295803-114295803 on Build GRCh38
rs896620338 Chr.10: 114295676-114295676 on Build GRCh38
rs112535868 Chr.10: 114295523-114295524 on Build GRCh38
rs950567940 Chr.10: 114295752-114295752 on Build GRCh38
rs926940217 Chr.10: 114295823-114295823 on Build GRCh38
rs909137196 Chr.10: 114295517-114295517 on Build GRCh38
rs550909528 Chr.10: 114295910-114295910 on Build GRCh38
rs3750821 Chr.10: 114295658-114295658 on Build GRCh38
rs1045400025 Chr.10: 114295851-114295851 on Build GRCh38
rs933903238 Chr.10: 114295733-114295733 on Build GRCh38
rs528047160 Chr.10: 114295968-114295968 on Build GRCh38
rs778318895 Chr.10: 114295659-114295659 on Build GRCh38
rs889639349 Chr.10: 114295560-114295561 on Build GRCh38
rs1038100605 Chr.10: 114295595-114295595 on Build GRCh38
rs535604768 Chr.10: 114295440-114295440 on Build GRCh38
rs543949491 Chr.10: 114295856-114295857 on Build GRCh38
rs1044075827 Chr.10: 114295540-114295540 on Build GRCh38
rs1056903294 Chr.10: 114295675-114295675 on Build GRCh38
rs545926736 Chr.10: 114295924-114295924 on Build GRCh38
rs906887723 Chr.10: 114295882-114295882 on Build GRCh38
rs779371163 Chr.10: 114295609-114295609 on Build GRCh38
rs896804799 Chr.10: 114295617-114295617 on Build GRCh38
rs1001764825 Chr.10: 114295919-114295919 on Build GRCh38
rs983345172 Chr.10: 114295763-114295763 on Build GRCh38
rs772255256 Chr.10: 114295933-114295933 on Build GRCh38
rs370490636 Chr.10: 114295826-114295826 on Build GRCh38
rs181346166 Chr.10: 114295899-114295899 on Build GRCh38
rs968326729 Chr.10: 114295689-114295689 on Build GRCh38
rs771370830 Chr.10: 114295838-114295838 on Build GRCh38
rs147901415 Chr.10: 114295478-114295492 on Build GRCh38
rs971666888 Chr.10: 114295649-114295649 on Build GRCh38
rs762554544 Chr.10: 114295558-114295558 on Build GRCh38
rs923392582 Chr.10: 114295743-114295743 on Build GRCh38
rs542022010 Chr.10: 114295794-114295794 on Build GRCh38

View Full Product Details