Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 118694910-118695434 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACUL1
Entrez Gene ID
143384
Gene Name
CDK2 associated cullin domain 1
Gene Aliases
C10orf46, CAC1
UniGene
Hs.420024
Gene Chromosome Location
Chr.10: 118680982-118755246 on Build GRCh38
Transcripts
NM_153810.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7205096 c.801A>G p.L267L Chr.10: 118695226-118695226 on Build GRCh38
5527487 c.829C>T p.Q277* Chr.10: 118695198-118695198 on Build GRCh38
1188064 c.849G>T p.M283I Chr.10: 118695178-118695178 on Build GRCh38
6618789 c.813C>A p.A271A Chr.10: 118695214-118695214 on Build GRCh38
364282 c.844A>T p.T282S Chr.10: 118695183-118695183 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763931296 Chr.10: 118695182-118695182 on Build GRCh38
rs368121522 Chr.10: 118695169-118695169 on Build GRCh38
rs370275185 Chr.10: 118695394-118695394 on Build GRCh38
rs772340882 Chr.10: 118695271-118695271 on Build GRCh38
rs747773011 Chr.10: 118695265-118695265 on Build GRCh38
rs752707623 Chr.10: 118695266-118695267 on Build GRCh38
rs779751492 Chr.10: 118695238-118695238 on Build GRCh38
rs376898192 Chr.10: 118695089-118695089 on Build GRCh38
rs556431653 Chr.10: 118695117-118695117 on Build GRCh38
rs554745243 Chr.10: 118694995-118694995 on Build GRCh38
rs377687662 Chr.10: 118695276-118695276 on Build GRCh38
rs772706490 Chr.10: 118695127-118695127 on Build GRCh38
rs779661577 Chr.10: 118695094-118695094 on Build GRCh38
rs753377716 Chr.10: 118695239-118695239 on Build GRCh38
rs765318952 Chr.10: 118695260-118695261 on Build GRCh38
rs937129144 Chr.10: 118695266-118695266 on Build GRCh38
rs900470632 Chr.10: 118695088-118695088 on Build GRCh38
rs766473161 Chr.10: 118695157-118695157 on Build GRCh38
rs750349565 Chr.10: 118695229-118695229 on Build GRCh38
rs774906652 Chr.10: 118695112-118695112 on Build GRCh38
rs757006988 Chr.10: 118695197-118695197 on Build GRCh38
rs773600997 Chr.10: 118695138-118695138 on Build GRCh38
rs762889068 Chr.10: 118695052-118695052 on Build GRCh38
rs141552171 Chr.10: 118695415-118695415 on Build GRCh38
rs199654505 Chr.10: 118695191-118695191 on Build GRCh38
rs375683137 Chr.10: 118695236-118695236 on Build GRCh38
rs1047580712 Chr.10: 118695020-118695020 on Build GRCh38
rs973092635 Chr.10: 118695287-118695287 on Build GRCh38
rs748980834 Chr.10: 118695108-118695108 on Build GRCh38
rs767284846 Chr.10: 118695203-118695203 on Build GRCh38
rs181227204 Chr.10: 118694972-118694972 on Build GRCh38
rs1055200900 Chr.10: 118695314-118695314 on Build GRCh38
rs536051138 Chr.10: 118695072-118695072 on Build GRCh38
rs757151740 Chr.10: 118695403-118695403 on Build GRCh38
rs756448817 Chr.10: 118694966-118694967 on Build GRCh38
rs1020927446 Chr.10: 118694948-118694948 on Build GRCh38
rs1054624144 Chr.10: 118695143-118695143 on Build GRCh38
rs778155115 Chr.10: 118695275-118695275 on Build GRCh38
rs564831351 Chr.10: 118695357-118695357 on Build GRCh38
rs776621612 Chr.10: 118695110-118695111 on Build GRCh38
rs761191651 Chr.10: 118695144-118695144 on Build GRCh38
rs759396942 Chr.10: 118695237-118695238 on Build GRCh38
rs768393011 Chr.10: 118695111-118695111 on Build GRCh38
rs751577828 Chr.10: 118695206-118695206 on Build GRCh38
rs929117891 Chr.10: 118695004-118695004 on Build GRCh38
rs139043037 Chr.10: 118695063-118695063 on Build GRCh38
rs369575855 Chr.10: 118695259-118695259 on Build GRCh38
rs372058175 Chr.10: 118695170-118695170 on Build GRCh38
rs201261327 Chr.10: 118695244-118695244 on Build GRCh38
rs911731563 Chr.10: 118695033-118695033 on Build GRCh38

View Full Product Details