Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6823336-6823906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR162
Entrez Gene ID
27239
Gene Name
G protein-coupled receptor 162
Gene Aliases
A-2, GRCA
UniGene
Hs.631654
Gene Chromosome Location
Chr.12: 6821797-6827418 on Build GRCh38
Transcripts
NM_014449.1 NM_019858.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695323 c.8G>A p.R3Q Chr.12: 6823906-6823906 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1022263895 Chr.12: 6823581-6823581 on Build GRCh38
rs782775930 Chr.12: 6823758-6823758 on Build GRCh38
rs782425347 Chr.12: 6823859-6823859 on Build GRCh38
rs916748134 Chr.12: 6823640-6823640 on Build GRCh38
rs2040353 Chr.12: 6823801-6823801 on Build GRCh38
rs1036936179 Chr.12: 6823711-6823711 on Build GRCh38
rs781896772 Chr.12: 6823761-6823761 on Build GRCh38
rs782585258 Chr.12: 6823850-6823850 on Build GRCh38
rs1048017253 Chr.12: 6823417-6823417 on Build GRCh38
rs782433054 Chr.12: 6823544-6823544 on Build GRCh38
rs782140614 Chr.12: 6823667-6823667 on Build GRCh38
rs954132531 Chr.12: 6823564-6823564 on Build GRCh38
rs188128709 Chr.12: 6823625-6823625 on Build GRCh38
rs149274182 Chr.12: 6823809-6823809 on Build GRCh38
rs948462075 Chr.12: 6823651-6823651 on Build GRCh38
rs998122082 Chr.12: 6823725-6823725 on Build GRCh38
rs781857750 Chr.12: 6823794-6823794 on Build GRCh38
rs967149808 Chr.12: 6823599-6823599 on Build GRCh38
rs960956533 Chr.12: 6823626-6823626 on Build GRCh38
rs1036989948 Chr.12: 6823461-6823461 on Build GRCh38
rs782801621 Chr.12: 6823659-6823659 on Build GRCh38
rs201631838 Chr.12: 6823874-6823874 on Build GRCh38
rs781858035 Chr.12: 6823882-6823882 on Build GRCh38
rs1022842969 Chr.12: 6823785-6823785 on Build GRCh38
rs559862372 Chr.12: 6823701-6823701 on Build GRCh38
rs909547583 Chr.12: 6823660-6823660 on Build GRCh38
rs978366531 Chr.12: 6823601-6823601 on Build GRCh38
rs377755632 Chr.12: 6823797-6823797 on Build GRCh38
rs782281571 Chr.12: 6823882-6823882 on Build GRCh38
rs782618981 Chr.12: 6823784-6823784 on Build GRCh38
rs782752764 Chr.12: 6823870-6823870 on Build GRCh38
rs781917111 Chr.12: 6823727-6823727 on Build GRCh38
rs915631185 Chr.12: 6823412-6823412 on Build GRCh38
rs998046828 Chr.12: 6823506-6823506 on Build GRCh38
rs888116432 Chr.12: 6823447-6823447 on Build GRCh38
rs990923262 Chr.12: 6823634-6823634 on Build GRCh38
rs782363399 Chr.12: 6823709-6823709 on Build GRCh38
rs1028174110 Chr.12: 6823556-6823556 on Build GRCh38
rs782045695 Chr.12: 6823423-6823423 on Build GRCh38
rs781924116 Chr.12: 6823860-6823860 on Build GRCh38
rs782062580 Chr.12: 6823728-6823728 on Build GRCh38
rs900986736 Chr.12: 6823491-6823491 on Build GRCh38
rs1048652353 Chr.12: 6823655-6823655 on Build GRCh38
rs960904235 Chr.12: 6823840-6823840 on Build GRCh38
rs200468818 Chr.12: 6823745-6823745 on Build GRCh38
rs782167225 Chr.12: 6823767-6823767 on Build GRCh38
rs782638257 Chr.12: 6823710-6823710 on Build GRCh38
rs782174284 Chr.12: 6823798-6823798 on Build GRCh38
rs928244894 Chr.12: 6823621-6823621 on Build GRCh38
rs782119221 Chr.12: 6823875-6823875 on Build GRCh38
rs1011411251 Chr.12: 6823568-6823568 on Build GRCh38
rs948492602 Chr.12: 6823416-6823416 on Build GRCh38
rs782414445 Chr.12: 6823722-6823722 on Build GRCh38
rs782219158 Chr.12: 6823853-6823853 on Build GRCh38
rs782088732 Chr.12: 6823774-6823774 on Build GRCh38
rs782296011 Chr.12: 6823861-6823861 on Build GRCh38
rs1004124925 Chr.12: 6823460-6823460 on Build GRCh38
rs1035000000 Chr.12: 6823825-6823825 on Build GRCh38
rs782044419 Chr.12: 6823703-6823704 on Build GRCh38
rs782698097 Chr.12: 6823770-6823770 on Build GRCh38
rs199644244 Chr.12: 6823771-6823771 on Build GRCh38
rs782501498 Chr.12: 6823733-6823733 on Build GRCh38
rs782112510 Chr.12: 6823756-6823756 on Build GRCh38
rs117711782 Chr.12: 6823389-6823389 on Build GRCh38
rs966718948 Chr.12: 6823787-6823787 on Build GRCh38
rs11539232 Chr.12: 6823714-6823714 on Build GRCh38
rs901062011 Chr.12: 6823715-6823715 on Build GRCh38

View Full Product Details