Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 32031290-32031821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KHDRBS1
Entrez Gene ID
10657
Gene Name
KH RNA binding domain containing, signal transduction associated 1
Gene Aliases
Sam68, p62, p68
UniGene
Hs.445893
Gene Chromosome Location
Chr.1: 32013694-32060859 on Build GRCh38
Transcripts
NM_001271878.1 NM_006559.2 NR_073498.1 NR_073499.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1644702 c.512A>G p.N171S Chr.1: 32031528-32031528 on Build GRCh38
908226 c.529C>A p.L177I Chr.1: 32031545-32031545 on Build GRCh38
908227 c.552C>G p.I184M Chr.1: 32031568-32031568 on Build GRCh38
6126115 c.568G>C p.E190Q Chr.1: 32031584-32031584 on Build GRCh38
2077522 c.520G>T p.G174W Chr.1: 32031536-32031536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs41312631 Chr.1: 32031348-32031348 on Build GRCh38
rs369355024 Chr.1: 32031475-32031475 on Build GRCh38
rs373088925 Chr.1: 32031497-32031497 on Build GRCh38
rs542418391 Chr.1: 32031753-32031753 on Build GRCh38
rs900977113 Chr.1: 32031411-32031411 on Build GRCh38
rs577649541 Chr.1: 32031460-32031460 on Build GRCh38
rs559835305 Chr.1: 32031748-32031748 on Build GRCh38
rs753283735 Chr.1: 32031521-32031521 on Build GRCh38
rs778548188 Chr.1: 32031473-32031473 on Build GRCh38
rs771221823 Chr.1: 32031507-32031507 on Build GRCh38
rs758223817 Chr.1: 32031481-32031481 on Build GRCh38
rs746901910 Chr.1: 32031489-32031489 on Build GRCh38
rs376888111 Chr.1: 32031689-32031689 on Build GRCh38
rs527445328 Chr.1: 32031750-32031750 on Build GRCh38
rs533853816 Chr.1: 32031354-32031354 on Build GRCh38
rs373475108 Chr.1: 32031623-32031623 on Build GRCh38
rs952602016 Chr.1: 32031670-32031670 on Build GRCh38
rs1041585268 Chr.1: 32031453-32031453 on Build GRCh38
rs919335896 Chr.1: 32031334-32031334 on Build GRCh38
rs531551134 Chr.1: 32031797-32031797 on Build GRCh38
rs764832191 Chr.1: 32031583-32031583 on Build GRCh38
rs756931104 Chr.1: 32031726-32031726 on Build GRCh38
rs1028200894 Chr.1: 32031667-32031667 on Build GRCh38
rs1041542704 Chr.1: 32031393-32031393 on Build GRCh38
rs749438000 Chr.1: 32031506-32031506 on Build GRCh38
rs761077941 Chr.1: 32031517-32031517 on Build GRCh38
rs771156139 Chr.1: 32031692-32031692 on Build GRCh38
rs555923154 Chr.1: 32031425-32031425 on Build GRCh38
rs200585423 Chr.1: 32031604-32031604 on Build GRCh38
rs188954247 Chr.1: 32031715-32031715 on Build GRCh38
rs761314139 Chr.1: 32031554-32031554 on Build GRCh38
rs775962905 Chr.1: 32031514-32031514 on Build GRCh38
rs963704757 Chr.1: 32031722-32031722 on Build GRCh38
rs376286662 Chr.1: 32031505-32031505 on Build GRCh38
rs949530950 Chr.1: 32031341-32031341 on Build GRCh38
rs759929273 Chr.1: 32031508-32031508 on Build GRCh38
rs779996884 Chr.1: 32031485-32031485 on Build GRCh38
rs772301856 Chr.1: 32031510-32031510 on Build GRCh38
rs779686927 Chr.1: 32031674-32031674 on Build GRCh38
rs961051840 Chr.1: 32031713-32031713 on Build GRCh38
rs971201789 Chr.1: 32031785-32031785 on Build GRCh38
rs764715808 Chr.1: 32031519-32031519 on Build GRCh38
rs58430927 Chr.1: 32031384-32031384 on Build GRCh38
rs970882423 Chr.1: 32031728-32031728 on Build GRCh38
rs758099293 Chr.1: 32031650-32031650 on Build GRCh38
rs937219402 Chr.1: 32031373-32031373 on Build GRCh38
rs749339024 Chr.1: 32031690-32031690 on Build GRCh38
rs867524504 Chr.1: 32031385-32031385 on Build GRCh38
rs751439232 Chr.1: 32031685-32031685 on Build GRCh38
rs972635262 Chr.1: 32031734-32031734 on Build GRCh38
rs754859442 Chr.1: 32031687-32031687 on Build GRCh38
rs561235322 Chr.1: 32031793-32031793 on Build GRCh38

View Full Product Details