Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 29058816-29059271 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C22orf31
Entrez Gene ID
25770
Gene Name
chromosome 22 open reading frame 31
Gene Aliases
HS747E2A, bK747E2.1
UniGene
Hs.50891
Gene Chromosome Location
Chr.22: 29058672-29073974 on Build GRCh38
Transcripts
NM_015370.1 XM_005261491.3 XM_011530096.2 XM_017028741.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5907777 c.451G>A p.E151K Chr.22: 29059164-29059164 on Build GRCh38
1033098 c.606G>A p.T202T Chr.22: 29059009-29059009 on Build GRCh38
4004884 c.656C>T p.A219V Chr.22: 29058959-29058959 on Build GRCh38
1033099 c.588G>T p.Q196H Chr.22: 29059027-29059027 on Build GRCh38
6615679 c.539C>T p.T180I Chr.22: 29059076-29059076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368571887 Chr.22: 29058920-29058920 on Build GRCh38
rs773173970 Chr.22: 29058890-29058890 on Build GRCh38
rs771618915 Chr.22: 29058971-29058971 on Build GRCh38
rs137960357 Chr.22: 29058960-29058960 on Build GRCh38
rs137894229 Chr.22: 29059010-29059010 on Build GRCh38
rs774767456 Chr.22: 29059003-29059003 on Build GRCh38
rs781782390 Chr.22: 29058848-29058848 on Build GRCh38
rs759439524 Chr.22: 29059049-29059049 on Build GRCh38
rs376741034 Chr.22: 29059102-29059102 on Build GRCh38
rs780413015 Chr.22: 29058853-29058853 on Build GRCh38
rs143362009 Chr.22: 29059015-29059015 on Build GRCh38
rs201956712 Chr.22: 29058921-29058921 on Build GRCh38
rs756600462 Chr.22: 29058930-29058930 on Build GRCh38
rs777243259 Chr.22: 29059097-29059097 on Build GRCh38
rs762082122 Chr.22: 29059005-29059005 on Build GRCh38
rs763099609 Chr.22: 29058911-29058911 on Build GRCh38
rs145497257 Chr.22: 29058895-29058895 on Build GRCh38
rs754960376 Chr.22: 29059149-29059150 on Build GRCh38
rs374421493 Chr.22: 29058876-29058876 on Build GRCh38
rs777278729 Chr.22: 29059052-29059052 on Build GRCh38
rs759987285 Chr.22: 29059098-29059098 on Build GRCh38
rs757402176 Chr.22: 29058838-29058838 on Build GRCh38
rs775381148 Chr.22: 29059076-29059076 on Build GRCh38
rs372284586 Chr.22: 29059185-29059185 on Build GRCh38
rs377069164 Chr.22: 29058976-29058976 on Build GRCh38
rs149467740 Chr.22: 29059060-29059060 on Build GRCh38
rs374309411 Chr.22: 29058944-29058944 on Build GRCh38
rs1034646156 Chr.22: 29058995-29058995 on Build GRCh38
rs747175576 Chr.22: 29059205-29059205 on Build GRCh38
rs755409332 Chr.22: 29059019-29059019 on Build GRCh38
rs367892930 Chr.22: 29058961-29058961 on Build GRCh38
rs751143451 Chr.22: 29059142-29059142 on Build GRCh38
rs764436670 Chr.22: 29058917-29058917 on Build GRCh38
rs767733864 Chr.22: 29059008-29059008 on Build GRCh38
rs777225440 Chr.22: 29058977-29058977 on Build GRCh38
rs764440938 Chr.22: 29058908-29058909 on Build GRCh38
rs776532720 Chr.22: 29059217-29059217 on Build GRCh38
rs527877688 Chr.22: 29059146-29059146 on Build GRCh38
rs368048142 Chr.22: 29059114-29059114 on Build GRCh38
rs760873872 Chr.22: 29059092-29059092 on Build GRCh38
rs267606214 Chr.22: 29059079-29059079 on Build GRCh38
rs746589758 Chr.22: 29059054-29059054 on Build GRCh38
rs773861622 Chr.22: 29058869-29058869 on Build GRCh38
rs150118170 Chr.22: 29058877-29058877 on Build GRCh38
rs766118867 Chr.22: 29058897-29058897 on Build GRCh38
rs757978840 Chr.22: 29059036-29059036 on Build GRCh38
rs987442195 Chr.22: 29058947-29058947 on Build GRCh38
rs773519100 Chr.22: 29059229-29059229 on Build GRCh38
rs768284900 Chr.22: 29058868-29058868 on Build GRCh38
rs143333996 Chr.22: 29059183-29059183 on Build GRCh38
rs770845028 Chr.22: 29058985-29058985 on Build GRCh38
rs754295174 Chr.22: 29059018-29059018 on Build GRCh38
rs146939582 Chr.22: 29058998-29058998 on Build GRCh38
rs772575388 Chr.22: 29058870-29058870 on Build GRCh38
rs753747712 Chr.22: 29059083-29059083 on Build GRCh38
rs994703484 Chr.22: 29058939-29058939 on Build GRCh38
rs1048850105 Chr.22: 29059112-29059112 on Build GRCh38
rs193162306 Chr.22: 29059070-29059070 on Build GRCh38
rs574570058 Chr.22: 29059172-29059172 on Build GRCh38
rs769684370 Chr.22: 29059001-29059001 on Build GRCh38
rs779096368 Chr.22: 29059034-29059035 on Build GRCh38
rs973466110 Chr.22: 29059231-29059231 on Build GRCh38
rs1029517082 Chr.22: 29059134-29059134 on Build GRCh38
rs762126996 Chr.22: 29059077-29059077 on Build GRCh38
rs756488608 Chr.22: 29058852-29058852 on Build GRCh38
rs768322801 Chr.22: 29059186-29059186 on Build GRCh38
rs987326625 Chr.22: 29059040-29059040 on Build GRCh38
rs769738587 Chr.22: 29059062-29059062 on Build GRCh38
rs201380718 Chr.22: 29059029-29059029 on Build GRCh38
rs148807209 Chr.22: 29059151-29059151 on Build GRCh38
rs714136 Chr.22: 29058986-29058986 on Build GRCh38
rs781233120 Chr.22: 29059059-29059059 on Build GRCh38
rs746350169 Chr.22: 29058851-29058851 on Build GRCh38
rs773521720 Chr.22: 29059189-29059189 on Build GRCh38
rs765790267 Chr.22: 29059228-29059228 on Build GRCh38
rs776057378 Chr.22: 29059009-29059009 on Build GRCh38
rs1009764848 Chr.22: 29059006-29059006 on Build GRCh38
rs866133658 Chr.22: 29059014-29059014 on Build GRCh38
rs150647312 Chr.22: 29059145-29059145 on Build GRCh38
rs752604444 Chr.22: 29059179-29059183 on Build GRCh38
rs143862587 Chr.22: 29059096-29059096 on Build GRCh38
rs374345401 Chr.22: 29058910-29058910 on Build GRCh38
rs748921003 Chr.22: 29058858-29058858 on Build GRCh38
rs750985136 Chr.22: 29058926-29058926 on Build GRCh38
rs760048821 Chr.22: 29059219-29059219 on Build GRCh38
rs141928677 Chr.22: 29059123-29059123 on Build GRCh38
rs780283742 Chr.22: 29058942-29058942 on Build GRCh38
rs763805355 Chr.22: 29059138-29059138 on Build GRCh38
rs761907073 Chr.22: 29059010-29059011 on Build GRCh38
rs771063997 Chr.22: 29059209-29059209 on Build GRCh38
rs751732458 Chr.22: 29058965-29058973 on Build GRCh38
rs372405935 Chr.22: 29058940-29058940 on Build GRCh38
rs747767260 Chr.22: 29058968-29058968 on Build GRCh38
rs778666885 Chr.22: 29059151-29059151 on Build GRCh38
rs752368086 Chr.22: 29059031-29059031 on Build GRCh38
rs746926059 Chr.22: 29058984-29058984 on Build GRCh38
rs773290339 Chr.22: 29059083-29059083 on Build GRCh38
rs770345617 Chr.22: 29059058-29059058 on Build GRCh38

View Full Product Details