Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65268462-65268995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACTR2
Entrez Gene ID
10097
Gene Name
ARP2 actin related protein 2 homolog
Gene Aliases
ARP2
UniGene
Hs.643727
Gene Chromosome Location
Chr.2: 65227695-65271253 on Build GRCh38
Transcripts
NM_001005386.2 NM_005722.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3055333 c.1028G>T p.R343L Chr.2: 65268577-65268577 on Build GRCh38
1022285 c.1124G>A p.R375Q Chr.2: 65268673-65268673 on Build GRCh38
6291420 c.1066C>T p.L356L Chr.2: 65268615-65268615 on Build GRCh38
3055334 c.1060C>T p.R354C Chr.2: 65268594-65268594 on Build GRCh38
3426606 c.1061G>A p.R354H Chr.2: 65268595-65268595 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1009714391 Chr.2: 65268929-65268929 on Build GRCh38
rs750130999 Chr.2: 65268496-65268496 on Build GRCh38
rs11424 Chr.2: 65268884-65268884 on Build GRCh38
rs759145997 Chr.2: 65268611-65268611 on Build GRCh38
rs574904518 Chr.2: 65268635-65268635 on Build GRCh38
rs776047161 Chr.2: 65268744-65268744 on Build GRCh38
rs895709974 Chr.2: 65268858-65268858 on Build GRCh38
rs17038564 Chr.2: 65268924-65268924 on Build GRCh38
rs17851187 Chr.2: 65268692-65268692 on Build GRCh38
rs268875 Chr.2: 65268753-65268753 on Build GRCh38
rs772692650 Chr.2: 65268547-65268547 on Build GRCh38
rs369526875 Chr.2: 65268530-65268530 on Build GRCh38
rs766862717 Chr.2: 65268548-65268548 on Build GRCh38
rs751755650 Chr.2: 65268514-65268514 on Build GRCh38
rs987854705 Chr.2: 65268768-65268768 on Build GRCh38
rs199819019 Chr.2: 65268747-65268747 on Build GRCh38
rs760116016 Chr.2: 65268556-65268556 on Build GRCh38
rs561798444 Chr.2: 65268840-65268840 on Build GRCh38
rs113084011 Chr.2: 65268837-65268837 on Build GRCh38
rs560183107 Chr.2: 65268689-65268689 on Build GRCh38
rs762512287 Chr.2: 65268522-65268523 on Build GRCh38
rs768458921 Chr.2: 65268529-65268529 on Build GRCh38
rs527787336 Chr.2: 65268849-65268849 on Build GRCh38
rs372771813 Chr.2: 65268534-65268534 on Build GRCh38
rs764765098 Chr.2: 65268666-65268666 on Build GRCh38
rs772043747 Chr.2: 65268540-65268540 on Build GRCh38
rs757353258 Chr.2: 65268673-65268673 on Build GRCh38
rs777633748 Chr.2: 65268700-65268700 on Build GRCh38
rs570506885 Chr.2: 65268890-65268890 on Build GRCh38
rs139637582 Chr.2: 65268803-65268803 on Build GRCh38
rs770390466 Chr.2: 65268741-65268741 on Build GRCh38
rs200356120 Chr.2: 65268561-65268561 on Build GRCh38
rs964782156 Chr.2: 65268960-65268960 on Build GRCh38
rs905529829 Chr.2: 65268893-65268893 on Build GRCh38
rs955102221 Chr.2: 65268937-65268937 on Build GRCh38
rs8771 Chr.2: 65268941-65268941 on Build GRCh38
rs1000349797 Chr.2: 65268902-65268902 on Build GRCh38
rs368220890 Chr.2: 65268699-65268699 on Build GRCh38
rs775919693 Chr.2: 65268761-65268761 on Build GRCh38
rs746974315 Chr.2: 65268730-65268730 on Build GRCh38
rs746083946 Chr.2: 65268520-65268520 on Build GRCh38
rs1032758839 Chr.2: 65268910-65268910 on Build GRCh38
rs548385656 Chr.2: 65268830-65268830 on Build GRCh38
rs770445383 Chr.2: 65268557-65268557 on Build GRCh38
rs910506290 Chr.2: 65268955-65268955 on Build GRCh38
rs753053609 Chr.2: 65268630-65268630 on Build GRCh38
rs531370847 Chr.2: 65268828-65268828 on Build GRCh38
rs781507626 Chr.2: 65268518-65268518 on Build GRCh38
rs562834420 Chr.2: 65268808-65268808 on Build GRCh38
rs752116809 Chr.2: 65268668-65268668 on Build GRCh38
rs765606484 Chr.2: 65268614-65268614 on Build GRCh38
rs1021097952 Chr.2: 65268933-65268933 on Build GRCh38
rs188171224 Chr.2: 65268770-65268770 on Build GRCh38
rs752112550 Chr.2: 65268758-65268762 on Build GRCh38
rs943417335 Chr.2: 65268806-65268806 on Build GRCh38
rs368001778 Chr.2: 65268762-65268762 on Build GRCh38
rs778743679 Chr.2: 65268696-65268696 on Build GRCh38
rs112224060 Chr.2: 65268563-65268563 on Build GRCh38
rs1014183016 Chr.2: 65268882-65268882 on Build GRCh38
rs929150431 Chr.2: 65268846-65268846 on Build GRCh38
rs757539701 Chr.2: 65268516-65268516 on Build GRCh38

View Full Product Details