Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 47379509-47380042 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPCAM
Entrez Gene ID
4072
Gene Name
epithelial cell adhesion molecule
Gene Aliases
DIAR5, EGP-2, EGP314, EGP40, ESA, HNPCC8, KS1/4, KSA, M4S1, MIC18, MK-1, TACSTD1, TROP1
UniGene
Hs.542050
Gene Chromosome Location
Chr.2: 47369148-47387028 on Build GRCh38
Transcripts
NM_002354.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6952447 c.658G>A p.V220I Chr.2: 47379769-47379769 on Build GRCh38
7177053 c.687G>C p.K229N Chr.2: 47379798-47379798 on Build GRCh38
7190291 c.858+6T>C p.? Chr.2: 47379975-47379975 on Build GRCh38
3910372 c.694G>A p.D232N Chr.2: 47379805-47379805 on Build GRCh38
6954704 c.808G>T p.A270S Chr.2: 47379919-47379919 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1005676020 Chr.2: 47379694-47379694 on Build GRCh38
rs202201524 Chr.2: 47379882-47379882 on Build GRCh38
rs534760114 Chr.2: 47379613-47379614 on Build GRCh38
rs749054225 Chr.2: 47379843-47379843 on Build GRCh38
rs760783078 Chr.2: 47379779-47379779 on Build GRCh38
rs778825812 Chr.2: 47379944-47379944 on Build GRCh38
rs753520097 Chr.2: 47379933-47379933 on Build GRCh38
rs775866056 Chr.2: 47379977-47379977 on Build GRCh38
rs748292053 Chr.2: 47379773-47379773 on Build GRCh38
rs565682940 Chr.2: 47379831-47379831 on Build GRCh38
rs954882431 Chr.2: 47379702-47379702 on Build GRCh38
rs763729606 Chr.2: 47379956-47379956 on Build GRCh38
rs759960861 Chr.2: 47379895-47379895 on Build GRCh38
rs758153782 Chr.2: 47380013-47380013 on Build GRCh38
rs864622463 Chr.2: 47379801-47379801 on Build GRCh38
rs776027650 Chr.2: 47379903-47379903 on Build GRCh38
rs377276151 Chr.2: 47379926-47379926 on Build GRCh38
rs989290527 Chr.2: 47379578-47379578 on Build GRCh38
rs557572119 Chr.2: 47379757-47379757 on Build GRCh38
rs200600253 Chr.2: 47379879-47379879 on Build GRCh38
rs746068800 Chr.2: 47379970-47379970 on Build GRCh38
rs532123511 Chr.2: 47379731-47379731 on Build GRCh38
rs182423282 Chr.2: 47379657-47379657 on Build GRCh38
rs766367240 Chr.2: 47380011-47380012 on Build GRCh38
rs769956389 Chr.2: 47379976-47379976 on Build GRCh38
rs779056963 Chr.2: 47379852-47379852 on Build GRCh38
rs864622420 Chr.2: 47379840-47379840 on Build GRCh38
rs138718438 Chr.2: 47379969-47379969 on Build GRCh38
rs587780771 Chr.2: 47379875-47379875 on Build GRCh38
rs776854951 Chr.2: 47379884-47379884 on Build GRCh38
rs758439465 Chr.2: 47379957-47379957 on Build GRCh38
rs542512735 Chr.2: 47379676-47379676 on Build GRCh38
rs863224390 Chr.2: 47379873-47379873 on Build GRCh38
rs756801261 Chr.2: 47379799-47379799 on Build GRCh38
rs749908516 Chr.2: 47379925-47379925 on Build GRCh38
rs878996857 Chr.2: 47379974-47379974 on Build GRCh38
rs864622089 Chr.2: 47379877-47379877 on Build GRCh38
rs759359621 Chr.2: 47379743-47379743 on Build GRCh38
rs147958220 Chr.2: 47379850-47379850 on Build GRCh38
rs568079223 Chr.2: 47379802-47379802 on Build GRCh38
rs202126213 Chr.2: 47379809-47379809 on Build GRCh38
rs3923560 Chr.2: 47379544-47379544 on Build GRCh38
rs915113822 Chr.2: 47379818-47379818 on Build GRCh38
rs886832889 Chr.2: 47379693-47379693 on Build GRCh38
rs772270298 Chr.2: 47379777-47379777 on Build GRCh38
rs753197812 Chr.2: 47379794-47379794 on Build GRCh38
rs989321602 Chr.2: 47379805-47379805 on Build GRCh38
rs541430701 Chr.2: 47379626-47379626 on Build GRCh38
rs768723264 Chr.2: 47379763-47379763 on Build GRCh38
rs933635236 Chr.2: 47379633-47379634 on Build GRCh38
rs765184472 Chr.2: 47379746-47379753 on Build GRCh38
rs900879192 Chr.2: 47379627-47379627 on Build GRCh38
rs780889839 Chr.2: 47379730-47379730 on Build GRCh38
rs1046988477 Chr.2: 47379592-47379592 on Build GRCh38
rs375966033 Chr.2: 47379725-47379725 on Build GRCh38
rs1005250854 Chr.2: 47379615-47379615 on Build GRCh38
rs115283528 Chr.2: 47379942-47379942 on Build GRCh38
rs755988611 Chr.2: 47379738-47379738 on Build GRCh38
rs765113243 Chr.2: 47379817-47379817 on Build GRCh38
rs1017022186 Chr.2: 47379706-47379706 on Build GRCh38
rs769060285 Chr.2: 47379989-47379989 on Build GRCh38
rs115618704 Chr.2: 47379715-47379715 on Build GRCh38
rs752488919 Chr.2: 47379952-47379952 on Build GRCh38
rs776452722 Chr.2: 47379688-47379688 on Build GRCh38
rs777628310 Chr.2: 47380017-47380017 on Build GRCh38
rs771029207 Chr.2: 47379968-47379968 on Build GRCh38
rs369828181 Chr.2: 47380002-47380002 on Build GRCh38
rs752696226 Chr.2: 47379820-47379820 on Build GRCh38
rs774911872 Chr.2: 47379994-47379994 on Build GRCh38
rs879226247 Chr.2: 47379972-47379972 on Build GRCh38
rs766571766 Chr.2: 47379780-47379780 on Build GRCh38
rs191194897 Chr.2: 47380005-47380005 on Build GRCh38
rs755757555 Chr.2: 47379841-47379841 on Build GRCh38
rs13398742 Chr.2: 47379798-47379798 on Build GRCh38
rs776888900 Chr.2: 47379783-47379783 on Build GRCh38
rs879272145 Chr.2: 47379678-47379678 on Build GRCh38
rs752696672 Chr.2: 47379616-47379616 on Build GRCh38
rs369770948 Chr.2: 47379999-47379999 on Build GRCh38
rs770114313 Chr.2: 47379899-47379899 on Build GRCh38
rs199875139 Chr.2: 47379766-47379766 on Build GRCh38
rs148106293 Chr.2: 47379928-47379928 on Build GRCh38
rs762765995 Chr.2: 47379914-47379916 on Build GRCh38
rs564616792 Chr.2: 47379718-47379718 on Build GRCh38
rs779738050 Chr.2: 47379740-47379740 on Build GRCh38
rs779694526 Chr.2: 47379842-47379842 on Build GRCh38
rs1013024828 Chr.2: 47379982-47379982 on Build GRCh38
rs969122017 Chr.2: 47379726-47379726 on Build GRCh38
rs999234141 Chr.2: 47379736-47379736 on Build GRCh38
rs773185578 Chr.2: 47379865-47379865 on Build GRCh38
rs763475158 Chr.2: 47379911-47379911 on Build GRCh38
rs535401857 Chr.2: 47379864-47379864 on Build GRCh38
rs751145659 Chr.2: 47379985-47379985 on Build GRCh38
rs1038082340 Chr.2: 47379964-47379964 on Build GRCh38
rs766993531 Chr.2: 47379813-47379813 on Build GRCh38
rs748065050 Chr.2: 47379863-47379863 on Build GRCh38
rs552390311 Chr.2: 47379717-47379717 on Build GRCh38
rs1803936 Chr.2: 47379854-47379854 on Build GRCh38
rs770690819 Chr.2: 47379661-47379661 on Build GRCh38
rs372674836 Chr.2: 47379778-47379778 on Build GRCh38
rs747071468 Chr.2: 47379966-47379966 on Build GRCh38
rs927296654 Chr.2: 47379753-47379753 on Build GRCh38
rs771063031 Chr.2: 47379872-47379872 on Build GRCh38
rs975353107 Chr.2: 47379880-47379880 on Build GRCh38
rs563702338 Chr.2: 47379711-47379711 on Build GRCh38
rs763240638 Chr.2: 47379988-47379988 on Build GRCh38
rs942560883 Chr.2: 47379594-47379594 on Build GRCh38
rs878854495 Chr.2: 47379946-47379946 on Build GRCh38
rs191046709 Chr.2: 47379604-47379604 on Build GRCh38
rs3923559 Chr.2: 47379635-47379635 on Build GRCh38
rs558832554 Chr.2: 47380003-47380003 on Build GRCh38
rs994019980 Chr.2: 47379585-47379585 on Build GRCh38
rs764624596 Chr.2: 47379917-47379917 on Build GRCh38
rs924675912 Chr.2: 47379537-47379537 on Build GRCh38

View Full Product Details