Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 86454692-86455281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KDM3A
Entrez Gene ID
55818
Gene Name
lysine demethylase 3A
Gene Aliases
JHDM2A, JHMD2A, JMJD1, JMJD1A, TSGA
UniGene
Hs.557425
Gene Chromosome Location
Chr.2: 86440647-86492716 on Build GRCh38
Transcripts
NM_001146688.1 NM_018433.5 XM_006712051.3 XM_017004493.1 XM_017004494.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7164555 c.505G>A p.E169K Chr.2: 86455136-86455136 on Build GRCh38
1752704 c.469C>T p.R157* Chr.2: 86455100-86455100 on Build GRCh38
1023488 c.505G>T p.E169* Chr.2: 86455136-86455136 on Build GRCh38
6383012 c.511C>A p.Q171K Chr.2: 86455142-86455142 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375986346 Chr.2: 86455218-86455218 on Build GRCh38
rs756972130 Chr.2: 86455056-86455056 on Build GRCh38
rs777133848 Chr.2: 86455048-86455048 on Build GRCh38
rs533854706 Chr.2: 86455247-86455247 on Build GRCh38
rs551976167 Chr.2: 86455005-86455005 on Build GRCh38
rs769446587 Chr.2: 86455070-86455070 on Build GRCh38
rs373154030 Chr.2: 86455034-86455034 on Build GRCh38
rs113942794 Chr.2: 86454875-86454875 on Build GRCh38
rs901596374 Chr.2: 86454800-86454800 on Build GRCh38
rs751145946 Chr.2: 86455153-86455153 on Build GRCh38
rs1006795069 Chr.2: 86454840-86454840 on Build GRCh38
rs746636892 Chr.2: 86454746-86454746 on Build GRCh38
rs753108933 Chr.2: 86455041-86455041 on Build GRCh38
rs535766302 Chr.2: 86455213-86455213 on Build GRCh38
rs765513133 Chr.2: 86455127-86455127 on Build GRCh38
rs1040722252 Chr.2: 86454789-86454789 on Build GRCh38
rs1050573964 Chr.2: 86455249-86455249 on Build GRCh38
rs1035631337 Chr.2: 86454882-86454882 on Build GRCh38
rs143285228 Chr.2: 86454929-86454929 on Build GRCh38
rs34746063 Chr.2: 86454974-86454974 on Build GRCh38
rs371044560 Chr.2: 86455101-86455101 on Build GRCh38
rs993726946 Chr.2: 86454809-86454809 on Build GRCh38
rs537293422 Chr.2: 86455066-86455068 on Build GRCh38
rs779867462 Chr.2: 86454722-86454722 on Build GRCh38
rs1024774875 Chr.2: 86455012-86455012 on Build GRCh38
rs746061643 Chr.2: 86455234-86455234 on Build GRCh38
rs572781114 Chr.2: 86455222-86455222 on Build GRCh38
rs146656604 Chr.2: 86454966-86454966 on Build GRCh38
rs368315222 Chr.2: 86455071-86455071 on Build GRCh38
rs537980682 Chr.2: 86455108-86455108 on Build GRCh38
rs764592204 Chr.2: 86455144-86455144 on Build GRCh38
rs567690611 Chr.2: 86455093-86455093 on Build GRCh38
rs967557272 Chr.2: 86455195-86455195 on Build GRCh38
rs376781531 Chr.2: 86455027-86455030 on Build GRCh38
rs776633096 Chr.2: 86454807-86454807 on Build GRCh38
rs993327910 Chr.2: 86454999-86454999 on Build GRCh38
rs753301591 Chr.2: 86455128-86455128 on Build GRCh38
rs562143584 Chr.2: 86454841-86454841 on Build GRCh38
rs1035658997 Chr.2: 86455230-86455230 on Build GRCh38
rs887918277 Chr.2: 86454837-86454837 on Build GRCh38
rs753059744 Chr.2: 86455132-86455132 on Build GRCh38
rs970533635 Chr.2: 86454873-86454873 on Build GRCh38
rs879448032 Chr.2: 86454831-86454831 on Build GRCh38
rs1006417228 Chr.2: 86455154-86455154 on Build GRCh38
rs985680710 Chr.2: 86455015-86455015 on Build GRCh38
rs368560015 Chr.2: 86455229-86455229 on Build GRCh38
rs529925195 Chr.2: 86454866-86454866 on Build GRCh38
rs767319225 Chr.2: 86455159-86455159 on Build GRCh38
rs759981549 Chr.2: 86455125-86455125 on Build GRCh38
rs929553099 Chr.2: 86455103-86455103 on Build GRCh38
rs139842789 Chr.2: 86454744-86454744 on Build GRCh38
rs755996913 Chr.2: 86455165-86455165 on Build GRCh38
rs750098775 Chr.2: 86455162-86455162 on Build GRCh38
rs149847163 Chr.2: 86455141-86455141 on Build GRCh38
rs914464346 Chr.2: 86454987-86454987 on Build GRCh38
rs776807992 Chr.2: 86455116-86455116 on Build GRCh38
rs1048365471 Chr.2: 86454818-86454818 on Build GRCh38
rs1048010526 Chr.2: 86455126-86455126 on Build GRCh38
rs774546682 Chr.2: 86454926-86454926 on Build GRCh38
rs971515007 Chr.2: 86455021-86455021 on Build GRCh38
rs751055972 Chr.2: 86455053-86455053 on Build GRCh38
rs72844691 Chr.2: 86454783-86454783 on Build GRCh38
rs780951414 Chr.2: 86455063-86455063 on Build GRCh38
rs968618167 Chr.2: 86455002-86455002 on Build GRCh38
rs72930366 Chr.2: 86455042-86455042 on Build GRCh38
rs568096784 Chr.2: 86455185-86455185 on Build GRCh38
rs749170478 Chr.2: 86455075-86455075 on Build GRCh38
rs144363423 Chr.2: 86455113-86455113 on Build GRCh38
rs745707605 Chr.2: 86455067-86455067 on Build GRCh38
rs768583500 Chr.2: 86455087-86455087 on Build GRCh38
rs914767436 Chr.2: 86454770-86454770 on Build GRCh38
rs946208409 Chr.2: 86454780-86454780 on Build GRCh38
rs756777757 Chr.2: 86455158-86455158 on Build GRCh38
rs749048694 Chr.2: 86455193-86455193 on Build GRCh38

View Full Product Details