Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96897685-96898283 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM178B
Entrez Gene ID
51252
Gene Name
family with sequence similarity 178 member B
Gene Aliases
-
UniGene
Hs.107922
Gene Chromosome Location
Chr.2: 96875882-96986600 on Build GRCh38
Transcripts
NM_001122646.2 NM_016490.4 XM_011511294.2 XM_017004263.1 XM_017004264.1 XM_017004265.1 XM_017004266.1

SNP Detail

SNP ID Chromosome Location
rs4243786 Chr.2: 96898008-96898008 on Build GRCh38
rs749455093 Chr.2: 96898204-96898204 on Build GRCh38
rs899483560 Chr.2: 96898203-96898203 on Build GRCh38
rs541860582 Chr.2: 96898178-96898178 on Build GRCh38
rs904871451 Chr.2: 96898256-96898256 on Build GRCh38
rs979225122 Chr.2: 96897721-96897721 on Build GRCh38
rs556347919 Chr.2: 96898229-96898229 on Build GRCh38
rs562264212 Chr.2: 96897765-96897765 on Build GRCh38
rs898771286 Chr.2: 96898126-96898126 on Build GRCh38
rs141519147 Chr.2: 96898246-96898246 on Build GRCh38
rs745494656 Chr.2: 96898023-96898023 on Build GRCh38
rs748476183 Chr.2: 96898129-96898129 on Build GRCh38
rs1001747249 Chr.2: 96897797-96897797 on Build GRCh38
rs548142232 Chr.2: 96897930-96897930 on Build GRCh38
rs568549381 Chr.2: 96897875-96897875 on Build GRCh38
rs577109438 Chr.2: 96898066-96898066 on Build GRCh38
rs950045387 Chr.2: 96897792-96897792 on Build GRCh38
rs1047679787 Chr.2: 96898131-96898131 on Build GRCh38
rs541706393 Chr.2: 96898238-96898238 on Build GRCh38
rs770350283 Chr.2: 96898149-96898149 on Build GRCh38
rs752447302 Chr.2: 96897777-96897777 on Build GRCh38
rs532119488 Chr.2: 96897836-96897836 on Build GRCh38
rs1051584679 Chr.2: 96897936-96897936 on Build GRCh38
rs1023285318 Chr.2: 96898255-96898255 on Build GRCh38
rs568269331 Chr.2: 96897964-96897964 on Build GRCh38
rs756065862 Chr.2: 96897839-96897839 on Build GRCh38
rs911140288 Chr.2: 96897714-96897714 on Build GRCh38
rs994766336 Chr.2: 96897837-96897837 on Build GRCh38
rs556709902 Chr.2: 96898125-96898125 on Build GRCh38
rs34638877 Chr.2: 96898134-96898134 on Build GRCh38
rs1038914942 Chr.2: 96897811-96897811 on Build GRCh38
rs890845711 Chr.2: 96898153-96898153 on Build GRCh38
rs925156342 Chr.2: 96897745-96897745 on Build GRCh38
rs148361304 Chr.2: 96897997-96897997 on Build GRCh38
rs963928398 Chr.2: 96897988-96897988 on Build GRCh38
rs1024769504 Chr.2: 96898258-96898258 on Build GRCh38
rs930158150 Chr.2: 96898130-96898130 on Build GRCh38
rs950138237 Chr.2: 96898065-96898065 on Build GRCh38
rs1017932466 Chr.2: 96898201-96898201 on Build GRCh38
rs564328118 Chr.2: 96897796-96897796 on Build GRCh38
rs186863599 Chr.2: 96898230-96898230 on Build GRCh38
rs767265540 Chr.2: 96897753-96897753 on Build GRCh38
rs937570652 Chr.2: 96898074-96898074 on Build GRCh38
rs969651240 Chr.2: 96898009-96898009 on Build GRCh38
rs768618466 Chr.2: 96898129-96898130 on Build GRCh38
rs890259968 Chr.2: 96898208-96898208 on Build GRCh38
rs527938971 Chr.2: 96897781-96897781 on Build GRCh38
rs181656521 Chr.2: 96898089-96898089 on Build GRCh38
rs534053672 Chr.2: 96897996-96897996 on Build GRCh38
rs1052127556 Chr.2: 96898191-96898191 on Build GRCh38
rs982790172 Chr.2: 96898019-96898019 on Build GRCh38
rs551831367 Chr.2: 96897853-96897853 on Build GRCh38
rs773527542 Chr.2: 96898195-96898195 on Build GRCh38
rs913311728 Chr.2: 96898062-96898062 on Build GRCh38
rs755435282 Chr.2: 96898058-96898058 on Build GRCh38
rs898681727 Chr.2: 96897813-96897813 on Build GRCh38
rs927309208 Chr.2: 96898070-96898070 on Build GRCh38
rs898904990 Chr.2: 96898237-96898237 on Build GRCh38
rs963959101 Chr.2: 96897716-96897716 on Build GRCh38
rs996525280 Chr.2: 96897880-96897880 on Build GRCh38
rs781714783 Chr.2: 96898121-96898121 on Build GRCh38
rs1017961383 Chr.2: 96897886-96897886 on Build GRCh38
rs973616074 Chr.2: 96897739-96897739 on Build GRCh38
rs190778042 Chr.2: 96897795-96897795 on Build GRCh38
rs576577990 Chr.2: 96898147-96898147 on Build GRCh38
rs370774476 Chr.2: 96897775-96897775 on Build GRCh38
rs763878535 Chr.2: 96897857-96897857 on Build GRCh38
rs975371750 Chr.2: 96898096-96898096 on Build GRCh38
rs900419888 Chr.2: 96897878-96897878 on Build GRCh38
rs537462859 Chr.2: 96897884-96897884 on Build GRCh38
rs753391534 Chr.2: 96897876-96897876 on Build GRCh38
rs1004606160 Chr.2: 96897965-96897965 on Build GRCh38

View Full Product Details