Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159319129-159319517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

SNP Detail

SNP ID Chromosome Location
rs929870720 Chr.2: 159319250-159319250 on Build GRCh38
rs948034732 Chr.2: 159319289-159319289 on Build GRCh38
rs1047313019 Chr.2: 159319409-159319409 on Build GRCh38
rs917873297 Chr.2: 159319262-159319262 on Build GRCh38
rs899559497 Chr.2: 159319406-159319406 on Build GRCh38
rs1008522347 Chr.2: 159319451-159319451 on Build GRCh38
rs748842719 Chr.2: 159319444-159319444 on Build GRCh38
rs1021351418 Chr.2: 159319487-159319487 on Build GRCh38
rs150700748 Chr.2: 159319467-159319467 on Build GRCh38
rs898568577 Chr.2: 159319326-159319326 on Build GRCh38
rs997896878 Chr.2: 159319442-159319442 on Build GRCh38
rs935550910 Chr.2: 159319240-159319240 on Build GRCh38
rs1039386674 Chr.2: 159319401-159319401 on Build GRCh38
rs1020977826 Chr.2: 159319489-159319489 on Build GRCh38
rs888102998 Chr.2: 159319272-159319272 on Build GRCh38
rs541393596 Chr.2: 159319407-159319407 on Build GRCh38
rs917095841 Chr.2: 159319181-159319181 on Build GRCh38
rs1045210395 Chr.2: 159319302-159319302 on Build GRCh38
rs527608016 Chr.2: 159319361-159319361 on Build GRCh38
rs968216028 Chr.2: 159319171-159319171 on Build GRCh38
rs996454672 Chr.2: 159319485-159319485 on Build GRCh38
rs924233927 Chr.2: 159319189-159319189 on Build GRCh38
rs1047053378 Chr.2: 159319254-159319254 on Build GRCh38
rs1010440243 Chr.2: 159319488-159319488 on Build GRCh38
rs561344530 Chr.2: 159319459-159319459 on Build GRCh38
rs1056475478 Chr.2: 159319249-159319249 on Build GRCh38
rs963714190 Chr.2: 159319468-159319468 on Build GRCh38
rs992637260 Chr.2: 159319156-159319156 on Build GRCh38

View Full Product Details