Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 44506581-44507180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSCB
Entrez Gene ID
84075
Gene Name
fibrous sheath CABYR binding protein
Gene Aliases
C14orf155
UniGene
Hs.307086
Gene Chromosome Location
Chr.14: 44504149-44507296 on Build GRCh38
Transcripts
NM_032135.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6001383 c.128C>A p.A43D Chr.14: 44506860-44506860 on Build GRCh38
140028 c.268G>A p.E90K Chr.14: 44506720-44506720 on Build GRCh38
5591040 c.68C>T p.S23F Chr.14: 44506920-44506920 on Build GRCh38
6075811 c.206C>A p.T69N Chr.14: 44506782-44506782 on Build GRCh38
4966278 c.136T>C p.Y46H Chr.14: 44506852-44506852 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759643320 Chr.14: 44506961-44506961 on Build GRCh38
rs952273069 Chr.14: 44506974-44506974 on Build GRCh38
rs548381970 Chr.14: 44507077-44507077 on Build GRCh38
rs779123363 Chr.14: 44507014-44507014 on Build GRCh38
rs137911374 Chr.14: 44506653-44506653 on Build GRCh38
rs369591164 Chr.14: 44506627-44506627 on Build GRCh38
rs530257531 Chr.14: 44507061-44507061 on Build GRCh38
rs11629125 Chr.14: 44507038-44507038 on Build GRCh38
rs756196550 Chr.14: 44506987-44506987 on Build GRCh38
rs185783175 Chr.14: 44507048-44507048 on Build GRCh38
rs950324032 Chr.14: 44506689-44506689 on Build GRCh38
rs376297816 Chr.14: 44506726-44506726 on Build GRCh38
rs1001178044 Chr.14: 44507092-44507092 on Build GRCh38
rs564059828 Chr.14: 44506970-44506970 on Build GRCh38
rs1042053062 Chr.14: 44507068-44507068 on Build GRCh38
rs971428204 Chr.14: 44507058-44507058 on Build GRCh38
rs753738083 Chr.14: 44506760-44506760 on Build GRCh38
rs926075860 Chr.14: 44506602-44506602 on Build GRCh38
rs781615055 Chr.14: 44506778-44506778 on Build GRCh38
rs761606590 Chr.14: 44506937-44506937 on Build GRCh38
rs764939371 Chr.14: 44506938-44506938 on Build GRCh38
rs755030267 Chr.14: 44506774-44506774 on Build GRCh38
rs750293647 Chr.14: 44506949-44506949 on Build GRCh38
rs770331182 Chr.14: 44506784-44506784 on Build GRCh38
rs747098128 Chr.14: 44506640-44506640 on Build GRCh38
rs759364392 Chr.14: 44506697-44506701 on Build GRCh38
rs143704152 Chr.14: 44506714-44506714 on Build GRCh38
rs541538463 Chr.14: 44506854-44506854 on Build GRCh38
rs201109046 Chr.14: 44506761-44506761 on Build GRCh38
rs889617822 Chr.14: 44507140-44507141 on Build GRCh38
rs34414518 Chr.14: 44507080-44507081 on Build GRCh38
rs758608988 Chr.14: 44506628-44506628 on Build GRCh38
rs966012218 Chr.14: 44506915-44506915 on Build GRCh38
rs527889835 Chr.14: 44507148-44507148 on Build GRCh38
rs777029779 Chr.14: 44506663-44506663 on Build GRCh38
rs759019044 Chr.14: 44506812-44506812 on Build GRCh38
rs572138282 Chr.14: 44506929-44506929 on Build GRCh38
rs764168539 Chr.14: 44506851-44506851 on Build GRCh38
rs567888781 Chr.14: 44506914-44506914 on Build GRCh38
rs919384215 Chr.14: 44506738-44506738 on Build GRCh38
rs1049283470 Chr.14: 44506845-44506845 on Build GRCh38
rs370125808 Chr.14: 44506850-44506850 on Build GRCh38
rs556553779 Chr.14: 44507136-44507136 on Build GRCh38
rs945114807 Chr.14: 44507047-44507047 on Build GRCh38
rs757839022 Chr.14: 44507013-44507013 on Build GRCh38
rs968281598 Chr.14: 44506939-44506939 on Build GRCh38
rs751736996 Chr.14: 44506873-44506873 on Build GRCh38
rs768666313 Chr.14: 44506935-44506935 on Build GRCh38
rs748652888 Chr.14: 44506780-44506780 on Build GRCh38
rs760777002 Chr.14: 44507027-44507027 on Build GRCh38
rs762771265 Chr.14: 44506900-44506900 on Build GRCh38
rs771204342 Chr.14: 44506911-44506911 on Build GRCh38
rs930906179 Chr.14: 44506639-44506639 on Build GRCh38
rs141248167 Chr.14: 44506847-44506847 on Build GRCh38
rs267603990 Chr.14: 44506894-44506894 on Build GRCh38
rs180887451 Chr.14: 44507030-44507030 on Build GRCh38
rs548710567 Chr.14: 44506796-44506796 on Build GRCh38
rs775110991 Chr.14: 44506930-44506930 on Build GRCh38
rs917260090 Chr.14: 44506669-44506669 on Build GRCh38
rs780172171 Chr.14: 44506762-44506762 on Build GRCh38
rs770151089 Chr.14: 44506678-44506678 on Build GRCh38
rs748025761 Chr.14: 44507039-44507039 on Build GRCh38
rs150784772 Chr.14: 44506897-44506897 on Build GRCh38
rs745493677 Chr.14: 44506922-44506922 on Build GRCh38
rs866351969 Chr.14: 44507131-44507131 on Build GRCh38
rs770564938 Chr.14: 44506805-44506805 on Build GRCh38
rs774252014 Chr.14: 44507005-44507005 on Build GRCh38
rs554125229 Chr.14: 44506899-44506899 on Build GRCh38
rs868136157 Chr.14: 44506615-44506615 on Build GRCh38
rs545555565 Chr.14: 44506936-44506936 on Build GRCh38
rs36083807 Chr.14: 44506633-44506633 on Build GRCh38
rs911868742 Chr.14: 44506603-44506603 on Build GRCh38
rs755297110 Chr.14: 44506876-44506876 on Build GRCh38
rs752811535 Chr.14: 44506895-44506895 on Build GRCh38
rs771476208 Chr.14: 44506825-44506825 on Build GRCh38
rs201908847 Chr.14: 44506750-44506750 on Build GRCh38
rs761539695 Chr.14: 44506862-44506862 on Build GRCh38
rs763969899 Chr.14: 44506751-44506751 on Build GRCh38
rs958698008 Chr.14: 44506624-44506624 on Build GRCh38
rs758990775 Chr.14: 44506727-44506727 on Build GRCh38
rs752337735 Chr.14: 44507003-44507005 on Build GRCh38
rs45586531 Chr.14: 44506715-44506715 on Build GRCh38
rs374177028 Chr.14: 44507031-44507031 on Build GRCh38
rs767430118 Chr.14: 44506744-44506744 on Build GRCh38
rs374018207 Chr.14: 44506677-44506677 on Build GRCh38
rs930788989 Chr.14: 44507106-44507106 on Build GRCh38
rs752721220 Chr.14: 44506984-44506984 on Build GRCh38
rs763819335 Chr.14: 44506933-44506933 on Build GRCh38
rs753646333 Chr.14: 44506853-44506853 on Build GRCh38
rs368475663 Chr.14: 44507011-44507011 on Build GRCh38
rs148860855 Chr.14: 44506618-44506618 on Build GRCh38
rs374629243 Chr.14: 44506948-44506948 on Build GRCh38
rs746285503 Chr.14: 44507024-44507024 on Build GRCh38
rs752684975 Chr.14: 44506749-44506749 on Build GRCh38
rs552778592 Chr.14: 44507155-44507155 on Build GRCh38
rs560342635 Chr.14: 44506889-44506889 on Build GRCh38
rs377202903 Chr.14: 44506695-44506695 on Build GRCh38
rs766569606 Chr.14: 44506866-44506866 on Build GRCh38
rs765983593 Chr.14: 44506723-44506723 on Build GRCh38
rs62013252 Chr.14: 44506768-44506768 on Build GRCh38
rs773887213 Chr.14: 44506810-44506810 on Build GRCh38
rs749679703 Chr.14: 44506804-44506804 on Build GRCh38
rs926828144 Chr.14: 44506671-44506671 on Build GRCh38
rs78484213 Chr.14: 44507127-44507127 on Build GRCh38
rs767423379 Chr.14: 44506840-44506841 on Build GRCh38

View Full Product Details