Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 76173406-76173978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH2L
Entrez Gene ID
55668
Gene Name
G-patch domain containing 2 like
Gene Aliases
C14orf118
UniGene
Hs.410231
Gene Chromosome Location
Chr.14: 76151885-76214473 on Build GRCh38
Transcripts
NM_001322026.1 NM_001322027.1 NM_001322028.1 NM_001322029.1 NM_001322030.1 NM_001322032.1 NM_017926.3 NM_017972.3 NR_110314.1 XM_006720191.2 XM_017021427.1 XM_017021428.1 XM_017021429.1 XM_017021430.1 XM_017021431.1 XM_017021432.1 XM_017021433.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6673022 c.905-10_905-9insT p.? Chr.14: 76173536-76173537 on Build GRCh38
4601696 c.916C>T p.R306C Chr.14: 76173557-76173557 on Build GRCh38
4601698 c.916C>T p.R306C Chr.14: 76173557-76173557 on Build GRCh38
6673025 c.905-9_905-8delTT p.? Chr.14: 76173537-76173538 on Build GRCh38
6673043 c.905-9delT p.? Chr.14: 76173537-76173537 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752048317 Chr.14: 76173564-76173564 on Build GRCh38
rs946945767 Chr.14: 76173786-76173786 on Build GRCh38
rs777584926 Chr.14: 76173597-76173597 on Build GRCh38
rs776448859 Chr.14: 76173626-76173626 on Build GRCh38
rs377474002 Chr.14: 76173587-76173587 on Build GRCh38
rs530845364 Chr.14: 76173678-76173678 on Build GRCh38
rs778609402 Chr.14: 76173521-76173521 on Build GRCh38
rs775483043 Chr.14: 76173790-76173790 on Build GRCh38
rs549475757 Chr.14: 76173529-76173529 on Build GRCh38
rs539542110 Chr.14: 76173441-76173441 on Build GRCh38
rs760387653 Chr.14: 76173538-76173538 on Build GRCh38
rs535704138 Chr.14: 76173607-76173607 on Build GRCh38
rs962947274 Chr.14: 76173578-76173578 on Build GRCh38
rs578009938 Chr.14: 76173723-76173723 on Build GRCh38
rs778295927 Chr.14: 76173499-76173499 on Build GRCh38
rs746470289 Chr.14: 76173712-76173712 on Build GRCh38
rs35631432 Chr.14: 76173622-76173622 on Build GRCh38
rs995762696 Chr.14: 76173914-76173914 on Build GRCh38
rs747025642 Chr.14: 76173602-76173602 on Build GRCh38
rs1043954527 Chr.14: 76173788-76173788 on Build GRCh38
rs61465599 Chr.14: 76173848-76173849 on Build GRCh38
rs796895080 Chr.14: 76173951-76173951 on Build GRCh38
rs781733578 Chr.14: 76173567-76173567 on Build GRCh38
rs750836950 Chr.14: 76173682-76173682 on Build GRCh38
rs757971706 Chr.14: 76173710-76173710 on Build GRCh38
rs1045525109 Chr.14: 76173703-76173703 on Build GRCh38
rs78121241 Chr.14: 76173848-76173848 on Build GRCh38
rs759819620 Chr.14: 76173553-76173553 on Build GRCh38
rs867722201 Chr.14: 76173693-76173693 on Build GRCh38
rs776397521 Chr.14: 76173552-76173552 on Build GRCh38
rs527614879 Chr.14: 76173897-76173897 on Build GRCh38
rs759279925 Chr.14: 76173645-76173645 on Build GRCh38
rs398025756 Chr.14: 76173849-76173849 on Build GRCh38
rs770110438 Chr.14: 76173696-76173699 on Build GRCh38
rs765413540 Chr.14: 76173557-76173557 on Build GRCh38
rs36047526 Chr.14: 76173679-76173679 on Build GRCh38
rs367834139 Chr.14: 76173656-76173656 on Build GRCh38
rs555637280 Chr.14: 76173640-76173640 on Build GRCh38
rs750789629 Chr.14: 76173570-76173570 on Build GRCh38
rs752332781 Chr.14: 76173697-76173697 on Build GRCh38
rs921819895 Chr.14: 76173902-76173902 on Build GRCh38
rs183315508 Chr.14: 76173791-76173791 on Build GRCh38
rs1006686103 Chr.14: 76173853-76173853 on Build GRCh38
rs547528888 Chr.14: 76173857-76173857 on Build GRCh38
rs371327893 Chr.14: 76173667-76173667 on Build GRCh38
rs895297002 Chr.14: 76173801-76173801 on Build GRCh38
rs999203676 Chr.14: 76173488-76173488 on Build GRCh38
rs764514578 Chr.14: 76173562-76173562 on Build GRCh38
rs1004813197 Chr.14: 76173876-76173876 on Build GRCh38
rs557916468 Chr.14: 76173933-76173933 on Build GRCh38
rs770036194 Chr.14: 76173647-76173647 on Build GRCh38
rs983436027 Chr.14: 76173748-76173748 on Build GRCh38
rs1050755297 Chr.14: 76173822-76173822 on Build GRCh38
rs1055213400 Chr.14: 76173778-76173778 on Build GRCh38
rs866153515 Chr.14: 76173724-76173724 on Build GRCh38
rs769523003 Chr.14: 76173734-76173734 on Build GRCh38
rs763051565 Chr.14: 76173560-76173560 on Build GRCh38
rs762956350 Chr.14: 76173660-76173660 on Build GRCh38
rs749011474 Chr.14: 76173518-76173518 on Build GRCh38
rs569318072 Chr.14: 76173575-76173575 on Build GRCh38
rs374329052 Chr.14: 76173531-76173531 on Build GRCh38
rs955109085 Chr.14: 76173516-76173516 on Build GRCh38
rs771540003 Chr.14: 76173594-76173594 on Build GRCh38
rs957395950 Chr.14: 76173927-76173927 on Build GRCh38
rs928014131 Chr.14: 76173763-76173763 on Build GRCh38
rs746159848 Chr.14: 76173720-76173720 on Build GRCh38
rs563319475 Chr.14: 76173493-76173493 on Build GRCh38
rs778656471 Chr.14: 76173584-76173584 on Build GRCh38
rs765653180 Chr.14: 76173938-76173938 on Build GRCh38
rs150861843 Chr.14: 76173877-76173877 on Build GRCh38
rs386778945 Chr.14: 76173849-76173854 on Build GRCh38
rs756504053 Chr.14: 76173691-76173691 on Build GRCh38
rs2289836 Chr.14: 76173782-76173782 on Build GRCh38
rs747637759 Chr.14: 76173588-76173588 on Build GRCh38
rs866889375 Chr.14: 76173755-76173755 on Build GRCh38
rs371069179 Chr.14: 76173514-76173514 on Build GRCh38
rs558356363 Chr.14: 76173722-76173722 on Build GRCh38
rs539512913 Chr.14: 76173921-76173921 on Build GRCh38
rs774294010 Chr.14: 76173672-76173672 on Build GRCh38
rs531233041 Chr.14: 76173592-76173592 on Build GRCh38
rs1025537331 Chr.14: 76173925-76173925 on Build GRCh38
rs377564638 Chr.14: 76173500-76173500 on Build GRCh38
rs777724644 Chr.14: 76173444-76173444 on Build GRCh38
rs566069795 Chr.14: 76173681-76173681 on Build GRCh38
rs748685426 Chr.14: 76173644-76173644 on Build GRCh38
rs35998762 Chr.14: 76173604-76173604 on Build GRCh38
rs79425319 Chr.14: 76173849-76173849 on Build GRCh38
rs59407603 Chr.14: 76173838-76173838 on Build GRCh38
rs995107736 Chr.14: 76173815-76173815 on Build GRCh38
rs902204314 Chr.14: 76173463-76173463 on Build GRCh38
rs372967613 Chr.14: 76173494-76173494 on Build GRCh38
rs939446512 Chr.14: 76173769-76173769 on Build GRCh38
rs770654112 Chr.14: 76173540-76173540 on Build GRCh38
rs75230612 Chr.14: 76173850-76173850 on Build GRCh38
rs534888463 Chr.14: 76173692-76173692 on Build GRCh38
rs974043625 Chr.14: 76173612-76173612 on Build GRCh38
rs749098512 Chr.14: 76173517-76173517 on Build GRCh38
rs757695553 Chr.14: 76173566-76173566 on Build GRCh38
rs572862892 Chr.14: 76173727-76173727 on Build GRCh38
rs373935229 Chr.14: 76173669-76173669 on Build GRCh38
rs777407114 Chr.14: 76173711-76173711 on Build GRCh38
rs752797272 Chr.14: 76173559-76173559 on Build GRCh38

View Full Product Details