Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78590258-78590771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6850859 c.984T>C p.I328I Chr.15: 78590375-78590375 on Build GRCh38
6838857 c.920C>G p.S307* Chr.15: 78590311-78590311 on Build GRCh38
5565797 c.1100G>C p.R367T Chr.15: 78590491-78590491 on Build GRCh38
3504322 c.1239C>T p.V413V Chr.15: 78590630-78590630 on Build GRCh38
1189078 c.1075A>T p.T359S Chr.15: 78590466-78590466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759551526 Chr.15: 78590670-78590670 on Build GRCh38
rs944978839 Chr.15: 78590322-78590322 on Build GRCh38
rs200449882 Chr.15: 78590457-78590457 on Build GRCh38
rs200127699 Chr.15: 78590631-78590631 on Build GRCh38
rs200414546 Chr.15: 78590303-78590303 on Build GRCh38
rs767284826 Chr.15: 78590558-78590558 on Build GRCh38
rs370722027 Chr.15: 78590468-78590468 on Build GRCh38
rs200102110 Chr.15: 78590361-78590361 on Build GRCh38
rs757408983 Chr.15: 78590291-78590291 on Build GRCh38
rs139612394 Chr.15: 78590394-78590394 on Build GRCh38
rs780560672 Chr.15: 78590351-78590351 on Build GRCh38
rs201385812 Chr.15: 78590449-78590449 on Build GRCh38
rs753213192 Chr.15: 78590674-78590674 on Build GRCh38
rs200369775 Chr.15: 78590438-78590438 on Build GRCh38
rs769498175 Chr.15: 78590518-78590518 on Build GRCh38
rs150696868 Chr.15: 78590295-78590295 on Build GRCh38
rs949514581 Chr.15: 78590743-78590743 on Build GRCh38
rs543578175 Chr.15: 78590546-78590546 on Build GRCh38
rs768162469 Chr.15: 78590612-78590612 on Build GRCh38
rs867013198 Chr.15: 78590383-78590383 on Build GRCh38
rs750645471 Chr.15: 78590561-78590561 on Build GRCh38
rs771827809 Chr.15: 78590632-78590632 on Build GRCh38
rs151003606 Chr.15: 78590579-78590579 on Build GRCh38
rs781009027 Chr.15: 78590507-78590507 on Build GRCh38
rs748922437 Chr.15: 78590402-78590404 on Build GRCh38
rs757107988 Chr.15: 78590457-78590458 on Build GRCh38
rs201775979 Chr.15: 78590589-78590589 on Build GRCh38
rs760933898 Chr.15: 78590657-78590657 on Build GRCh38
rs770904584 Chr.15: 78590750-78590750 on Build GRCh38
rs957002666 Chr.15: 78590698-78590698 on Build GRCh38
rs769011950 Chr.15: 78590376-78590376 on Build GRCh38
rs762980943 Chr.15: 78590410-78590410 on Build GRCh38
rs767663657 Chr.15: 78590439-78590439 on Build GRCh38
rs200010345 Chr.15: 78590287-78590287 on Build GRCh38
rs375168271 Chr.15: 78590306-78590306 on Build GRCh38
rs76766434 Chr.15: 78590592-78590592 on Build GRCh38
rs201720688 Chr.15: 78590540-78590540 on Build GRCh38
rs745918259 Chr.15: 78590305-78590305 on Build GRCh38
rs201916656 Chr.15: 78590529-78590529 on Build GRCh38
rs772437201 Chr.15: 78590502-78590502 on Build GRCh38
rs74865777 Chr.15: 78590352-78590352 on Build GRCh38
rs116099178 Chr.15: 78590340-78590340 on Build GRCh38
rs554878059 Chr.15: 78590387-78590387 on Build GRCh38
rs79721430 Chr.15: 78590467-78590467 on Build GRCh38
rs141180754 Chr.15: 78590593-78590593 on Build GRCh38
rs867769219 Chr.15: 78590630-78590630 on Build GRCh38
rs201995384 Chr.15: 78590455-78590455 on Build GRCh38
rs185109067 Chr.15: 78590729-78590729 on Build GRCh38
rs780040957 Chr.15: 78590454-78590454 on Build GRCh38
rs768325287 Chr.15: 78590586-78590586 on Build GRCh38
rs372560481 Chr.15: 78590414-78590414 on Build GRCh38
rs778281973 Chr.15: 78590587-78590587 on Build GRCh38
rs72648879 Chr.15: 78590744-78590744 on Build GRCh38
rs144306535 Chr.15: 78590432-78590432 on Build GRCh38
rs1042500 Chr.15: 78590605-78590605 on Build GRCh38
rs778735334 Chr.15: 78590518-78590521 on Build GRCh38
rs79518498 Chr.15: 78590289-78590289 on Build GRCh38
rs977099173 Chr.15: 78590568-78590568 on Build GRCh38
rs918008475 Chr.15: 78590711-78590711 on Build GRCh38
rs201569673 Chr.15: 78590328-78590328 on Build GRCh38
rs1018181118 Chr.15: 78590595-78590595 on Build GRCh38
rs558752984 Chr.15: 78590437-78590437 on Build GRCh38
rs867154905 Chr.15: 78590296-78590296 on Build GRCh38
rs772975768 Chr.15: 78590642-78590642 on Build GRCh38
rs964002776 Chr.15: 78590427-78590427 on Build GRCh38
rs200627083 Chr.15: 78590483-78590483 on Build GRCh38
rs763035811 Chr.15: 78590533-78590533 on Build GRCh38
rs79109919 Chr.15: 78590479-78590479 on Build GRCh38
rs146801604 Chr.15: 78590390-78590390 on Build GRCh38
rs745841148 Chr.15: 78590595-78590599 on Build GRCh38
rs140785516 Chr.15: 78590564-78590564 on Build GRCh38
rs748780333 Chr.15: 78590594-78590594 on Build GRCh38
rs759142711 Chr.15: 78590392-78590392 on Build GRCh38
rs201117838 Chr.15: 78590658-78590658 on Build GRCh38
rs745450184 Chr.15: 78590513-78590513 on Build GRCh38
rs373336975 Chr.15: 78590638-78590638 on Build GRCh38
rs776310455 Chr.15: 78590391-78590391 on Build GRCh38
rs200644647 Chr.15: 78590567-78590567 on Build GRCh38
rs61740655 Chr.15: 78590384-78590384 on Build GRCh38
rs150108684 Chr.15: 78590582-78590582 on Build GRCh38
rs202178144 Chr.15: 78590626-78590626 on Build GRCh38
rs16969968 Chr.15: 78590583-78590583 on Build GRCh38
rs775575449 Chr.15: 78590400-78590400 on Build GRCh38
rs202070829 Chr.15: 78590418-78590418 on Build GRCh38
rs143586773 Chr.15: 78590578-78590578 on Build GRCh38
rs776789728 Chr.15: 78590665-78590665 on Build GRCh38
rs979829227 Chr.15: 78590654-78590654 on Build GRCh38
rs1036440224 Chr.15: 78590692-78590692 on Build GRCh38
rs565162745 Chr.15: 78590695-78590695 on Build GRCh38
rs369392765 Chr.15: 78590673-78590673 on Build GRCh38
rs988276785 Chr.15: 78590708-78590708 on Build GRCh38
rs200946954 Chr.15: 78590379-78590379 on Build GRCh38
rs547608751 Chr.15: 78590665-78590667 on Build GRCh38
rs868388135 Chr.15: 78590338-78590338 on Build GRCh38
rs201259246 Chr.15: 78590385-78590385 on Build GRCh38
rs570647862 Chr.15: 78590448-78590448 on Build GRCh38

View Full Product Details