Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 83891226-83891600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTSL3
Entrez Gene ID
57188
Gene Name
ADAMTS like 3
Gene Aliases
ADAMTSL-3
UniGene
Hs.459162
Gene Chromosome Location
Chr.15: 83654086-84039842 on Build GRCh38
Transcripts
NM_001301110.1 NM_207517.2 XM_011521822.2 XM_011521823.2 XM_011521824.2 XM_011521825.2 XM_017022434.1 XM_017022435.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7087482 c.1258C>A p.P420T Chr.15: 83891375-83891375 on Build GRCh38
7082159 c.1248C>T p.F416F Chr.15: 83891365-83891365 on Build GRCh38
216354 c.1243C>A p.H415N Chr.15: 83891360-83891360 on Build GRCh38
272313 c.1262G>A p.R421H Chr.15: 83891379-83891379 on Build GRCh38
7087483 c.1258C>A p.P420T Chr.15: 83891375-83891375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1003421784 Chr.15: 83891287-83891287 on Build GRCh38
rs772258043 Chr.15: 83891370-83891370 on Build GRCh38
rs780859128 Chr.15: 83891325-83891325 on Build GRCh38
rs987382523 Chr.15: 83891554-83891554 on Build GRCh38
rs773594221 Chr.15: 83891378-83891378 on Build GRCh38
rs770165561 Chr.15: 83891344-83891344 on Build GRCh38
rs1044875684 Chr.15: 83891273-83891276 on Build GRCh38
rs573005080 Chr.15: 83891487-83891487 on Build GRCh38
rs200042144 Chr.15: 83891313-83891313 on Build GRCh38
rs1034353813 Chr.15: 83891477-83891477 on Build GRCh38
rs115111662 Chr.15: 83891305-83891305 on Build GRCh38
rs375927670 Chr.15: 83891387-83891387 on Build GRCh38
rs185235494 Chr.15: 83891452-83891452 on Build GRCh38
rs549043745 Chr.15: 83891265-83891265 on Build GRCh38
rs141215018 Chr.15: 83891353-83891353 on Build GRCh38
rs764154681 Chr.15: 83891430-83891430 on Build GRCh38
rs1051327311 Chr.15: 83891266-83891266 on Build GRCh38
rs535493447 Chr.15: 83891259-83891259 on Build GRCh38
rs951539063 Chr.15: 83891519-83891519 on Build GRCh38
rs144512205 Chr.15: 83891453-83891453 on Build GRCh38
rs761445175 Chr.15: 83891379-83891379 on Build GRCh38
rs1018406457 Chr.15: 83891448-83891448 on Build GRCh38
rs762638995 Chr.15: 83891429-83891429 on Build GRCh38
rs150330504 Chr.15: 83891360-83891360 on Build GRCh38
rs767078132 Chr.15: 83891532-83891532 on Build GRCh38
rs573090819 Chr.15: 83891330-83891330 on Build GRCh38
rs756714987 Chr.15: 83891317-83891317 on Build GRCh38
rs912804664 Chr.15: 83891566-83891566 on Build GRCh38
rs775300698 Chr.15: 83891417-83891417 on Build GRCh38
rs867148343 Chr.15: 83891333-83891333 on Build GRCh38
rs541918283 Chr.15: 83891579-83891579 on Build GRCh38
rs929635296 Chr.15: 83891255-83891259 on Build GRCh38
rs922852290 Chr.15: 83891463-83891463 on Build GRCh38
rs557744424 Chr.15: 83891348-83891348 on Build GRCh38
rs368431268 Chr.15: 83891497-83891497 on Build GRCh38
rs765079834 Chr.15: 83891280-83891280 on Build GRCh38
rs769264280 Chr.15: 83891357-83891357 on Build GRCh38
rs1001483967 Chr.15: 83891458-83891458 on Build GRCh38
rs1039420571 Chr.15: 83891307-83891307 on Build GRCh38
rs891374308 Chr.15: 83891277-83891277 on Build GRCh38
rs111728418 Chr.15: 83891553-83891553 on Build GRCh38
rs758291082 Chr.15: 83891298-83891298 on Build GRCh38
rs202075227 Chr.15: 83891368-83891368 on Build GRCh38
rs962753715 Chr.15: 83891483-83891483 on Build GRCh38
rs941484269 Chr.15: 83891250-83891250 on Build GRCh38
rs918790633 Chr.15: 83891261-83891261 on Build GRCh38
rs745493564 Chr.15: 83891339-83891339 on Build GRCh38
rs752612140 Chr.15: 83891296-83891296 on Build GRCh38
rs918682767 Chr.15: 83891508-83891508 on Build GRCh38

View Full Product Details